Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 57652029-57652626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B1
Entrez Gene ID
5205
Gene Name
ATPase phospholipid transporting 8B1
Gene Aliases
ATPIC, BRIC, FIC1, ICP1, PFIC, PFIC1
UniGene
Hs.216623
Gene Chromosome Location
Chr.18: 57646426-57803828 on Build GRCh38
Transcripts
NM_005603.4 XM_006722481.3 XM_011526022.2 XM_011526023.2 NM_001242804.1

Gene Symbol
LOC100505549
Entrez Gene ID
100505549
Gene Name
uncharacterized LOC100505549
Gene Aliases
-
UniGene
Hs.660645
Gene Chromosome Location
Chr.18: 57630302-57669276 on Build GRCh38
Transcripts
NM_005603.4 XM_006722481.3 XM_011526022.2 XM_011526023.2 NM_001242804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6149328 c.3144C>T p.F1048F Chr.18: 57652601-57652601 on Build GRCh38
5612010 c.3244A>G p.I1082V Chr.18: 57652501-57652501 on Build GRCh38
4817264 c.3305C>A p.S1102* Chr.18: 57652129-57652129 on Build GRCh38
3796462 c.3141C>G p.L1047L Chr.18: 57652604-57652604 on Build GRCh38
3787520 c.3266G>A p.G1089D Chr.18: 57652168-57652168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776611415 Chr.18: 57652221-57652221 on Build GRCh38
rs937398042 Chr.18: 57652226-57652226 on Build GRCh38
rs112868885 Chr.18: 57652074-57652074 on Build GRCh38
rs766292913 Chr.18: 57652121-57652121 on Build GRCh38
rs753887767 Chr.18: 57652436-57652436 on Build GRCh38
rs968352318 Chr.18: 57652366-57652366 on Build GRCh38
rs901687134 Chr.18: 57652345-57652345 on Build GRCh38
rs746384145 Chr.18: 57652152-57652152 on Build GRCh38
rs79391613 Chr.18: 57652150-57652150 on Build GRCh38
rs754259464 Chr.18: 57652505-57652505 on Build GRCh38
rs34547890 Chr.18: 57652253-57652254 on Build GRCh38
rs917293206 Chr.18: 57652443-57652443 on Build GRCh38
rs754583409 Chr.18: 57652525-57652525 on Build GRCh38
rs965126957 Chr.18: 57652378-57652378 on Build GRCh38
rs756419398 Chr.18: 57652153-57652153 on Build GRCh38
rs771488259 Chr.18: 57652204-57652204 on Build GRCh38
rs758728677 Chr.18: 57652127-57652127 on Build GRCh38
rs1033007908 Chr.18: 57652430-57652430 on Build GRCh38
rs11875799 Chr.18: 57652328-57652328 on Build GRCh38
rs17759233 Chr.18: 57652212-57652212 on Build GRCh38
rs183371188 Chr.18: 57652501-57652501 on Build GRCh38
rs756652647 Chr.18: 57652479-57652479 on Build GRCh38
rs761969261 Chr.18: 57652578-57652578 on Build GRCh38
rs367988012 Chr.18: 57652581-57652581 on Build GRCh38
rs200614310 Chr.18: 57652463-57652463 on Build GRCh38
rs769485522 Chr.18: 57652570-57652570 on Build GRCh38
rs768112395 Chr.18: 57652056-57652056 on Build GRCh38
rs767548754 Chr.18: 57652111-57652111 on Build GRCh38
rs996386509 Chr.18: 57652355-57652355 on Build GRCh38
rs778273036 Chr.18: 57652200-57652200 on Build GRCh38
rs750888321 Chr.18: 57652464-57652464 on Build GRCh38
rs1044664913 Chr.18: 57652080-57652080 on Build GRCh38
rs761915146 Chr.18: 57652466-57652466 on Build GRCh38
rs377048061 Chr.18: 57652246-57652246 on Build GRCh38
rs771044045 Chr.18: 57652549-57652549 on Build GRCh38
rs767601611 Chr.18: 57652580-57652580 on Build GRCh38
rs886109835 Chr.18: 57652585-57652585 on Build GRCh38
rs747760731 Chr.18: 57652201-57652201 on Build GRCh38
rs139083067 Chr.18: 57652590-57652590 on Build GRCh38
rs564218494 Chr.18: 57652367-57652367 on Build GRCh38
rs531722791 Chr.18: 57652419-57652419 on Build GRCh38
rs746914580 Chr.18: 57652546-57652546 on Build GRCh38
rs138594092 Chr.18: 57652359-57652359 on Build GRCh38
rs994395369 Chr.18: 57652289-57652289 on Build GRCh38
rs577380060 Chr.18: 57652336-57652336 on Build GRCh38
rs896966503 Chr.18: 57652281-57652281 on Build GRCh38
rs895912676 Chr.18: 57652247-57652247 on Build GRCh38
rs771564318 Chr.18: 57652541-57652541 on Build GRCh38
rs950099457 Chr.18: 57652260-57652260 on Build GRCh38
rs1036657695 Chr.18: 57652283-57652283 on Build GRCh38
rs370876900 Chr.18: 57652594-57652594 on Build GRCh38
rs761633819 Chr.18: 57652093-57652093 on Build GRCh38
rs114260754 Chr.18: 57652309-57652309 on Build GRCh38
rs773951157 Chr.18: 57652076-57652076 on Build GRCh38
rs752920462 Chr.18: 57652126-57652126 on Build GRCh38
rs1020476911 Chr.18: 57652417-57652417 on Build GRCh38
rs540921535 Chr.18: 57652310-57652310 on Build GRCh38
rs988899891 Chr.18: 57652441-57652441 on Build GRCh38
rs777459332 Chr.18: 57652544-57652544 on Build GRCh38
rs1046235167 Chr.18: 57652273-57652273 on Build GRCh38
rs866362278 Chr.18: 57652537-57652537 on Build GRCh38
rs768403395 Chr.18: 57652167-57652167 on Build GRCh38
rs140005450 Chr.18: 57652116-57652116 on Build GRCh38
rs111931116 Chr.18: 57652456-57652456 on Build GRCh38
rs75586146 Chr.18: 57652164-57652164 on Build GRCh38
rs190425045 Chr.18: 57652222-57652222 on Build GRCh38
rs780425796 Chr.18: 57652159-57652159 on Build GRCh38
rs955982898 Chr.18: 57652435-57652435 on Build GRCh38
rs757695758 Chr.18: 57652142-57652142 on Build GRCh38
rs146782236 Chr.18: 57652584-57652584 on Build GRCh38
rs7226404 Chr.18: 57652240-57652240 on Build GRCh38
rs759303818 Chr.18: 57652565-57652565 on Build GRCh38
rs906084726 Chr.18: 57652123-57652123 on Build GRCh38
rs146191639 Chr.18: 57652545-57652545 on Build GRCh38
rs991453254 Chr.18: 57652543-57652543 on Build GRCh38
rs773145325 Chr.18: 57652209-57652209 on Build GRCh38
rs144084458 Chr.18: 57652454-57652454 on Build GRCh38
rs761245441 Chr.18: 57652461-57652461 on Build GRCh38
rs774580560 Chr.18: 57652571-57652571 on Build GRCh38
rs751713854 Chr.18: 57652141-57652141 on Build GRCh38
rs926260260 Chr.18: 57652496-57652496 on Build GRCh38
rs764440498 Chr.18: 57652450-57652450 on Build GRCh38
rs199499992 Chr.18: 57652526-57652526 on Build GRCh38
rs776558169 Chr.18: 57652550-57652550 on Build GRCh38
rs149423934 Chr.18: 57652134-57652134 on Build GRCh38
rs569790496 Chr.18: 57652218-57652218 on Build GRCh38
rs976473416 Chr.18: 57652415-57652415 on Build GRCh38
rs772879356 Chr.18: 57652112-57652112 on Build GRCh38
rs866794984 Chr.18: 57652163-57652164 on Build GRCh38

View Full Product Details