Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46608841-46609415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM3
Entrez Gene ID
808
Gene Name
calmodulin 3
Gene Aliases
HEL-S-72, PHKD, PHKD3
UniGene
Hs.515487
Gene Chromosome Location
Chr.19: 46601255-46610782 on Build GRCh38
Transcripts
NM_005184.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6003976 c.294T>G p.N98K Chr.19: 46608854-46608854 on Build GRCh38
6151436 c.348G>A p.K116K Chr.19: 46608908-46608908 on Build GRCh38
6539216 c.303C>G p.I101M Chr.19: 46608863-46608863 on Build GRCh38
4435102 c.320G>A p.R107H Chr.19: 46608880-46608880 on Build GRCh38
998446 c.393C>T p.I131I Chr.19: 46608953-46608953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745665173 Chr.19: 46609138-46609138 on Build GRCh38
rs758692040 Chr.19: 46608923-46608923 on Build GRCh38
rs370437245 Chr.19: 46609205-46609205 on Build GRCh38
rs368104478 Chr.19: 46609172-46609172 on Build GRCh38
rs376034110 Chr.19: 46609074-46609074 on Build GRCh38
rs765760249 Chr.19: 46609075-46609075 on Build GRCh38
rs754643698 Chr.19: 46609196-46609196 on Build GRCh38
rs373393026 Chr.19: 46609100-46609100 on Build GRCh38
rs1021714998 Chr.19: 46609049-46609049 on Build GRCh38
rs771635378 Chr.19: 46609009-46609009 on Build GRCh38
rs757183576 Chr.19: 46609105-46609105 on Build GRCh38
rs778763138 Chr.19: 46609107-46609107 on Build GRCh38
rs143416579 Chr.19: 46609207-46609207 on Build GRCh38
rs928586014 Chr.19: 46609340-46609340 on Build GRCh38
rs369762540 Chr.19: 46609082-46609082 on Build GRCh38
rs763914591 Chr.19: 46609194-46609194 on Build GRCh38
rs750712140 Chr.19: 46608914-46608914 on Build GRCh38
rs2229577 Chr.19: 46609174-46609174 on Build GRCh38
rs879043246 Chr.19: 46608988-46608988 on Build GRCh38
rs938513010 Chr.19: 46608968-46608968 on Build GRCh38
rs750311120 Chr.19: 46609180-46609180 on Build GRCh38
rs751768955 Chr.19: 46609090-46609090 on Build GRCh38
rs182901112 Chr.19: 46609024-46609024 on Build GRCh38
rs1049107500 Chr.19: 46608885-46608885 on Build GRCh38
rs1007562909 Chr.19: 46609212-46609212 on Build GRCh38
rs1056801056 Chr.19: 46609373-46609373 on Build GRCh38
rs1010013527 Chr.19: 46609016-46609016 on Build GRCh38
rs528957115 Chr.19: 46609210-46609210 on Build GRCh38
rs35617141 Chr.19: 46608950-46608950 on Build GRCh38
rs1021871838 Chr.19: 46609157-46609157 on Build GRCh38
rs775140748 Chr.19: 46609023-46609023 on Build GRCh38
rs751608096 Chr.19: 46608953-46608953 on Build GRCh38
rs369796184 Chr.19: 46608995-46608995 on Build GRCh38
rs372521150 Chr.19: 46609070-46609070 on Build GRCh38
rs879168639 Chr.19: 46608989-46608989 on Build GRCh38
rs370101432 Chr.19: 46609126-46609126 on Build GRCh38
rs969737189 Chr.19: 46609261-46609261 on Build GRCh38
rs760289559 Chr.19: 46609181-46609181 on Build GRCh38
rs758987936 Chr.19: 46609213-46609214 on Build GRCh38
rs370109570 Chr.19: 46609097-46609097 on Build GRCh38
rs746703455 Chr.19: 46609183-46609183 on Build GRCh38
rs10113 Chr.19: 46609391-46609391 on Build GRCh38
rs887745567 Chr.19: 46608896-46608896 on Build GRCh38
rs78240569 Chr.19: 46609059-46609059 on Build GRCh38
rs371492420 Chr.19: 46609203-46609203 on Build GRCh38
rs1002168064 Chr.19: 46609357-46609357 on Build GRCh38
rs745577081 Chr.19: 46608999-46608999 on Build GRCh38
rs113567530 Chr.19: 46609122-46609122 on Build GRCh38
rs780937181 Chr.19: 46609200-46609200 on Build GRCh38
rs771860813 Chr.19: 46609139-46609139 on Build GRCh38
rs373639939 Chr.19: 46609158-46609158 on Build GRCh38
rs767653657 Chr.19: 46609173-46609173 on Build GRCh38
rs764512871 Chr.19: 46608892-46608892 on Build GRCh38
rs186233475 Chr.19: 46609217-46609217 on Build GRCh38
rs772262461 Chr.19: 46609156-46609157 on Build GRCh38
rs748321909 Chr.19: 46609101-46609104 on Build GRCh38
rs747668761 Chr.19: 46609201-46609201 on Build GRCh38
rs147880865 Chr.19: 46608867-46608867 on Build GRCh38
rs779037661 Chr.19: 46609100-46609101 on Build GRCh38
rs745378856 Chr.19: 46609004-46609004 on Build GRCh38
rs376694389 Chr.19: 46608986-46608986 on Build GRCh38
rs902082630 Chr.19: 46609219-46609219 on Build GRCh38
rs370746699 Chr.19: 46609235-46609235 on Build GRCh38
rs879235161 Chr.19: 46608984-46608984 on Build GRCh38
rs766718548 Chr.19: 46609171-46609171 on Build GRCh38
rs779594484 Chr.19: 46609147-46609147 on Build GRCh38
rs538139194 Chr.19: 46609242-46609242 on Build GRCh38
rs533341328 Chr.19: 46609204-46609204 on Build GRCh38
rs763262846 Chr.19: 46609170-46609170 on Build GRCh38
rs45587137 Chr.19: 46608884-46608884 on Build GRCh38
rs762466010 Chr.19: 46609162-46609162 on Build GRCh38
rs1026461427 Chr.19: 46609236-46609236 on Build GRCh38
rs770764196 Chr.19: 46608987-46608987 on Build GRCh38
rs566425717 Chr.19: 46609209-46609209 on Build GRCh38
rs746647108 Chr.19: 46609150-46609150 on Build GRCh38
rs1044212599 Chr.19: 46608944-46608944 on Build GRCh38
rs773943649 Chr.19: 46608878-46608878 on Build GRCh38
rs143699789 Chr.19: 46608899-46608899 on Build GRCh38
rs750808046 Chr.19: 46609078-46609078 on Build GRCh38
rs959956569 Chr.19: 46609353-46609353 on Build GRCh38
rs1039326977 Chr.19: 46609214-46609214 on Build GRCh38
rs1043729277 Chr.19: 46609301-46609301 on Build GRCh38
rs202189304 Chr.19: 46608902-46608902 on Build GRCh38
rs767096742 Chr.19: 46608870-46608870 on Build GRCh38
rs200095251 Chr.19: 46608869-46608869 on Build GRCh38
rs373139363 Chr.19: 46609028-46609028 on Build GRCh38
rs111606037 Chr.19: 46608982-46608982 on Build GRCh38
rs375625847 Chr.19: 46609376-46609376 on Build GRCh38
rs375150686 Chr.19: 46609260-46609260 on Build GRCh38
rs776273060 Chr.19: 46609159-46609159 on Build GRCh38
rs773792527 Chr.19: 46609163-46609163 on Build GRCh38
rs768160180 Chr.19: 46609025-46609025 on Build GRCh38
rs546364141 Chr.19: 46609051-46609051 on Build GRCh38
rs146819515 Chr.19: 46608866-46608866 on Build GRCh38
rs373845333 Chr.19: 46609206-46609206 on Build GRCh38
rs979722071 Chr.19: 46609313-46609313 on Build GRCh38
rs879785123 Chr.19: 46609355-46609355 on Build GRCh38
rs373529765 Chr.19: 46608985-46608985 on Build GRCh38
rs1007778611 Chr.19: 46609067-46609067 on Build GRCh38
rs138439676 Chr.19: 46609087-46609087 on Build GRCh38
rs972571879 Chr.19: 46609079-46609079 on Build GRCh38
rs750435544 Chr.19: 46609305-46609305 on Build GRCh38
rs200365925 Chr.19: 46609101-46609102 on Build GRCh38
rs755022859 Chr.19: 46608962-46608962 on Build GRCh38
rs1005936586 Chr.19: 46609247-46609247 on Build GRCh38
rs758159285 Chr.19: 46609157-46609157 on Build GRCh38
rs371974172 Chr.19: 46609091-46609091 on Build GRCh38

View Full Product Details