Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 107860678-107861204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL17
Entrez Gene ID
64839
Gene Name
F-box and leucine rich repeat protein 17
Gene Aliases
FBXO13, Fbl17, Fbx13
UniGene
Hs.657225
Gene Chromosome Location
Chr.5: 107859033-108382098 on Build GRCh38
Transcripts
NM_001163315.2

SNP Detail

SNP ID Chromosome Location
rs772544158 Chr.5: 107861079-107861079 on Build GRCh38
rs150249233 Chr.5: 107861138-107861138 on Build GRCh38
rs1004686994 Chr.5: 107860753-107860753 on Build GRCh38
rs1057058907 Chr.5: 107861046-107861046 on Build GRCh38
rs929497372 Chr.5: 107861029-107861029 on Build GRCh38
rs948824215 Chr.5: 107861061-107861061 on Build GRCh38
rs574445530 Chr.5: 107860725-107860725 on Build GRCh38
rs891705601 Chr.5: 107861116-107861116 on Build GRCh38
rs1029142218 Chr.5: 107861130-107861130 on Build GRCh38
rs909545198 Chr.5: 107860858-107860858 on Build GRCh38
rs953923788 Chr.5: 107860843-107860843 on Build GRCh38
rs1020262764 Chr.5: 107860883-107860883 on Build GRCh38
rs78138161 Chr.5: 107860778-107860778 on Build GRCh38
rs1019875467 Chr.5: 107861167-107861167 on Build GRCh38
rs760261557 Chr.5: 107861101-107861101 on Build GRCh38
rs540240902 Chr.5: 107860749-107860749 on Build GRCh38
rs886320711 Chr.5: 107861067-107861067 on Build GRCh38
rs938639666 Chr.5: 107860712-107860712 on Build GRCh38
rs184178364 Chr.5: 107861052-107861052 on Build GRCh38
rs1044458318 Chr.5: 107861066-107861066 on Build GRCh38
rs879901084 Chr.5: 107860928-107860928 on Build GRCh38
rs553814413 Chr.5: 107861044-107861044 on Build GRCh38
rs6873336 Chr.5: 107860821-107860821 on Build GRCh38
rs954514522 Chr.5: 107860853-107860853 on Build GRCh38
rs988907346 Chr.5: 107860929-107860929 on Build GRCh38
rs914662854 Chr.5: 107860938-107860938 on Build GRCh38
rs938656241 Chr.5: 107861039-107861039 on Build GRCh38
rs905858768 Chr.5: 107860708-107860708 on Build GRCh38
rs751515126 Chr.5: 107860768-107860768 on Build GRCh38
rs757248885 Chr.5: 107860790-107860790 on Build GRCh38
rs777093314 Chr.5: 107860758-107860758 on Build GRCh38
rs757951682 Chr.5: 107860718-107860718 on Build GRCh38
rs895946272 Chr.5: 107861058-107861058 on Build GRCh38
rs765921999 Chr.5: 107861031-107861031 on Build GRCh38
rs956247876 Chr.5: 107860900-107860900 on Build GRCh38
rs771779793 Chr.5: 107860777-107860777 on Build GRCh38
rs531444013 Chr.5: 107861180-107861180 on Build GRCh38
rs886290384 Chr.5: 107860737-107860737 on Build GRCh38
rs1029447158 Chr.5: 107860832-107860832 on Build GRCh38
rs576944235 Chr.5: 107861070-107861070 on Build GRCh38
rs919498440 Chr.5: 107861014-107861014 on Build GRCh38
rs112826035 Chr.5: 107861148-107861148 on Build GRCh38
rs980140910 Chr.5: 107860988-107860988 on Build GRCh38
rs141394549 Chr.5: 107861108-107861108 on Build GRCh38
rs898989787 Chr.5: 107861094-107861094 on Build GRCh38
rs987095059 Chr.5: 107860876-107860876 on Build GRCh38
rs890147533 Chr.5: 107861139-107861139 on Build GRCh38
rs868590127 Chr.5: 107860731-107860731 on Build GRCh38
rs973676052 Chr.5: 107860947-107860947 on Build GRCh38
rs138909446 Chr.5: 107861181-107861181 on Build GRCh38
rs1057411967 Chr.5: 107860726-107860726 on Build GRCh38
rs927338085 Chr.5: 107861024-107861024 on Build GRCh38
rs947515856 Chr.5: 107860958-107860958 on Build GRCh38

View Full Product Details