Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107504907-107505421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH8
Entrez Gene ID
91801
Gene Name
alkB homolog 8, tRNA methyltransferase
Gene Aliases
ABH8, TRM9, TRMT9
UniGene
Hs.503763
Gene Chromosome Location
Chr.11: 107502726-107565740 on Build GRCh38
Transcripts
NM_001301010.1 NM_138775.2 XM_005271728.2 XM_005271729.3 XM_017018553.1 XM_017018554.1 XM_017018555.1 XM_017018556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6198396 c.1462G>A p.E488K Chr.11: 107505191-107505191 on Build GRCh38
2109907 c.1673C>G p.P558R Chr.11: 107504980-107504980 on Build GRCh38
4388142 c.1513G>A p.A505T Chr.11: 107505140-107505140 on Build GRCh38
922596 c.1488T>G p.G496G Chr.11: 107505165-107505165 on Build GRCh38
7355233 c.1552C>T p.L518F Chr.11: 107505101-107505101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547745993 Chr.11: 107505238-107505238 on Build GRCh38
rs928794967 Chr.11: 107505063-107505063 on Build GRCh38
rs563635396 Chr.11: 107505117-107505117 on Build GRCh38
rs188662625 Chr.11: 107504953-107504953 on Build GRCh38
rs577135985 Chr.11: 107504978-107504978 on Build GRCh38
rs865927357 Chr.11: 107505009-107505009 on Build GRCh38
rs773207996 Chr.11: 107505177-107505177 on Build GRCh38
rs970208973 Chr.11: 107505375-107505375 on Build GRCh38
rs771898321 Chr.11: 107505109-107505109 on Build GRCh38
rs202023150 Chr.11: 107504991-107504991 on Build GRCh38
rs1031492235 Chr.11: 107504982-107504982 on Build GRCh38
rs776510729 Chr.11: 107505147-107505147 on Build GRCh38
rs541296766 Chr.11: 107504980-107504980 on Build GRCh38
rs768954087 Chr.11: 107504944-107504944 on Build GRCh38
rs778320324 Chr.11: 107505219-107505219 on Build GRCh38
rs553235868 Chr.11: 107504981-107504981 on Build GRCh38
rs890920186 Chr.11: 107505218-107505218 on Build GRCh38
rs375189195 Chr.11: 107505002-107505002 on Build GRCh38
rs755755364 Chr.11: 107505238-107505239 on Build GRCh38
rs73551686 Chr.11: 107505318-107505318 on Build GRCh38
rs765246505 Chr.11: 107504975-107504975 on Build GRCh38
rs532327456 Chr.11: 107505211-107505211 on Build GRCh38
rs543270040 Chr.11: 107505182-107505182 on Build GRCh38
rs767887255 Chr.11: 107505199-107505199 on Build GRCh38
rs184105011 Chr.11: 107505241-107505241 on Build GRCh38
rs961383680 Chr.11: 107505286-107505286 on Build GRCh38
rs769733031 Chr.11: 107505167-107505167 on Build GRCh38
rs921968873 Chr.11: 107505016-107505016 on Build GRCh38
rs537250015 Chr.11: 107504960-107504960 on Build GRCh38
rs139390837 Chr.11: 107504977-107504977 on Build GRCh38
rs368075378 Chr.11: 107505006-107505006 on Build GRCh38
rs370393432 Chr.11: 107504994-107504994 on Build GRCh38
rs866520413 Chr.11: 107505131-107505131 on Build GRCh38
rs756590462 Chr.11: 107505058-107505058 on Build GRCh38
rs188908701 Chr.11: 107505359-107505359 on Build GRCh38
rs1015496533 Chr.11: 107505260-107505260 on Build GRCh38
rs1011908909 Chr.11: 107505240-107505240 on Build GRCh38
rs145683517 Chr.11: 107505249-107505249 on Build GRCh38
rs562387234 Chr.11: 107505239-107505240 on Build GRCh38
rs777499341 Chr.11: 107505095-107505095 on Build GRCh38
rs371673860 Chr.11: 107505350-107505350 on Build GRCh38
rs761680732 Chr.11: 107505153-107505153 on Build GRCh38
rs753153965 Chr.11: 107505007-107505007 on Build GRCh38
rs530996811 Chr.11: 107505125-107505125 on Build GRCh38
rs570746407 Chr.11: 107505367-107505367 on Build GRCh38
rs1043358733 Chr.11: 107505138-107505138 on Build GRCh38
rs61748874 Chr.11: 107504941-107504941 on Build GRCh38
rs751780444 Chr.11: 107505187-107505187 on Build GRCh38
rs756617052 Chr.11: 107505213-107505213 on Build GRCh38
rs553976674 Chr.11: 107505036-107505036 on Build GRCh38
rs752172434 Chr.11: 107505221-107505222 on Build GRCh38
rs763847489 Chr.11: 107505240-107505241 on Build GRCh38
rs948804281 Chr.11: 107504995-107504995 on Build GRCh38
rs989864551 Chr.11: 107505011-107505011 on Build GRCh38
rs548119655 Chr.11: 107505302-107505302 on Build GRCh38
rs940830316 Chr.11: 107505163-107505163 on Build GRCh38
rs199521158 Chr.11: 107505073-107505073 on Build GRCh38
rs753342751 Chr.11: 107505106-107505106 on Build GRCh38
rs193041881 Chr.11: 107505101-107505101 on Build GRCh38
rs759693036 Chr.11: 107505193-107505193 on Build GRCh38
rs779839216 Chr.11: 107505115-107505115 on Build GRCh38
rs569659333 Chr.11: 107505315-107505315 on Build GRCh38
rs996997657 Chr.11: 107505181-107505181 on Build GRCh38
rs200270363 Chr.11: 107505269-107505269 on Build GRCh38
rs565048041 Chr.11: 107505185-107505185 on Build GRCh38
rs931093569 Chr.11: 107505212-107505212 on Build GRCh38
rs1030852356 Chr.11: 107505304-107505304 on Build GRCh38

View Full Product Details