Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54537434-54537971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGRRF1
Entrez Gene ID
10668
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
CGR19, RNF197
UniGene
Hs.59106
Gene Chromosome Location
Chr.14: 54509869-54539310 on Build GRCh38
Transcripts
NM_006568.2 XM_011536362.2 XM_017020931.1 XM_017020932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361083 c.658G>C p.G220R Chr.14: 54537809-54537809 on Build GRCh38
3700875 c.678+7C>G p.? Chr.14: 54537836-54537836 on Build GRCh38
372758 c.650T>A p.L217* Chr.14: 54537801-54537801 on Build GRCh38
2245118 c.639A>G p.Q213Q Chr.14: 54537790-54537790 on Build GRCh38
6307933 c.678+8G>C p.? Chr.14: 54537837-54537837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562695090 Chr.14: 54537661-54537661 on Build GRCh38
rs1006997168 Chr.14: 54537680-54537680 on Build GRCh38
rs771299427 Chr.14: 54537803-54537803 on Build GRCh38
rs893469110 Chr.14: 54537627-54537627 on Build GRCh38
rs573259109 Chr.14: 54537460-54537460 on Build GRCh38
rs544897139 Chr.14: 54537555-54537555 on Build GRCh38
rs528088515 Chr.14: 54537784-54537784 on Build GRCh38
rs764877085 Chr.14: 54537843-54537843 on Build GRCh38
rs1010856662 Chr.14: 54537654-54537655 on Build GRCh38
rs780350368 Chr.14: 54537763-54537763 on Build GRCh38
rs368828165 Chr.14: 54537515-54537515 on Build GRCh38
rs1052461904 Chr.14: 54537913-54537913 on Build GRCh38
rs753457851 Chr.14: 54537735-54537735 on Build GRCh38
rs749691923 Chr.14: 54537751-54537761 on Build GRCh38
rs1011304832 Chr.14: 54537923-54537923 on Build GRCh38
rs757331684 Chr.14: 54537887-54537887 on Build GRCh38
rs563870382 Chr.14: 54537739-54537739 on Build GRCh38
rs1050123416 Chr.14: 54537937-54537937 on Build GRCh38
rs374420600 Chr.14: 54537824-54537824 on Build GRCh38
rs149482494 Chr.14: 54537728-54537728 on Build GRCh38
rs1052071070 Chr.14: 54537622-54537622 on Build GRCh38
rs965460712 Chr.14: 54537490-54537490 on Build GRCh38
rs550917185 Chr.14: 54537873-54537873 on Build GRCh38
rs749496888 Chr.14: 54537799-54537799 on Build GRCh38
rs777752778 Chr.14: 54537790-54537790 on Build GRCh38
rs373396471 Chr.14: 54537563-54537563 on Build GRCh38
rs890583718 Chr.14: 54537850-54537850 on Build GRCh38
rs529130707 Chr.14: 54537860-54537860 on Build GRCh38
rs1007096027 Chr.14: 54537462-54537462 on Build GRCh38
rs774246276 Chr.14: 54537806-54537806 on Build GRCh38
rs1019647075 Chr.14: 54537489-54537489 on Build GRCh38
rs1025605926 Chr.14: 54537753-54537753 on Build GRCh38
rs751954483 Chr.14: 54537767-54537767 on Build GRCh38
rs370150557 Chr.14: 54537771-54537771 on Build GRCh38
rs935085277 Chr.14: 54537596-54537596 on Build GRCh38
rs199511847 Chr.14: 54537610-54537611 on Build GRCh38
rs946365029 Chr.14: 54537910-54537910 on Build GRCh38
rs774491017 Chr.14: 54537863-54537864 on Build GRCh38
rs115333865 Chr.14: 54537731-54537731 on Build GRCh38
rs372012447 Chr.14: 54537677-54537677 on Build GRCh38
rs774334133 Chr.14: 54537703-54537703 on Build GRCh38
rs893844386 Chr.14: 54537921-54537921 on Build GRCh38
rs890527908 Chr.14: 54537611-54537611 on Build GRCh38
rs984173455 Chr.14: 54537770-54537770 on Build GRCh38
rs981924858 Chr.14: 54537497-54537497 on Build GRCh38
rs556021626 Chr.14: 54537532-54537532 on Build GRCh38
rs771831993 Chr.14: 54537525-54537531 on Build GRCh38
rs1030962722 Chr.14: 54537633-54537633 on Build GRCh38
rs771846035 Chr.14: 54537812-54537812 on Build GRCh38
rs916540889 Chr.14: 54537564-54537564 on Build GRCh38
rs958244383 Chr.14: 54537652-54537652 on Build GRCh38
rs1046291545 Chr.14: 54537580-54537580 on Build GRCh38
rs927679528 Chr.14: 54537585-54537585 on Build GRCh38
rs761417839 Chr.14: 54537839-54537839 on Build GRCh38
rs372258888 Chr.14: 54537853-54537853 on Build GRCh38
rs916426548 Chr.14: 54537458-54537458 on Build GRCh38
rs771138217 Chr.14: 54537826-54537830 on Build GRCh38
rs577773314 Chr.14: 54537710-54537710 on Build GRCh38
rs952626215 Chr.14: 54537548-54537548 on Build GRCh38
rs368440719 Chr.14: 54537837-54537837 on Build GRCh38
rs763639476 Chr.14: 54537836-54537836 on Build GRCh38
rs996745930 Chr.14: 54537605-54537605 on Build GRCh38
rs915032916 Chr.14: 54537889-54537889 on Build GRCh38
rs947873384 Chr.14: 54537575-54537575 on Build GRCh38
rs969270120 Chr.14: 54537663-54537663 on Build GRCh38
rs770815181 Chr.14: 54537696-54537696 on Build GRCh38
rs371449555 Chr.14: 54537809-54537809 on Build GRCh38
rs1037997081 Chr.14: 54537797-54537797 on Build GRCh38
rs546321805 Chr.14: 54537816-54537816 on Build GRCh38
rs1023579332 Chr.14: 54537939-54537939 on Build GRCh38
rs375850699 Chr.14: 54537769-54537769 on Build GRCh38
rs763579500 Chr.14: 54537734-54537734 on Build GRCh38
rs1046775647 Chr.14: 54537546-54537546 on Build GRCh38
rs760569494 Chr.14: 54537830-54537830 on Build GRCh38
rs947622898 Chr.14: 54537642-54537642 on Build GRCh38
rs972686735 Chr.14: 54537517-54537517 on Build GRCh38
rs981916353 Chr.14: 54537804-54537804 on Build GRCh38
rs576108137 Chr.14: 54537539-54537539 on Build GRCh38
rs984449018 Chr.14: 54537560-54537560 on Build GRCh38

View Full Product Details