Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54599256-54599685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNAP
Entrez Gene ID
284254
Gene Name
dynactin associated protein
Gene Aliases
C18orf26
UniGene
Hs.376146
Gene Chromosome Location
Chr.18: 54532713-54599544 on Build GRCh38
Transcripts
NM_001307955.1 NM_173629.2 XM_011525923.2 XM_011525924.2 XM_011525925.2 XM_017025709.1

SNP Detail

SNP ID Chromosome Location
rs534263327 Chr.18: 54599598-54599598 on Build GRCh38
rs985483324 Chr.18: 54599510-54599510 on Build GRCh38
rs1014158305 Chr.18: 54599304-54599304 on Build GRCh38
rs772158352 Chr.18: 54599541-54599541 on Build GRCh38
rs1005527363 Chr.18: 54599366-54599366 on Build GRCh38
rs961648036 Chr.18: 54599423-54599423 on Build GRCh38
rs945089326 Chr.18: 54599597-54599601 on Build GRCh38
rs369080658 Chr.18: 54599604-54599604 on Build GRCh38
rs537312132 Chr.18: 54599338-54599338 on Build GRCh38
rs550806528 Chr.18: 54599396-54599396 on Build GRCh38
rs761174905 Chr.18: 54599620-54599620 on Build GRCh38
rs950047193 Chr.18: 54599595-54599595 on Build GRCh38
rs573509544 Chr.18: 54599472-54599472 on Build GRCh38
rs977029671 Chr.18: 54599436-54599436 on Build GRCh38
rs1021892590 Chr.18: 54599314-54599314 on Build GRCh38
rs562830994 Chr.18: 54599402-54599402 on Build GRCh38
rs985616804 Chr.18: 54599485-54599485 on Build GRCh38
rs552398242 Chr.18: 54599627-54599627 on Build GRCh38
rs903540697 Chr.18: 54599344-54599344 on Build GRCh38
rs935680238 Chr.18: 54599542-54599542 on Build GRCh38
rs530008381 Chr.18: 54599481-54599481 on Build GRCh38
rs771231254 Chr.18: 54599413-54599413 on Build GRCh38
rs912879131 Chr.18: 54599582-54599582 on Build GRCh38
rs992731626 Chr.18: 54599411-54599411 on Build GRCh38
rs376086082 Chr.18: 54599320-54599320 on Build GRCh38
rs1040572395 Chr.18: 54599547-54599547 on Build GRCh38
rs868532407 Chr.18: 54599310-54599310 on Build GRCh38
rs974174069 Chr.18: 54599478-54599478 on Build GRCh38
rs1001853546 Chr.18: 54599345-54599345 on Build GRCh38
rs144845589 Chr.18: 54599349-54599349 on Build GRCh38
rs1015772168 Chr.18: 54599451-54599451 on Build GRCh38
rs1002708320 Chr.18: 54599346-54599346 on Build GRCh38
rs892317683 Chr.18: 54599560-54599560 on Build GRCh38
rs910462813 Chr.18: 54599521-54599521 on Build GRCh38
rs1024581440 Chr.18: 54599308-54599308 on Build GRCh38
rs553044773 Chr.18: 54599607-54599608 on Build GRCh38
rs191094707 Chr.18: 54599483-54599483 on Build GRCh38
rs775490090 Chr.18: 54599551-54599551 on Build GRCh38
rs530357336 Chr.18: 54599405-54599405 on Build GRCh38
rs952711116 Chr.18: 54599356-54599356 on Build GRCh38
rs865799994 Chr.18: 54599525-54599525 on Build GRCh38

View Full Product Details