Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43869203-43869652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC122
Entrez Gene ID
160857
Gene Name
coiled-coil domain containing 122
Gene Aliases
-
UniGene
Hs.170849
Gene Chromosome Location
Chr.13: 43821959-43879690 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020398.1 XM_017020399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947604 c.9C>G p.D3E Chr.13: 43869368-43869368 on Build GRCh38
6349414 c.24G>C p.K8N Chr.13: 43869353-43869353 on Build GRCh38
4909009 c.40A>G p.K14E Chr.13: 43869337-43869337 on Build GRCh38
6349415 c.24G>C p.K8N Chr.13: 43869353-43869353 on Build GRCh38
4907345 c.43G>A p.E15K Chr.13: 43869334-43869334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915339457 Chr.13: 43869601-43869601 on Build GRCh38
rs968036047 Chr.13: 43869389-43869389 on Build GRCh38
rs778067049 Chr.13: 43869391-43869391 on Build GRCh38
rs956753231 Chr.13: 43869576-43869576 on Build GRCh38
rs184663196 Chr.13: 43869475-43869475 on Build GRCh38
rs957684829 Chr.13: 43869264-43869264 on Build GRCh38
rs748241412 Chr.13: 43869392-43869393 on Build GRCh38
rs550438423 Chr.13: 43869398-43869398 on Build GRCh38
rs748267723 Chr.13: 43869290-43869290 on Build GRCh38
rs57755231 Chr.13: 43869426-43869426 on Build GRCh38
rs764412585 Chr.13: 43869393-43869393 on Build GRCh38
rs557717870 Chr.13: 43869481-43869481 on Build GRCh38
rs765839782 Chr.13: 43869361-43869361 on Build GRCh38
rs542821122 Chr.13: 43869281-43869281 on Build GRCh38
rs762138887 Chr.13: 43869396-43869396 on Build GRCh38
rs1005348762 Chr.13: 43869442-43869442 on Build GRCh38
rs776072708 Chr.13: 43869626-43869626 on Build GRCh38
rs772138271 Chr.13: 43869322-43869322 on Build GRCh38
rs538450840 Chr.13: 43869363-43869363 on Build GRCh38
rs900866366 Chr.13: 43869320-43869320 on Build GRCh38
rs1023512309 Chr.13: 43869464-43869464 on Build GRCh38
rs781584959 Chr.13: 43869357-43869360 on Build GRCh38
rs71429465 Chr.13: 43869287-43869287 on Build GRCh38
rs779766946 Chr.13: 43869282-43869282 on Build GRCh38
rs746347176 Chr.13: 43869589-43869589 on Build GRCh38
rs928800301 Chr.13: 43869417-43869417 on Build GRCh38
rs771041405 Chr.13: 43869340-43869340 on Build GRCh38
rs1011018600 Chr.13: 43869305-43869305 on Build GRCh38
rs772602674 Chr.13: 43869604-43869604 on Build GRCh38
rs771319117 Chr.13: 43869440-43869440 on Build GRCh38
rs763430315 Chr.13: 43869365-43869365 on Build GRCh38
rs17065161 Chr.13: 43869587-43869587 on Build GRCh38
rs879694141 Chr.13: 43869523-43869523 on Build GRCh38
rs756352864 Chr.13: 43869342-43869342 on Build GRCh38
rs776636659 Chr.13: 43869347-43869347 on Build GRCh38
rs77941962 Chr.13: 43869559-43869559 on Build GRCh38
rs889371052 Chr.13: 43869257-43869257 on Build GRCh38
rs747238701 Chr.13: 43869328-43869328 on Build GRCh38
rs1021874956 Chr.13: 43869323-43869323 on Build GRCh38
rs533914335 Chr.13: 43869286-43869286 on Build GRCh38
rs760066211 Chr.13: 43869349-43869349 on Build GRCh38
rs993860023 Chr.13: 43869394-43869394 on Build GRCh38
rs375265750 Chr.13: 43869435-43869435 on Build GRCh38
rs773464221 Chr.13: 43869326-43869326 on Build GRCh38
rs1041102373 Chr.13: 43869318-43869318 on Build GRCh38
rs573458013 Chr.13: 43869600-43869600 on Build GRCh38
rs572765391 Chr.13: 43869497-43869497 on Build GRCh38

View Full Product Details