Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111789994-111790525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH2
Entrez Gene ID
217
Gene Name
aldehyde dehydrogenase 2 family (mitochondrial)
Gene Aliases
ALDH-E2, ALDHI, ALDM
UniGene
Hs.604551
Gene Chromosome Location
Chr.12: 111766887-111809985 on Build GRCh38
Transcripts
NM_000690.3 NM_001204889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598256 c.565C>T p.L189F Chr.12: 111790446-111790446 on Build GRCh38
1358791 c.571A>G p.M191V Chr.12: 111790452-111790452 on Build GRCh38
6331720 c.580T>A p.W194R Chr.12: 111790461-111790461 on Build GRCh38
80854 c.567C>G p.L189L Chr.12: 111790448-111790448 on Build GRCh38
5526513 c.555G>A p.W185* Chr.12: 111790436-111790436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371563129 Chr.12: 111790218-111790218 on Build GRCh38
rs868852015 Chr.12: 111790489-111790489 on Build GRCh38
rs138392178 Chr.12: 111790298-111790298 on Build GRCh38
rs917865245 Chr.12: 111790479-111790479 on Build GRCh38
rs1006282909 Chr.12: 111790364-111790364 on Build GRCh38
rs760265769 Chr.12: 111790219-111790219 on Build GRCh38
rs1056328713 Chr.12: 111790167-111790167 on Build GRCh38
rs766713154 Chr.12: 111790477-111790477 on Build GRCh38
rs1031032508 Chr.12: 111790294-111790294 on Build GRCh38
rs776360702 Chr.12: 111790303-111790303 on Build GRCh38
rs983540448 Chr.12: 111790051-111790051 on Build GRCh38
rs561726454 Chr.12: 111790073-111790073 on Build GRCh38
rs1000623830 Chr.12: 111790232-111790232 on Build GRCh38
rs907198254 Chr.12: 111790320-111790320 on Build GRCh38
rs749854544 Chr.12: 111790021-111790021 on Build GRCh38
rs767984978 Chr.12: 111790264-111790264 on Build GRCh38
rs577749293 Chr.12: 111790099-111790099 on Build GRCh38
rs149241562 Chr.12: 111790428-111790428 on Build GRCh38
rs370071768 Chr.12: 111790418-111790418 on Build GRCh38
rs896307451 Chr.12: 111790208-111790208 on Build GRCh38
rs927306164 Chr.12: 111790090-111790090 on Build GRCh38
rs909314522 Chr.12: 111790063-111790063 on Build GRCh38
rs1017239590 Chr.12: 111790246-111790246 on Build GRCh38
rs548961554 Chr.12: 111790350-111790350 on Build GRCh38
rs887879968 Chr.12: 111790352-111790352 on Build GRCh38
rs371655800 Chr.12: 111790075-111790075 on Build GRCh38
rs950896524 Chr.12: 111790295-111790295 on Build GRCh38
rs1014695177 Chr.12: 111790367-111790367 on Build GRCh38
rs527603470 Chr.12: 111790322-111790322 on Build GRCh38
rs75029020 Chr.12: 111790217-111790217 on Build GRCh38
rs925031301 Chr.12: 111790184-111790184 on Build GRCh38
rs540537301 Chr.12: 111790054-111790054 on Build GRCh38
rs980160197 Chr.12: 111790379-111790379 on Build GRCh38
rs764544132 Chr.12: 111790410-111790410 on Build GRCh38
rs984066293 Chr.12: 111790317-111790317 on Build GRCh38
rs922919610 Chr.12: 111790023-111790023 on Build GRCh38
rs532985612 Chr.12: 111790269-111790269 on Build GRCh38
rs927273521 Chr.12: 111790401-111790401 on Build GRCh38
rs770671499 Chr.12: 111790402-111790402 on Build GRCh38
rs761027843 Chr.12: 111790407-111790407 on Build GRCh38
rs775770152 Chr.12: 111790403-111790403 on Build GRCh38
rs754845817 Chr.12: 111790494-111790494 on Build GRCh38
rs771544832 Chr.12: 111790149-111790149 on Build GRCh38
rs79986087 Chr.12: 111790432-111790432 on Build GRCh38
rs903633560 Chr.12: 111790226-111790226 on Build GRCh38
rs58280059 Chr.12: 111790445-111790445 on Build GRCh38
rs942040448 Chr.12: 111790074-111790074 on Build GRCh38
rs780065997 Chr.12: 111790491-111790491 on Build GRCh38
rs986244307 Chr.12: 111790122-111790122 on Build GRCh38
rs963398174 Chr.12: 111790373-111790373 on Build GRCh38
rs751364520 Chr.12: 111790493-111790493 on Build GRCh38
rs550379014 Chr.12: 111790119-111790119 on Build GRCh38
rs150107349 Chr.12: 111790161-111790161 on Build GRCh38
rs762429882 Chr.12: 111790421-111790421 on Build GRCh38
rs964390486 Chr.12: 111790251-111790251 on Build GRCh38
rs758493011 Chr.12: 111790460-111790460 on Build GRCh38
rs1044410634 Chr.12: 111790088-111790088 on Build GRCh38
rs976052007 Chr.12: 111790022-111790022 on Build GRCh38
rs1001547573 Chr.12: 111790337-111790337 on Build GRCh38
rs953112420 Chr.12: 111790036-111790036 on Build GRCh38
rs566294630 Chr.12: 111790319-111790319 on Build GRCh38
rs562241287 Chr.12: 111790234-111790234 on Build GRCh38

View Full Product Details