Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78113911-78114411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
UniGene
Hs.503368
Gene Chromosome Location
Chr.11: 78100942-78139660 on Build GRCh38
Transcripts
NM_001007027.2 NM_024079.4 XM_005274247.3 XM_011545252.2 XM_017018274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1357153 c.571C>A p.L191I Chr.11: 78114368-78114368 on Build GRCh38
6070291 c.647C>G p.S216C Chr.11: 78114292-78114292 on Build GRCh38
5827056 c.571C>A p.L191I Chr.11: 78114368-78114368 on Build GRCh38
5827055 c.665A>G p.N222S Chr.11: 78114274-78114274 on Build GRCh38
6441994 c.551_552GG>AA p.R184K Chr.11: 78114387-78114388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141280441 Chr.11: 78113962-78113962 on Build GRCh38
rs756061099 Chr.11: 78113977-78113977 on Build GRCh38
rs767435809 Chr.11: 78114231-78114231 on Build GRCh38
rs941211985 Chr.11: 78114212-78114212 on Build GRCh38
rs780484107 Chr.11: 78114288-78114288 on Build GRCh38
rs531760134 Chr.11: 78114381-78114381 on Build GRCh38
rs747131228 Chr.11: 78114126-78114126 on Build GRCh38
rs754501798 Chr.11: 78114261-78114261 on Build GRCh38
rs774505315 Chr.11: 78114219-78114219 on Build GRCh38
rs750393405 Chr.11: 78113963-78113963 on Build GRCh38
rs766958404 Chr.11: 78114247-78114249 on Build GRCh38
rs775945856 Chr.11: 78114031-78114031 on Build GRCh38
rs1056348150 Chr.11: 78114343-78114343 on Build GRCh38
rs374436755 Chr.11: 78114215-78114215 on Build GRCh38
rs199603474 Chr.11: 78113996-78113996 on Build GRCh38
rs201602294 Chr.11: 78114295-78114295 on Build GRCh38
rs773541071 Chr.11: 78114327-78114327 on Build GRCh38
rs562915665 Chr.11: 78114351-78114351 on Build GRCh38
rs763258107 Chr.11: 78113953-78113953 on Build GRCh38
rs572693 Chr.11: 78114018-78114018 on Build GRCh38
rs61566223 Chr.11: 78114020-78114020 on Build GRCh38
rs759045647 Chr.11: 78114027-78114034 on Build GRCh38
rs979344807 Chr.11: 78114320-78114320 on Build GRCh38
rs547807006 Chr.11: 78113968-78113968 on Build GRCh38
rs769865988 Chr.11: 78113944-78113944 on Build GRCh38
rs751783361 Chr.11: 78113961-78113961 on Build GRCh38
rs907817618 Chr.11: 78114316-78114316 on Build GRCh38
rs150736564 Chr.11: 78113981-78113981 on Build GRCh38
rs755038379 Chr.11: 78114159-78114159 on Build GRCh38
rs1050014915 Chr.11: 78114156-78114156 on Build GRCh38
rs140571089 Chr.11: 78114203-78114203 on Build GRCh38
rs750895613 Chr.11: 78113938-78113938 on Build GRCh38
rs1036755831 Chr.11: 78114214-78114214 on Build GRCh38
rs911388071 Chr.11: 78114207-78114207 on Build GRCh38
rs759448781 Chr.11: 78114342-78114342 on Build GRCh38
rs758076775 Chr.11: 78114354-78114354 on Build GRCh38
rs774434021 Chr.11: 78114296-78114296 on Build GRCh38
rs888311317 Chr.11: 78114167-78114167 on Build GRCh38
rs769894472 Chr.11: 78114323-78114323 on Build GRCh38
rs201887966 Chr.11: 78114232-78114232 on Build GRCh38
rs780673624 Chr.11: 78114083-78114083 on Build GRCh38
rs754476593 Chr.11: 78113985-78113985 on Build GRCh38
rs192241801 Chr.11: 78114276-78114276 on Build GRCh38
rs886048687 Chr.11: 78114292-78114292 on Build GRCh38
rs760591590 Chr.11: 78114332-78114332 on Build GRCh38
rs533514438 Chr.11: 78114011-78114011 on Build GRCh38
rs1056292840 Chr.11: 78114084-78114084 on Build GRCh38
rs533704173 Chr.11: 78113978-78113978 on Build GRCh38
rs183585747 Chr.11: 78114312-78114312 on Build GRCh38
rs764796685 Chr.11: 78114266-78114266 on Build GRCh38
rs766260217 Chr.11: 78114334-78114334 on Build GRCh38
rs764263526 Chr.11: 78113956-78113956 on Build GRCh38
rs759036484 Chr.11: 78114072-78114072 on Build GRCh38
rs665278 Chr.11: 78114274-78114274 on Build GRCh38
rs748406490 Chr.11: 78114306-78114306 on Build GRCh38
rs775490122 Chr.11: 78113951-78113951 on Build GRCh38
rs1003406006 Chr.11: 78114080-78114080 on Build GRCh38
rs924711179 Chr.11: 78114047-78114047 on Build GRCh38
rs763785301 Chr.11: 78114369-78114369 on Build GRCh38
rs556076437 Chr.11: 78114141-78114141 on Build GRCh38
rs11237397 Chr.11: 78114016-78114016 on Build GRCh38
rs955606485 Chr.11: 78114175-78114175 on Build GRCh38
rs139106381 Chr.11: 78113988-78113988 on Build GRCh38
rs762991744 Chr.11: 78114036-78114036 on Build GRCh38
rs753787032 Chr.11: 78114243-78114243 on Build GRCh38
rs978844670 Chr.11: 78114035-78114035 on Build GRCh38
rs1023657363 Chr.11: 78114310-78114310 on Build GRCh38
rs752295244 Chr.11: 78114268-78114268 on Build GRCh38
rs201538885 Chr.11: 78114270-78114270 on Build GRCh38
rs932618101 Chr.11: 78114147-78114147 on Build GRCh38
rs967698642 Chr.11: 78114131-78114131 on Build GRCh38
rs768929600 Chr.11: 78114050-78114050 on Build GRCh38
rs1031676586 Chr.11: 78114322-78114322 on Build GRCh38
rs776497184 Chr.11: 78114340-78114340 on Build GRCh38
rs986953385 Chr.11: 78114194-78114194 on Build GRCh38
rs766456373 Chr.11: 78114233-78114233 on Build GRCh38
rs371571471 Chr.11: 78114009-78114009 on Build GRCh38
rs968885498 Chr.11: 78114317-78114317 on Build GRCh38
rs777227210 Chr.11: 78114088-78114088 on Build GRCh38
rs886287555 Chr.11: 78114073-78114073 on Build GRCh38
rs1011992878 Chr.11: 78114120-78114120 on Build GRCh38
rs10899440 Chr.11: 78114229-78114229 on Build GRCh38
rs886048688 Chr.11: 78114316-78114316 on Build GRCh38

View Full Product Details