Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93759721-93760277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf54
Entrez Gene ID
28970
Gene Name
chromosome 11 open reading frame 54
Gene Aliases
PTD012, PTOD012
UniGene
Hs.8360
Gene Chromosome Location
Chr.11: 93741594-93765323 on Build GRCh38
Transcripts
NM_001286067.1 NM_001286068.1 NM_001286069.1 NM_001286070.1 NM_001286071.1 NM_014039.3 XM_006718824.1 XM_011542781.2 XM_011542782.2 XM_017017613.1 XM_017017614.1 XM_017017615.1 XM_017017616.1 XM_017017617.1 XM_017017618.1 XM_017017619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3454694 c.539C>T p.S180F Chr.11: 93759773-93759773 on Build GRCh38
3810694 c.773C>G p.P258R Chr.11: 93759857-93759857 on Build GRCh38
3454695 c.689C>T p.S230F Chr.11: 93759773-93759773 on Build GRCh38
5479899 c.578T>A p.M193K Chr.11: 93759812-93759812 on Build GRCh38
3810693 c.623C>G p.P208R Chr.11: 93759857-93759857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957915799 Chr.11: 93760204-93760204 on Build GRCh38
rs11600142 Chr.11: 93760146-93760146 on Build GRCh38
rs750483653 Chr.11: 93760129-93760129 on Build GRCh38
rs970521243 Chr.11: 93760240-93760240 on Build GRCh38
rs534683737 Chr.11: 93759857-93759857 on Build GRCh38
rs199806624 Chr.11: 93759827-93759827 on Build GRCh38
rs546720382 Chr.11: 93759891-93759891 on Build GRCh38
rs898707260 Chr.11: 93760038-93760038 on Build GRCh38
rs759357177 Chr.11: 93759874-93759875 on Build GRCh38
rs749598202 Chr.11: 93760061-93760061 on Build GRCh38
rs753321529 Chr.11: 93759768-93759768 on Build GRCh38
rs80125365 Chr.11: 93760107-93760107 on Build GRCh38
rs11603174 Chr.11: 93760212-93760212 on Build GRCh38
rs768722332 Chr.11: 93759812-93759812 on Build GRCh38
rs624883 Chr.11: 93760236-93760236 on Build GRCh38
rs11602715 Chr.11: 93760197-93760197 on Build GRCh38
rs557646283 Chr.11: 93759978-93759978 on Build GRCh38
rs774300878 Chr.11: 93759781-93759785 on Build GRCh38
rs769927816 Chr.11: 93759792-93759792 on Build GRCh38
rs1002422919 Chr.11: 93760194-93760194 on Build GRCh38
rs771230635 Chr.11: 93760103-93760103 on Build GRCh38
rs772702735 Chr.11: 93760245-93760245 on Build GRCh38
rs191307246 Chr.11: 93760133-93760133 on Build GRCh38
rs770126324 Chr.11: 93759782-93759782 on Build GRCh38
rs748302157 Chr.11: 93759777-93759777 on Build GRCh38
rs925288076 Chr.11: 93759998-93759998 on Build GRCh38
rs77740618 Chr.11: 93760123-93760123 on Build GRCh38
rs186645027 Chr.11: 93759849-93759849 on Build GRCh38
rs139891911 Chr.11: 93759806-93759806 on Build GRCh38
rs565241737 Chr.11: 93760147-93760147 on Build GRCh38
rs11600144 Chr.11: 93760191-93760191 on Build GRCh38
rs181140249 Chr.11: 93759821-93759821 on Build GRCh38
rs963863190 Chr.11: 93760156-93760156 on Build GRCh38
rs753475113 Chr.11: 93759858-93759858 on Build GRCh38
rs996914269 Chr.11: 93760160-93760160 on Build GRCh38
rs541022879 Chr.11: 93760237-93760237 on Build GRCh38
rs1046211993 Chr.11: 93759903-93759903 on Build GRCh38
rs937331456 Chr.11: 93759953-93759953 on Build GRCh38
rs117997839 Chr.11: 93759911-93759911 on Build GRCh38
rs775738134 Chr.11: 93759802-93759802 on Build GRCh38
rs34454784 Chr.11: 93760082-93760083 on Build GRCh38
rs535362384 Chr.11: 93760050-93760050 on Build GRCh38
rs11602282 Chr.11: 93760184-93760184 on Build GRCh38
rs936634590 Chr.11: 93760090-93760090 on Build GRCh38
rs759029666 Chr.11: 93759890-93759890 on Build GRCh38
rs761734799 Chr.11: 93759834-93759834 on Build GRCh38
rs150664273 Chr.11: 93759749-93759749 on Build GRCh38
rs937442228 Chr.11: 93760246-93760246 on Build GRCh38
rs1055837114 Chr.11: 93760033-93760033 on Build GRCh38
rs11602712 Chr.11: 93760177-93760177 on Build GRCh38
rs747509218 Chr.11: 93759778-93759778 on Build GRCh38
rs201911102 Chr.11: 93759836-93759836 on Build GRCh38
rs555484131 Chr.11: 93760134-93760134 on Build GRCh38
rs764803015 Chr.11: 93759897-93759897 on Build GRCh38
rs867564773 Chr.11: 93760104-93760104 on Build GRCh38
rs754446298 Chr.11: 93759773-93759773 on Build GRCh38
rs766008621 Chr.11: 93759765-93759765 on Build GRCh38
rs746137441 Chr.11: 93759784-93759784 on Build GRCh38
rs932542987 Chr.11: 93759944-93759945 on Build GRCh38
rs982001551 Chr.11: 93760241-93760241 on Build GRCh38
rs543666638 Chr.11: 93760072-93760072 on Build GRCh38
rs11602719 Chr.11: 93760211-93760211 on Build GRCh38
rs78390159 Chr.11: 93760108-93760108 on Build GRCh38
rs1050447059 Chr.11: 93759945-93759947 on Build GRCh38
rs575876472 Chr.11: 93760092-93760092 on Build GRCh38
rs377240951 Chr.11: 93759966-93759966 on Build GRCh38
rs11600143 Chr.11: 93760157-93760157 on Build GRCh38
rs1032011136 Chr.11: 93760201-93760201 on Build GRCh38

View Full Product Details