Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66800592-66801140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf80
Entrez Gene ID
79703
Gene Name
chromosome 11 open reading frame 80
Gene Aliases
TOP6BL, TOPOVIBL
UniGene
Hs.292088
Gene Chromosome Location
Chr.11: 66744736-66843516 on Build GRCh38
Transcripts
NM_001302084.1 NM_024650.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7193865 c.746A>G p.N249S Chr.11: 66800669-66800669 on Build GRCh38
7195293 c.766A>G p.I256V Chr.11: 66800689-66800689 on Build GRCh38
6333822 c.718-4G>T p.? Chr.11: 66800637-66800637 on Build GRCh38
3869966 c.727C>T p.H243Y Chr.11: 66800650-66800650 on Build GRCh38
6998114 c.805A>G p.N269D Chr.11: 66801062-66801062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373406160 Chr.11: 66801073-66801073 on Build GRCh38
rs917674405 Chr.11: 66800925-66800925 on Build GRCh38
rs771486212 Chr.11: 66801078-66801078 on Build GRCh38
rs540750934 Chr.11: 66800907-66800907 on Build GRCh38
rs114055882 Chr.11: 66801077-66801077 on Build GRCh38
rs759977903 Chr.11: 66800914-66800914 on Build GRCh38
rs781019380 Chr.11: 66800683-66800683 on Build GRCh38
rs564710567 Chr.11: 66800807-66800807 on Build GRCh38
rs749060869 Chr.11: 66801068-66801068 on Build GRCh38
rs779020740 Chr.11: 66800646-66800646 on Build GRCh38
rs745766170 Chr.11: 66800689-66800689 on Build GRCh38
rs533528462 Chr.11: 66800813-66800813 on Build GRCh38
rs191651158 Chr.11: 66800676-66800676 on Build GRCh38
rs761442553 Chr.11: 66800728-66800728 on Build GRCh38
rs1008061430 Chr.11: 66800638-66800638 on Build GRCh38
rs964514593 Chr.11: 66800853-66800853 on Build GRCh38
rs772772197 Chr.11: 66801079-66801079 on Build GRCh38
rs868620595 Chr.11: 66801102-66801102 on Build GRCh38
rs758492034 Chr.11: 66801027-66801027 on Build GRCh38
rs769485214 Chr.11: 66800695-66800695 on Build GRCh38
rs560596554 Chr.11: 66800932-66800932 on Build GRCh38
rs777073140 Chr.11: 66800769-66800769 on Build GRCh38
rs765534660 Chr.11: 66801091-66801091 on Build GRCh38
rs767088039 Chr.11: 66800981-66800981 on Build GRCh38
rs768404905 Chr.11: 66800714-66800714 on Build GRCh38
rs765411695 Chr.11: 66801016-66801016 on Build GRCh38
rs754011225 Chr.11: 66800632-66800632 on Build GRCh38
rs761177351 Chr.11: 66801006-66801006 on Build GRCh38
rs561838350 Chr.11: 66801111-66801111 on Build GRCh38
rs777845483 Chr.11: 66801038-66801038 on Build GRCh38
rs971563581 Chr.11: 66800928-66800928 on Build GRCh38
rs369298599 Chr.11: 66801048-66801048 on Build GRCh38
rs150252276 Chr.11: 66801012-66801012 on Build GRCh38
rs1000545137 Chr.11: 66800793-66800793 on Build GRCh38
rs777016876 Chr.11: 66801085-66801085 on Build GRCh38
rs183737871 Chr.11: 66800764-66800764 on Build GRCh38
rs766295177 Chr.11: 66800620-66800624 on Build GRCh38
rs890105934 Chr.11: 66800701-66800701 on Build GRCh38
rs376103610 Chr.11: 66800709-66800709 on Build GRCh38
rs989237237 Chr.11: 66800936-66800936 on Build GRCh38
rs988332995 Chr.11: 66800885-66800886 on Build GRCh38
rs766886411 Chr.11: 66801083-66801083 on Build GRCh38
rs748977351 Chr.11: 66800872-66800873 on Build GRCh38
rs752926094 Chr.11: 66801103-66801103 on Build GRCh38
rs766718266 Chr.11: 66801010-66801010 on Build GRCh38
rs751640272 Chr.11: 66801112-66801112 on Build GRCh38
rs752872582 Chr.11: 66801021-66801021 on Build GRCh38
rs542244777 Chr.11: 66800636-66800636 on Build GRCh38
rs755187009 Chr.11: 66800645-66800645 on Build GRCh38
rs1026003343 Chr.11: 66800700-66800700 on Build GRCh38
rs927564111 Chr.11: 66800937-66800937 on Build GRCh38
rs1022649575 Chr.11: 66800969-66800969 on Build GRCh38
rs948874647 Chr.11: 66801089-66801089 on Build GRCh38
rs189077398 Chr.11: 66801110-66801110 on Build GRCh38
rs759921697 Chr.11: 66801090-66801090 on Build GRCh38
rs373448823 Chr.11: 66800630-66800630 on Build GRCh38
rs750906058 Chr.11: 66801001-66801001 on Build GRCh38
rs779764796 Chr.11: 66801066-66801066 on Build GRCh38
rs754215262 Chr.11: 66801011-66801011 on Build GRCh38
rs768531414 Chr.11: 66801076-66801076 on Build GRCh38
rs774045692 Chr.11: 66800719-66800719 on Build GRCh38
rs756050388 Chr.11: 66801117-66801117 on Build GRCh38
rs762755722 Chr.11: 66800713-66800713 on Build GRCh38
rs756458009 Chr.11: 66800622-66800622 on Build GRCh38
rs963942945 Chr.11: 66800773-66800773 on Build GRCh38
rs937570871 Chr.11: 66800966-66800966 on Build GRCh38
rs770893070 Chr.11: 66800680-66800680 on Build GRCh38
rs761143330 Chr.11: 66801082-66801082 on Build GRCh38
rs745863865 Chr.11: 66801045-66801045 on Build GRCh38
rs796730264 Chr.11: 66800874-66800874 on Build GRCh38
rs779402951 Chr.11: 66801039-66801040 on Build GRCh38
rs1017303739 Chr.11: 66800761-66800761 on Build GRCh38
rs374243212 Chr.11: 66800970-66800970 on Build GRCh38

View Full Product Details