Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 2623006-2623556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01600
Entrez Gene ID
154386
Gene Name
long intergenic non-protein coding RNA 1600
Gene Aliases
C6orf195, bA145H9.2
UniGene
Hs.680574
Gene Chromosome Location
Chr.6: 2621913-2634603 on Build GRCh38
Transcripts
NR_131168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3623476 c.360G>A p.R120R Chr.6: 2623463-2623463 on Build GRCh38
7156670 c.357G>A p.Q119Q Chr.6: 2623466-2623466 on Build GRCh38
5351303 c.290C>T p.P97L Chr.6: 2623533-2623533 on Build GRCh38
3623477 c.287C>T p.T96I Chr.6: 2623536-2623536 on Build GRCh38
5404965 c.348C>T p.I116I Chr.6: 2623475-2623475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375013794 Chr.6: 2623451-2623451 on Build GRCh38
rs73348509 Chr.6: 2623437-2623437 on Build GRCh38
rs773310816 Chr.6: 2623484-2623484 on Build GRCh38
rs1006163325 Chr.6: 2623058-2623058 on Build GRCh38
rs750929554 Chr.6: 2623197-2623197 on Build GRCh38
rs771038967 Chr.6: 2623268-2623268 on Build GRCh38
rs765386941 Chr.6: 2623481-2623481 on Build GRCh38
rs1005995764 Chr.6: 2623233-2623233 on Build GRCh38
rs901730383 Chr.6: 2623187-2623187 on Build GRCh38
rs543423323 Chr.6: 2623142-2623142 on Build GRCh38
rs768985354 Chr.6: 2623471-2623471 on Build GRCh38
rs1025532676 Chr.6: 2623092-2623092 on Build GRCh38
rs763086464 Chr.6: 2623496-2623496 on Build GRCh38
rs374802838 Chr.6: 2623232-2623232 on Build GRCh38
rs1015062275 Chr.6: 2623255-2623255 on Build GRCh38
rs751519596 Chr.6: 2623417-2623417 on Build GRCh38
rs980999034 Chr.6: 2623113-2623113 on Build GRCh38
rs182198110 Chr.6: 2623465-2623465 on Build GRCh38
rs10591789 Chr.6: 2623327-2623328 on Build GRCh38
rs926845664 Chr.6: 2623135-2623135 on Build GRCh38
rs201834992 Chr.6: 2623470-2623470 on Build GRCh38
rs889137208 Chr.6: 2623037-2623037 on Build GRCh38
rs761117514 Chr.6: 2623286-2623286 on Build GRCh38
rs957721440 Chr.6: 2623170-2623170 on Build GRCh38
rs570831553 Chr.6: 2623467-2623467 on Build GRCh38
rs1025161070 Chr.6: 2623326-2623326 on Build GRCh38
rs754810668 Chr.6: 2623420-2623420 on Build GRCh38
rs897919871 Chr.6: 2623310-2623310 on Build GRCh38
rs1024680257 Chr.6: 2623523-2623523 on Build GRCh38
rs774909924 Chr.6: 2623077-2623077 on Build GRCh38
rs550141268 Chr.6: 2623503-2623503 on Build GRCh38
rs550100942 Chr.6: 2623477-2623477 on Build GRCh38
rs762434303 Chr.6: 2623184-2623184 on Build GRCh38
rs533447499 Chr.6: 2623480-2623480 on Build GRCh38
rs969029636 Chr.6: 2623534-2623534 on Build GRCh38
rs752779533 Chr.6: 2623505-2623505 on Build GRCh38
rs1038064953 Chr.6: 2623309-2623309 on Build GRCh38
rs9501863 Chr.6: 2623189-2623189 on Build GRCh38
rs922415698 Chr.6: 2623262-2623262 on Build GRCh38
rs376820310 Chr.6: 2623475-2623475 on Build GRCh38
rs888354827 Chr.6: 2623287-2623287 on Build GRCh38
rs548860029 Chr.6: 2623344-2623344 on Build GRCh38
rs962433463 Chr.6: 2623074-2623074 on Build GRCh38
rs541776789 Chr.6: 2623388-2623388 on Build GRCh38
rs970984657 Chr.6: 2623112-2623112 on Build GRCh38
rs9378719 Chr.6: 2623353-2623353 on Build GRCh38
rs993962558 Chr.6: 2623325-2623325 on Build GRCh38
rs1016381833 Chr.6: 2623059-2623059 on Build GRCh38
rs913273818 Chr.6: 2623380-2623380 on Build GRCh38
rs766429372 Chr.6: 2623499-2623499 on Build GRCh38
rs946022112 Chr.6: 2623147-2623147 on Build GRCh38
rs1033966431 Chr.6: 2623392-2623392 on Build GRCh38
rs993680799 Chr.6: 2623080-2623080 on Build GRCh38
rs957752870 Chr.6: 2623352-2623352 on Build GRCh38
rs990264182 Chr.6: 2623377-2623377 on Build GRCh38
rs6901966 Chr.6: 2623299-2623299 on Build GRCh38
rs1028861968 Chr.6: 2623203-2623203 on Build GRCh38
rs776531474 Chr.6: 2623071-2623071 on Build GRCh38
rs774533245 Chr.6: 2623269-2623270 on Build GRCh38
rs932467938 Chr.6: 2623276-2623276 on Build GRCh38
rs891812861 Chr.6: 2623143-2623143 on Build GRCh38
rs199691784 Chr.6: 2623407-2623407 on Build GRCh38
rs755008271 Chr.6: 2623507-2623507 on Build GRCh38
rs777020887 Chr.6: 2623478-2623478 on Build GRCh38
rs781184984 Chr.6: 2623433-2623433 on Build GRCh38
rs751496368 Chr.6: 2623501-2623501 on Build GRCh38
rs777309751 Chr.6: 2623447-2623447 on Build GRCh38
rs755197728 Chr.6: 2623394-2623394 on Build GRCh38
rs1040393766 Chr.6: 2623409-2623409 on Build GRCh38
rs372677511 Chr.6: 2623438-2623438 on Build GRCh38
rs1040896181 Chr.6: 2623234-2623234 on Build GRCh38
rs371708638 Chr.6: 2623227-2623227 on Build GRCh38
rs927450302 Chr.6: 2623104-2623104 on Build GRCh38
rs769454366 Chr.6: 2623038-2623038 on Build GRCh38
rs191001500 Chr.6: 2623190-2623190 on Build GRCh38
rs775990888 Chr.6: 2623381-2623381 on Build GRCh38

View Full Product Details