Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69820877-69821436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf135
Entrez Gene ID
138255
Gene Name
chromosome 9 open reading frame 135
Gene Aliases
-
UniGene
Hs.444459
Gene Chromosome Location
Chr.9: 69820804-69906237 on Build GRCh38
Transcripts
NM_001010940.2 NM_001308084.1 NM_001308085.1 NM_001308086.1 NR_131756.1 NR_131757.1 XM_011518229.2 XM_011518230.2 XM_011518231.2 XM_011518234.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
80949 c.120G>A p.K40K Chr.9: 69820999-69820999 on Build GRCh38
3848700 c.86G>A p.R29H Chr.9: 69820965-69820965 on Build GRCh38
3731050 c.37G>A p.D13N Chr.9: 69820916-69820916 on Build GRCh38
1109743 c.106C>T p.R36C Chr.9: 69820985-69820985 on Build GRCh38
3907746 c.150C>T p.S50S Chr.9: 69821029-69821029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889816009 Chr.9: 69821329-69821329 on Build GRCh38
rs1007186204 Chr.9: 69821121-69821121 on Build GRCh38
rs760463078 Chr.9: 69820913-69820913 on Build GRCh38
rs545079705 Chr.9: 69821175-69821175 on Build GRCh38
rs974539234 Chr.9: 69821259-69821259 on Build GRCh38
rs747083996 Chr.9: 69821360-69821360 on Build GRCh38
rs375047820 Chr.9: 69821048-69821048 on Build GRCh38
rs768524639 Chr.9: 69820985-69820985 on Build GRCh38
rs745462491 Chr.9: 69820970-69820970 on Build GRCh38
rs1051107302 Chr.9: 69821320-69821320 on Build GRCh38
rs971145491 Chr.9: 69821405-69821405 on Build GRCh38
rs368950631 Chr.9: 69821067-69821067 on Build GRCh38
rs751642816 Chr.9: 69821015-69821015 on Build GRCh38
rs528660519 Chr.9: 69821209-69821209 on Build GRCh38
rs149641101 Chr.9: 69820968-69820968 on Build GRCh38
rs2026796 Chr.9: 69821081-69821081 on Build GRCh38
rs946143563 Chr.9: 69821342-69821342 on Build GRCh38
rs1019087023 Chr.9: 69821225-69821225 on Build GRCh38
rs1025445136 Chr.9: 69821404-69821404 on Build GRCh38
rs937515341 Chr.9: 69821390-69821390 on Build GRCh38
rs748566007 Chr.9: 69820961-69820961 on Build GRCh38
rs953989931 Chr.9: 69821097-69821097 on Build GRCh38
rs1006889754 Chr.9: 69821345-69821345 on Build GRCh38
rs991521023 Chr.9: 69821274-69821274 on Build GRCh38
rs778216844 Chr.9: 69821058-69821058 on Build GRCh38
rs935899738 Chr.9: 69821277-69821277 on Build GRCh38
rs776467459 Chr.9: 69821141-69821141 on Build GRCh38
rs143163232 Chr.9: 69820910-69820910 on Build GRCh38
rs755057937 Chr.9: 69821020-69821020 on Build GRCh38
rs898401699 Chr.9: 69821374-69821374 on Build GRCh38
rs768170659 Chr.9: 69821080-69821080 on Build GRCh38
rs539450293 Chr.9: 69821401-69821401 on Build GRCh38
rs753690197 Chr.9: 69820924-69820924 on Build GRCh38
rs746539745 Chr.9: 69821078-69821078 on Build GRCh38
rs753210256 Chr.9: 69821033-69821033 on Build GRCh38
rs529639038 Chr.9: 69821275-69821275 on Build GRCh38
rs755499542 Chr.9: 69820959-69820959 on Build GRCh38
rs747552571 Chr.9: 69820954-69820954 on Build GRCh38
rs750600894 Chr.9: 69820928-69820928 on Build GRCh38
rs747761453 Chr.9: 69821079-69821080 on Build GRCh38
rs781711958 Chr.9: 69820960-69820960 on Build GRCh38
rs778571754 Chr.9: 69820950-69820951 on Build GRCh38
rs779735236 Chr.9: 69821068-69821068 on Build GRCh38
rs144388278 Chr.9: 69821308-69821308 on Build GRCh38
rs540062790 Chr.9: 69821173-69821173 on Build GRCh38
rs1017287830 Chr.9: 69821351-69821351 on Build GRCh38
rs768452396 Chr.9: 69821096-69821096 on Build GRCh38
rs1016843339 Chr.9: 69821137-69821137 on Build GRCh38
rs780342434 Chr.9: 69821193-69821195 on Build GRCh38
rs766889158 Chr.9: 69820902-69820902 on Build GRCh38
rs1046347114 Chr.9: 69821034-69821034 on Build GRCh38
rs764088063 Chr.9: 69820916-69820916 on Build GRCh38
rs550689873 Chr.9: 69821307-69821307 on Build GRCh38
rs369486419 Chr.9: 69821362-69821362 on Build GRCh38
rs781582859 Chr.9: 69821028-69821028 on Build GRCh38
rs760218675 Chr.9: 69820979-69820979 on Build GRCh38
rs1042507745 Chr.9: 69821299-69821299 on Build GRCh38
rs547246746 Chr.9: 69821210-69821210 on Build GRCh38
rs562547982 Chr.9: 69821162-69821162 on Build GRCh38
rs946917212 Chr.9: 69821281-69821281 on Build GRCh38
rs185325139 Chr.9: 69821251-69821251 on Build GRCh38
rs779936251 Chr.9: 69820937-69820937 on Build GRCh38
rs370333172 Chr.9: 69820915-69820915 on Build GRCh38
rs934036543 Chr.9: 69821317-69821317 on Build GRCh38
rs994013943 Chr.9: 69821388-69821388 on Build GRCh38
rs867663881 Chr.9: 69820942-69820942 on Build GRCh38
rs758492391 Chr.9: 69820929-69820929 on Build GRCh38
rs373790286 Chr.9: 69821004-69821004 on Build GRCh38
rs560313115 Chr.9: 69821187-69821187 on Build GRCh38
rs757022096 Chr.9: 69820925-69820925 on Build GRCh38
rs150197009 Chr.9: 69820972-69820972 on Build GRCh38
rs971114415 Chr.9: 69821207-69821207 on Build GRCh38
rs990588843 Chr.9: 69821300-69821300 on Build GRCh38
rs772911183 Chr.9: 69821212-69821212 on Build GRCh38
rs981134195 Chr.9: 69821211-69821211 on Build GRCh38
rs928691039 Chr.9: 69821352-69821352 on Build GRCh38
rs761626492 Chr.9: 69820990-69820990 on Build GRCh38
rs766616336 Chr.9: 69821012-69821012 on Build GRCh38
rs750655220 Chr.9: 69820997-69820997 on Build GRCh38
rs926918409 Chr.9: 69821224-69821224 on Build GRCh38
rs994503826 Chr.9: 69821166-69821166 on Build GRCh38
rs138857531 Chr.9: 69820978-69820978 on Build GRCh38
rs148273758 Chr.9: 69820926-69820926 on Build GRCh38

View Full Product Details