Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 26213052-26213562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX3
Entrez Gene ID
11335
Gene Name
chromobox 3
Gene Aliases
HECH, HP1-GAMMA, HP1Hs-gamma
UniGene
Hs.381189
Gene Chromosome Location
Chr.7: 26201211-26213607 on Build GRCh38
Transcripts
NM_007276.4 NM_016587.3 XM_005249611.3

SNP Detail

SNP ID Chromosome Location
rs1038261658 Chr.7: 26213529-26213529 on Build GRCh38
rs541058585 Chr.7: 26213200-26213200 on Build GRCh38
rs913229770 Chr.7: 26213146-26213146 on Build GRCh38
rs766771132 Chr.7: 26213279-26213279 on Build GRCh38
rs7186 Chr.7: 26213182-26213182 on Build GRCh38
rs1042589781 Chr.7: 26213401-26213401 on Build GRCh38
rs1026570331 Chr.7: 26213486-26213486 on Build GRCh38
rs181300726 Chr.7: 26213365-26213365 on Build GRCh38
rs897083688 Chr.7: 26213530-26213530 on Build GRCh38
rs537161081 Chr.7: 26213512-26213512 on Build GRCh38
rs957594392 Chr.7: 26213113-26213113 on Build GRCh38
rs1024036095 Chr.7: 26213398-26213398 on Build GRCh38
rs754181630 Chr.7: 26213337-26213337 on Build GRCh38
rs923328061 Chr.7: 26213470-26213470 on Build GRCh38
rs142267744 Chr.7: 26213351-26213351 on Build GRCh38
rs530117129 Chr.7: 26213268-26213268 on Build GRCh38
rs754532052 Chr.7: 26213141-26213141 on Build GRCh38
rs546499173 Chr.7: 26213294-26213298 on Build GRCh38
rs893896515 Chr.7: 26213384-26213384 on Build GRCh38
rs369134863 Chr.7: 26213385-26213385 on Build GRCh38
rs747720619 Chr.7: 26213177-26213177 on Build GRCh38
rs889651306 Chr.7: 26213341-26213341 on Build GRCh38
rs868458558 Chr.7: 26213320-26213320 on Build GRCh38
rs1012580571 Chr.7: 26213463-26213463 on Build GRCh38
rs111891412 Chr.7: 26213448-26213448 on Build GRCh38
rs1034507887 Chr.7: 26213423-26213423 on Build GRCh38
rs780776847 Chr.7: 26213167-26213167 on Build GRCh38
rs766588991 Chr.7: 26213328-26213331 on Build GRCh38
rs749155788 Chr.7: 26213461-26213461 on Build GRCh38
rs563164402 Chr.7: 26213354-26213354 on Build GRCh38
rs559982933 Chr.7: 26213152-26213152 on Build GRCh38
rs375726042 Chr.7: 26213169-26213169 on Build GRCh38
rs1049538305 Chr.7: 26213211-26213211 on Build GRCh38
rs994251777 Chr.7: 26213373-26213376 on Build GRCh38
rs945407009 Chr.7: 26213392-26213392 on Build GRCh38
rs927467860 Chr.7: 26213079-26213079 on Build GRCh38
rs561094204 Chr.7: 26213212-26213212 on Build GRCh38
rs377589440 Chr.7: 26213315-26213317 on Build GRCh38
rs568198570 Chr.7: 26213494-26213494 on Build GRCh38
rs911018000 Chr.7: 26213513-26213513 on Build GRCh38
rs1040423264 Chr.7: 26213168-26213168 on Build GRCh38
rs547853175 Chr.7: 26213451-26213451 on Build GRCh38
rs552155077 Chr.7: 26213465-26213465 on Build GRCh38
rs967373176 Chr.7: 26213452-26213452 on Build GRCh38
rs572413925 Chr.7: 26213192-26213192 on Build GRCh38
rs28407029 Chr.7: 26213346-26213346 on Build GRCh38
rs13247806 Chr.7: 26213339-26213339 on Build GRCh38
rs990434915 Chr.7: 26213139-26213139 on Build GRCh38
rs752431269 Chr.7: 26213150-26213150 on Build GRCh38
rs565392435 Chr.7: 26213183-26213183 on Build GRCh38
rs78619479 Chr.7: 26213256-26213256 on Build GRCh38
rs941206348 Chr.7: 26213516-26213516 on Build GRCh38
rs1001313421 Chr.7: 26213410-26213410 on Build GRCh38
rs373262978 Chr.7: 26213357-26213357 on Build GRCh38
rs144437057 Chr.7: 26213313-26213315 on Build GRCh38
rs115829318 Chr.7: 26213449-26213449 on Build GRCh38

View Full Product Details