Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80081448-80081954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC40
Entrez Gene ID
55036
Gene Name
coiled-coil domain containing 40
Gene Aliases
CILD15, FAP172
UniGene
Hs.202542
Gene Chromosome Location
Chr.17: 80036632-80100613 on Build GRCh38
Transcripts
NM_001243342.1 NM_017950.3 XM_011524963.2 XM_011524964.2 XM_011524965.2 XM_017024807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979730 c.1705A>T p.T569S Chr.17: 80081688-80081688 on Build GRCh38
1290775 c.1743C>T p.S581S Chr.17: 80081726-80081726 on Build GRCh38
5990730 c.1637delA p.K547fs*9 Chr.17: 80081620-80081620 on Build GRCh38
1683273 c.1735_1736insA p.L579fs*95 Chr.17: 80081718-80081719 on Build GRCh38
7201978 c.1713G>T p.Q571H Chr.17: 80081696-80081696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765427963 Chr.17: 80081915-80081915 on Build GRCh38
rs771469312 Chr.17: 80081525-80081525 on Build GRCh38
rs754603683 Chr.17: 80081668-80081668 on Build GRCh38
rs764252103 Chr.17: 80081547-80081547 on Build GRCh38
rs560402881 Chr.17: 80081744-80081744 on Build GRCh38
rs765420679 Chr.17: 80081765-80081765 on Build GRCh38
rs201164121 Chr.17: 80081586-80081586 on Build GRCh38
rs1014600453 Chr.17: 80081790-80081790 on Build GRCh38
rs762004514 Chr.17: 80081648-80081648 on Build GRCh38
rs754036161 Chr.17: 80081555-80081555 on Build GRCh38
rs751321015 Chr.17: 80081802-80081802 on Build GRCh38
rs746661400 Chr.17: 80081576-80081576 on Build GRCh38
rs753336334 Chr.17: 80081913-80081913 on Build GRCh38
rs200272404 Chr.17: 80081557-80081557 on Build GRCh38
rs975736018 Chr.17: 80081542-80081542 on Build GRCh38
rs752363930 Chr.17: 80081805-80081805 on Build GRCh38
rs965506812 Chr.17: 80081783-80081783 on Build GRCh38
rs878855041 Chr.17: 80081603-80081603 on Build GRCh38
rs368624849 Chr.17: 80081627-80081627 on Build GRCh38
rs936824026 Chr.17: 80081577-80081577 on Build GRCh38
rs371383125 Chr.17: 80081893-80081893 on Build GRCh38
rs750134735 Chr.17: 80081573-80081573 on Build GRCh38
rs370805578 Chr.17: 80081528-80081528 on Build GRCh38
rs182104049 Chr.17: 80081797-80081797 on Build GRCh38
rs752972291 Chr.17: 80081531-80081531 on Build GRCh38
rs200236937 Chr.17: 80081653-80081653 on Build GRCh38
rs375215801 Chr.17: 80081864-80081864 on Build GRCh38
rs373899109 Chr.17: 80081529-80081529 on Build GRCh38
rs767093580 Chr.17: 80081804-80081804 on Build GRCh38
rs1056918232 Chr.17: 80081620-80081620 on Build GRCh38
rs768215507 Chr.17: 80081501-80081501 on Build GRCh38
rs769644960 Chr.17: 80081562-80081562 on Build GRCh38
rs761918200 Chr.17: 80081747-80081747 on Build GRCh38
rs747620444 Chr.17: 80081679-80081679 on Build GRCh38
rs769950790 Chr.17: 80081848-80081848 on Build GRCh38
rs748943374 Chr.17: 80081652-80081652 on Build GRCh38
rs36033394 Chr.17: 80081536-80081537 on Build GRCh38
rs756747733 Chr.17: 80081920-80081920 on Build GRCh38
rs199786478 Chr.17: 80081697-80081697 on Build GRCh38
rs775891024 Chr.17: 80081712-80081712 on Build GRCh38
rs773232601 Chr.17: 80081796-80081796 on Build GRCh38
rs571288423 Chr.17: 80081929-80081929 on Build GRCh38
rs202093340 Chr.17: 80081704-80081704 on Build GRCh38
rs777329186 Chr.17: 80081817-80081817 on Build GRCh38
rs759273764 Chr.17: 80081872-80081872 on Build GRCh38
rs377057189 Chr.17: 80081530-80081530 on Build GRCh38
rs370166011 Chr.17: 80081580-80081580 on Build GRCh38
rs747335744 Chr.17: 80081828-80081828 on Build GRCh38
rs755826267 Chr.17: 80081808-80081808 on Build GRCh38
rs1006029303 Chr.17: 80081754-80081755 on Build GRCh38
rs899000567 Chr.17: 80081910-80081910 on Build GRCh38
rs766278175 Chr.17: 80081798-80081798 on Build GRCh38
rs905231208 Chr.17: 80081692-80081692 on Build GRCh38
rs746460749 Chr.17: 80081499-80081499 on Build GRCh38
rs768950130 Chr.17: 80081738-80081738 on Build GRCh38
rs757226965 Chr.17: 80081558-80081558 on Build GRCh38
rs191736683 Chr.17: 80081656-80081656 on Build GRCh38
rs745789752 Chr.17: 80081516-80081516 on Build GRCh38
rs765193858 Chr.17: 80081893-80081895 on Build GRCh38
rs944228531 Chr.17: 80081492-80081492 on Build GRCh38
rs200815406 Chr.17: 80081722-80081722 on Build GRCh38
rs759757268 Chr.17: 80081615-80081615 on Build GRCh38
rs758243927 Chr.17: 80081574-80081574 on Build GRCh38
rs760610292 Chr.17: 80081539-80081539 on Build GRCh38
rs530585863 Chr.17: 80081794-80081794 on Build GRCh38
rs377745657 Chr.17: 80081868-80081868 on Build GRCh38
rs767290092 Chr.17: 80081883-80081883 on Build GRCh38
rs753942872 Chr.17: 80081645-80081645 on Build GRCh38
rs779234130 Chr.17: 80081703-80081703 on Build GRCh38
rs538748486 Chr.17: 80081470-80081470 on Build GRCh38
rs774956477 Chr.17: 80081599-80081599 on Build GRCh38
rs780848862 Chr.17: 80081670-80081670 on Build GRCh38
rs201481334 Chr.17: 80081663-80081663 on Build GRCh38
rs561596077 Chr.17: 80081632-80081632 on Build GRCh38
rs748367580 Chr.17: 80081844-80081844 on Build GRCh38
rs1034184126 Chr.17: 80081471-80081471 on Build GRCh38
rs534787961 Chr.17: 80081849-80081849 on Build GRCh38
rs772502492 Chr.17: 80081616-80081616 on Build GRCh38
rs763446302 Chr.17: 80081521-80081521 on Build GRCh38
rs948532763 Chr.17: 80081876-80081876 on Build GRCh38
rs754801803 Chr.17: 80081803-80081803 on Build GRCh38
rs763700391 Chr.17: 80081912-80081912 on Build GRCh38
rs864622323 Chr.17: 80081709-80081709 on Build GRCh38
rs768363871 Chr.17: 80081579-80081579 on Build GRCh38
rs1030441385 Chr.17: 80081544-80081544 on Build GRCh38
rs934397972 Chr.17: 80081619-80081619 on Build GRCh38
rs764448400 Chr.17: 80081639-80081639 on Build GRCh38
rs752551759 Chr.17: 80081892-80081892 on Build GRCh38
rs545193924 Chr.17: 80081515-80081515 on Build GRCh38
rs947035837 Chr.17: 80081548-80081548 on Build GRCh38
rs199791020 Chr.17: 80081837-80081837 on Build GRCh38
rs375662034 Chr.17: 80081590-80081590 on Build GRCh38
rs761619279 Chr.17: 80081554-80081554 on Build GRCh38
rs373309118 Chr.17: 80081829-80081829 on Build GRCh38
rs752125203 Chr.17: 80081930-80081930 on Build GRCh38
rs779793965 Chr.17: 80081593-80081593 on Build GRCh38
rs748784362 Chr.17: 80081824-80081824 on Build GRCh38
rs577764470 Chr.17: 80081825-80081825 on Build GRCh38
rs113887654 Chr.17: 80081836-80081836 on Build GRCh38
rs779895005 Chr.17: 80081662-80081662 on Build GRCh38
rs995164303 Chr.17: 80081684-80081684 on Build GRCh38
rs779040745 Chr.17: 80081571-80081571 on Build GRCh38
rs201559168 Chr.17: 80081705-80081705 on Build GRCh38
rs887646328 Chr.17: 80081477-80081477 on Build GRCh38
rs369273325 Chr.17: 80081866-80081866 on Build GRCh38

View Full Product Details