Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1783465-1783953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
UniGene
Hs.127675
Gene Chromosome Location
Chr.8: 1755778-1786570 on Build GRCh38
Transcripts
NM_018941.3

SNP Detail

SNP ID Chromosome Location
rs767990670 Chr.8: 1783685-1783685 on Build GRCh38
rs981240824 Chr.8: 1783665-1783665 on Build GRCh38
rs934637419 Chr.8: 1783561-1783561 on Build GRCh38
rs953106236 Chr.8: 1783828-1783828 on Build GRCh38
rs970571822 Chr.8: 1783765-1783765 on Build GRCh38
rs1027651421 Chr.8: 1783628-1783628 on Build GRCh38
rs972975055 Chr.8: 1783624-1783624 on Build GRCh38
rs546107642 Chr.8: 1783811-1783811 on Build GRCh38
rs1005824721 Chr.8: 1783845-1783845 on Build GRCh38
rs1039850687 Chr.8: 1783831-1783831 on Build GRCh38
rs544100482 Chr.8: 1783758-1783758 on Build GRCh38
rs376610687 Chr.8: 1783497-1783497 on Build GRCh38
rs569798206 Chr.8: 1783705-1783706 on Build GRCh38
rs947354950 Chr.8: 1783742-1783742 on Build GRCh38
rs956856644 Chr.8: 1783810-1783810 on Build GRCh38
rs961921443 Chr.8: 1783623-1783623 on Build GRCh38
rs534991695 Chr.8: 1783627-1783627 on Build GRCh38
rs1016361683 Chr.8: 1783723-1783723 on Build GRCh38
rs916427906 Chr.8: 1783513-1783513 on Build GRCh38
rs1003356287 Chr.8: 1783933-1783933 on Build GRCh38
rs923933463 Chr.8: 1783589-1783589 on Build GRCh38
rs1053003040 Chr.8: 1783724-1783724 on Build GRCh38
rs934205567 Chr.8: 1783852-1783852 on Build GRCh38
rs1004575146 Chr.8: 1783698-1783698 on Build GRCh38
rs528187531 Chr.8: 1783861-1783861 on Build GRCh38
rs963162286 Chr.8: 1783495-1783495 on Build GRCh38
rs915649129 Chr.8: 1783649-1783649 on Build GRCh38
rs759897529 Chr.8: 1783648-1783648 on Build GRCh38
rs935378291 Chr.8: 1783605-1783605 on Build GRCh38
rs563222208 Chr.8: 1783606-1783606 on Build GRCh38
rs901344829 Chr.8: 1783654-1783654 on Build GRCh38
rs922758465 Chr.8: 1783850-1783850 on Build GRCh38
rs994761289 Chr.8: 1783813-1783813 on Build GRCh38
rs550844334 Chr.8: 1783889-1783889 on Build GRCh38
rs1023373893 Chr.8: 1783912-1783912 on Build GRCh38
rs1054263479 Chr.8: 1783608-1783608 on Build GRCh38
rs1019831957 Chr.8: 1783621-1783621 on Build GRCh38
rs942724751 Chr.8: 1783819-1783819 on Build GRCh38
rs998436783 Chr.8: 1783657-1783657 on Build GRCh38
rs575145815 Chr.8: 1783717-1783717 on Build GRCh38
rs1049779027 Chr.8: 1783753-1783753 on Build GRCh38
rs942777140 Chr.8: 1783616-1783616 on Build GRCh38
rs756975763 Chr.8: 1783725-1783725 on Build GRCh38
rs915255110 Chr.8: 1783814-1783814 on Build GRCh38
rs898837758 Chr.8: 1783883-1783883 on Build GRCh38
rs572603137 Chr.8: 1783541-1783541 on Build GRCh38
rs368257540 Chr.8: 1783878-1783878 on Build GRCh38
rs1011996314 Chr.8: 1783600-1783600 on Build GRCh38
rs765296138 Chr.8: 1783754-1783754 on Build GRCh38
rs928523946 Chr.8: 1783670-1783670 on Build GRCh38
rs970584859 Chr.8: 1783542-1783542 on Build GRCh38
rs532868505 Chr.8: 1783856-1783856 on Build GRCh38
rs893104920 Chr.8: 1783593-1783593 on Build GRCh38
rs778930314 Chr.8: 1783621-1783621 on Build GRCh38
rs1011959537 Chr.8: 1783757-1783757 on Build GRCh38
rs10089616 Chr.8: 1783689-1783689 on Build GRCh38
rs149877204 Chr.8: 1783801-1783801 on Build GRCh38
rs754718571 Chr.8: 1783863-1783863 on Build GRCh38
rs893055103 Chr.8: 1783737-1783737 on Build GRCh38
rs868843558 Chr.8: 1783514-1783514 on Build GRCh38
rs558996077 Chr.8: 1783853-1783853 on Build GRCh38
rs547729355 Chr.8: 1783922-1783922 on Build GRCh38
rs193228611 Chr.8: 1783767-1783767 on Build GRCh38
rs547740290 Chr.8: 1783877-1783877 on Build GRCh38
rs775958260 Chr.8: 1783787-1783787 on Build GRCh38
rs747012744 Chr.8: 1783846-1783846 on Build GRCh38
rs774641073 Chr.8: 1783496-1783496 on Build GRCh38
rs982052276 Chr.8: 1783544-1783544 on Build GRCh38
rs915681787 Chr.8: 1783511-1783511 on Build GRCh38
rs758353313 Chr.8: 1783781-1783781 on Build GRCh38
rs951722728 Chr.8: 1783646-1783646 on Build GRCh38
rs948497063 Chr.8: 1783653-1783653 on Build GRCh38
rs984465108 Chr.8: 1783506-1783506 on Build GRCh38
rs575341792 Chr.8: 1783610-1783610 on Build GRCh38
rs541925096 Chr.8: 1783582-1783582 on Build GRCh38
rs1047344081 Chr.8: 1783688-1783688 on Build GRCh38
rs530468873 Chr.8: 1783886-1783886 on Build GRCh38

View Full Product Details