Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100053969-100054464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPN1
Entrez Gene ID
1369
Gene Name
carboxypeptidase N subunit 1
Gene Aliases
CPN, SCPN
UniGene
Hs.2246
Gene Chromosome Location
Chr.10: 100042308-100081885 on Build GRCh38
Transcripts
NM_001308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6064858 c.1083C>A p.V361V Chr.10: 100054375-100054375 on Build GRCh38
325056 c.1030G>A p.G344R Chr.10: 100054428-100054428 on Build GRCh38
913867 c.1022G>A p.G341D Chr.10: 100054436-100054436 on Build GRCh38
6064859 c.1096C>G p.H366D Chr.10: 100054362-100054362 on Build GRCh38
3433505 c.1031G>A p.G344E Chr.10: 100054427-100054427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773195465 Chr.10: 100054157-100054157 on Build GRCh38
rs1038837742 Chr.10: 100054130-100054130 on Build GRCh38
rs902145298 Chr.10: 100054220-100054220 on Build GRCh38
rs941736061 Chr.10: 100054362-100054362 on Build GRCh38
rs141953664 Chr.10: 100054348-100054348 on Build GRCh38
rs150478398 Chr.10: 100054277-100054277 on Build GRCh38
rs538507253 Chr.10: 100054006-100054006 on Build GRCh38
rs921679127 Chr.10: 100054193-100054193 on Build GRCh38
rs553772101 Chr.10: 100054053-100054053 on Build GRCh38
rs779039367 Chr.10: 100054329-100054329 on Build GRCh38
rs753917948 Chr.10: 100054400-100054400 on Build GRCh38
rs771923245 Chr.10: 100054395-100054395 on Build GRCh38
rs543628582 Chr.10: 100054224-100054224 on Build GRCh38
rs748482664 Chr.10: 100054340-100054340 on Build GRCh38
rs932998993 Chr.10: 100054200-100054200 on Build GRCh38
rs765438662 Chr.10: 100054405-100054405 on Build GRCh38
rs557494565 Chr.10: 100054163-100054163 on Build GRCh38
rs917799539 Chr.10: 100054251-100054251 on Build GRCh38
rs1008207262 Chr.10: 100054068-100054068 on Build GRCh38
rs1037372375 Chr.10: 100054386-100054386 on Build GRCh38
rs778406963 Chr.10: 100054344-100054344 on Build GRCh38
rs758828408 Chr.10: 100054437-100054437 on Build GRCh38
rs1037444505 Chr.10: 100054025-100054025 on Build GRCh38
rs941769065 Chr.10: 100054020-100054020 on Build GRCh38
rs759543168 Chr.10: 100054403-100054403 on Build GRCh38
rs144489904 Chr.10: 100054123-100054123 on Build GRCh38
rs1051851418 Chr.10: 100054204-100054204 on Build GRCh38
rs762872157 Chr.10: 100054143-100054143 on Build GRCh38
rs769394039 Chr.10: 100054356-100054356 on Build GRCh38
rs949351881 Chr.10: 100054285-100054285 on Build GRCh38
rs1018269231 Chr.10: 100054115-100054115 on Build GRCh38
rs199635696 Chr.10: 100054396-100054396 on Build GRCh38
rs1026084213 Chr.10: 100054424-100054424 on Build GRCh38
rs748769668 Chr.10: 100054375-100054375 on Build GRCh38
rs368479939 Chr.10: 100054342-100054342 on Build GRCh38
rs527749314 Chr.10: 100054430-100054430 on Build GRCh38
rs1012026648 Chr.10: 100054347-100054347 on Build GRCh38
rs979503249 Chr.10: 100054154-100054154 on Build GRCh38
rs773878916 Chr.10: 100054391-100054391 on Build GRCh38
rs561817810 Chr.10: 100054226-100054226 on Build GRCh38
rs535091354 Chr.10: 100054046-100054046 on Build GRCh38
rs571805071 Chr.10: 100054058-100054058 on Build GRCh38
rs142351661 Chr.10: 100054009-100054009 on Build GRCh38
rs7084921 Chr.10: 100054045-100054045 on Build GRCh38
rs768313764 Chr.10: 100054384-100054384 on Build GRCh38
rs779662092 Chr.10: 100054361-100054361 on Build GRCh38
rs930234529 Chr.10: 100054042-100054042 on Build GRCh38
rs761514644 Chr.10: 100054117-100054117 on Build GRCh38
rs183581582 Chr.10: 100054172-100054172 on Build GRCh38
rs111804056 Chr.10: 100054228-100054228 on Build GRCh38

View Full Product Details