Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73836374-73836879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL6
Entrez Gene ID
6372
Gene Name
C-X-C motif chemokine ligand 6
Gene Aliases
CKA-3, GCP-2, GCP2, SCYB6
UniGene
Hs.164021
Gene Chromosome Location
Chr.4: 73836556-73838760 on Build GRCh38
Transcripts
NM_002993.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5784291 c.23C>A p.A8E Chr.4: 73836773-73836773 on Build GRCh38
6638453 c.65T>A p.L22H Chr.4: 73836815-73836815 on Build GRCh38
2831365 c.103G>A p.A35T Chr.4: 73836853-73836853 on Build GRCh38
6276606 c.109+1G>T p.? Chr.4: 73836860-73836860 on Build GRCh38
3826186 c.44C>T p.S15L Chr.4: 73836794-73836794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535506985 Chr.4: 73836424-73836424 on Build GRCh38
rs193257415 Chr.4: 73836581-73836581 on Build GRCh38
rs747748225 Chr.4: 73836844-73836844 on Build GRCh38
rs764229919 Chr.4: 73836804-73836804 on Build GRCh38
rs146902260 Chr.4: 73836855-73836855 on Build GRCh38
rs1008711538 Chr.4: 73836438-73836438 on Build GRCh38
rs79143912 Chr.4: 73836402-73836402 on Build GRCh38
rs575629842 Chr.4: 73836669-73836669 on Build GRCh38
rs1033662949 Chr.4: 73836542-73836542 on Build GRCh38
rs751664645 Chr.4: 73836809-73836809 on Build GRCh38
rs555237920 Chr.4: 73836666-73836666 on Build GRCh38
rs772873323 Chr.4: 73836791-73836791 on Build GRCh38
rs948467139 Chr.4: 73836527-73836527 on Build GRCh38
rs1056678399 Chr.4: 73836611-73836611 on Build GRCh38
rs777998565 Chr.4: 73836698-73836698 on Build GRCh38
rs559646591 Chr.4: 73836677-73836677 on Build GRCh38
rs961294275 Chr.4: 73836807-73836807 on Build GRCh38
rs955211392 Chr.4: 73836461-73836461 on Build GRCh38
rs778333716 Chr.4: 73836842-73836842 on Build GRCh38
rs967673021 Chr.4: 73836486-73836486 on Build GRCh38
rs879091251 Chr.4: 73836519-73836519 on Build GRCh38
rs748863270 Chr.4: 73836701-73836701 on Build GRCh38
rs752065844 Chr.4: 73836771-73836771 on Build GRCh38
rs527824063 Chr.4: 73836778-73836778 on Build GRCh38
rs992330889 Chr.4: 73836602-73836602 on Build GRCh38
rs775852013 Chr.4: 73836807-73836815 on Build GRCh38
rs200901978 Chr.4: 73836846-73836846 on Build GRCh38
rs543723730 Chr.4: 73836643-73836643 on Build GRCh38
rs948340096 Chr.4: 73836395-73836395 on Build GRCh38
rs944155629 Chr.4: 73836788-73836788 on Build GRCh38
rs774318977 Chr.4: 73836704-73836704 on Build GRCh38
rs960762460 Chr.4: 73836587-73836587 on Build GRCh38
rs999906122 Chr.4: 73836534-73836534 on Build GRCh38
rs112902654 Chr.4: 73836545-73836545 on Build GRCh38
rs968137817 Chr.4: 73836425-73836425 on Build GRCh38
rs1047111215 Chr.4: 73836429-73836429 on Build GRCh38
rs908458621 Chr.4: 73836479-73836479 on Build GRCh38
rs959017138 Chr.4: 73836547-73836547 on Build GRCh38
rs773879713 Chr.4: 73836784-73836784 on Build GRCh38
rs964617518 Chr.4: 73836465-73836465 on Build GRCh38
rs775906428 Chr.4: 73836801-73836801 on Build GRCh38
rs754627748 Chr.4: 73836840-73836840 on Build GRCh38
rs765528134 Chr.4: 73836800-73836800 on Build GRCh38
rs892706191 Chr.4: 73836629-73836629 on Build GRCh38
rs772055314 Chr.4: 73836786-73836786 on Build GRCh38
rs1022227121 Chr.4: 73836485-73836485 on Build GRCh38
rs553888501 Chr.4: 73836434-73836434 on Build GRCh38
rs200457897 Chr.4: 73836852-73836852 on Build GRCh38
rs767173197 Chr.4: 73836724-73836724 on Build GRCh38
rs377412241 Chr.4: 73836837-73836837 on Build GRCh38
rs17838982 Chr.4: 73836546-73836546 on Build GRCh38
rs1022743059 Chr.4: 73836423-73836423 on Build GRCh38
rs903860672 Chr.4: 73836667-73836667 on Build GRCh38
rs916785910 Chr.4: 73836617-73836617 on Build GRCh38
rs1010254074 Chr.4: 73836654-73836654 on Build GRCh38
rs761829624 Chr.4: 73836714-73836714 on Build GRCh38
rs929968094 Chr.4: 73836761-73836761 on Build GRCh38
rs759614893 Chr.4: 73836735-73836735 on Build GRCh38
rs763292477 Chr.4: 73836802-73836802 on Build GRCh38
rs576210432 Chr.4: 73836601-73836601 on Build GRCh38
rs768136002 Chr.4: 73836816-73836816 on Build GRCh38
rs747964724 Chr.4: 73836785-73836785 on Build GRCh38
rs919602852 Chr.4: 73836503-73836503 on Build GRCh38
rs994134605 Chr.4: 73836416-73836416 on Build GRCh38
rs765557722 Chr.4: 73836746-73836746 on Build GRCh38
rs879022227 Chr.4: 73836748-73836748 on Build GRCh38
rs1019224704 Chr.4: 73836447-73836447 on Build GRCh38
rs370080580 Chr.4: 73836795-73836795 on Build GRCh38
rs752824015 Chr.4: 73836751-73836751 on Build GRCh38
rs919566678 Chr.4: 73836834-73836835 on Build GRCh38
rs756549378 Chr.4: 73836821-73836821 on Build GRCh38
rs373291221 Chr.4: 73836810-73836810 on Build GRCh38
rs764679144 Chr.4: 73836768-73836768 on Build GRCh38
rs1951243 Chr.4: 73836413-73836413 on Build GRCh38
rs769013830 Chr.4: 73836857-73836857 on Build GRCh38
rs781345554 Chr.4: 73836772-73836772 on Build GRCh38
rs748771639 Chr.4: 73836781-73836781 on Build GRCh38
rs1022267378 Chr.4: 73836660-73836660 on Build GRCh38
rs770527395 Chr.4: 73836759-73836759 on Build GRCh38
rs375976441 Chr.4: 73836774-73836774 on Build GRCh38
rs996021234 Chr.4: 73836824-73836824 on Build GRCh38
rs746625197 Chr.4: 73836713-73836713 on Build GRCh38
rs966980062 Chr.4: 73836524-73836524 on Build GRCh38
rs749534611 Chr.4: 73836773-73836773 on Build GRCh38
rs996136385 Chr.4: 73836466-73836466 on Build GRCh38
rs1003761336 Chr.4: 73836397-73836397 on Build GRCh38
rs773061109 Chr.4: 73836731-73836731 on Build GRCh38
rs978175378 Chr.4: 73836487-73836487 on Build GRCh38
rs770802849 Chr.4: 73836853-73836853 on Build GRCh38
rs939969375 Chr.4: 73836480-73836480 on Build GRCh38
rs539437116 Chr.4: 73836579-73836579 on Build GRCh38
rs185031743 Chr.4: 73836764-73836764 on Build GRCh38
rs774289824 Chr.4: 73836859-73836859 on Build GRCh38
rs1040206395 Chr.4: 73836806-73836806 on Build GRCh38
rs768548132 Chr.4: 73836703-73836703 on Build GRCh38
rs565807756 Chr.4: 73836467-73836467 on Build GRCh38
rs568657784 Chr.4: 73836420-73836420 on Build GRCh38
rs776601097 Chr.4: 73836854-73836854 on Build GRCh38
rs768103843 Chr.4: 73836782-73836782 on Build GRCh38
rs532039148 Chr.4: 73836411-73836411 on Build GRCh38

View Full Product Details