Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42873868-42874413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP8B1
Entrez Gene ID
1582
Gene Name
cytochrome P450 family 8 subfamily B member 1
Gene Aliases
CP8B, CYP12
UniGene
Hs.447793
Gene Chromosome Location
Chr.3: 42872192-42876141 on Build GRCh38
Transcripts
NM_004391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1423150 c.1435C>T p.R479C Chr.3: 42874382-42874382 on Build GRCh38
145299 c.1456C>T p.Q486* Chr.3: 42874361-42874361 on Build GRCh38
4117664 c.1480C>T p.R494C Chr.3: 42874337-42874337 on Build GRCh38
5810859 c.1407C>T p.D469D Chr.3: 42874410-42874410 on Build GRCh38
1202907 c.1487G>A p.R496H Chr.3: 42874330-42874330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777422325 Chr.3: 42874271-42874271 on Build GRCh38
rs555376774 Chr.3: 42874181-42874181 on Build GRCh38
rs1029203861 Chr.3: 42873910-42873910 on Build GRCh38
rs991052018 Chr.3: 42874060-42874060 on Build GRCh38
rs543657145 Chr.3: 42874342-42874342 on Build GRCh38
rs1033075717 Chr.3: 42873986-42873986 on Build GRCh38
rs1020270681 Chr.3: 42873936-42873936 on Build GRCh38
rs772602532 Chr.3: 42874318-42874318 on Build GRCh38
rs565601077 Chr.3: 42874229-42874229 on Build GRCh38
rs889029866 Chr.3: 42873991-42873991 on Build GRCh38
rs35013899 Chr.3: 42874242-42874243 on Build GRCh38
rs372472190 Chr.3: 42874354-42874354 on Build GRCh38
rs1016086925 Chr.3: 42874022-42874022 on Build GRCh38
rs748292508 Chr.3: 42874305-42874305 on Build GRCh38
rs556582199 Chr.3: 42874055-42874055 on Build GRCh38
rs267599827 Chr.3: 42874349-42874349 on Build GRCh38
rs890696825 Chr.3: 42873913-42873913 on Build GRCh38
rs1033043019 Chr.3: 42874337-42874337 on Build GRCh38
rs940351051 Chr.3: 42874106-42874106 on Build GRCh38
rs774791142 Chr.3: 42874330-42874330 on Build GRCh38
rs542498923 Chr.3: 42874155-42874155 on Build GRCh38
rs769947294 Chr.3: 42874308-42874308 on Build GRCh38
rs141477865 Chr.3: 42873925-42873925 on Build GRCh38
rs761146146 Chr.3: 42874343-42874343 on Build GRCh38
rs571068760 Chr.3: 42873972-42873972 on Build GRCh38
rs1009153474 Chr.3: 42873924-42873924 on Build GRCh38
rs1050988231 Chr.3: 42874230-42874230 on Build GRCh38
rs150532676 Chr.3: 42874314-42874314 on Build GRCh38
rs561432475 Chr.3: 42874275-42874275 on Build GRCh38
rs572187445 Chr.3: 42874247-42874247 on Build GRCh38
rs761111073 Chr.3: 42874372-42874372 on Build GRCh38
rs563905678 Chr.3: 42874285-42874285 on Build GRCh38
rs933369618 Chr.3: 42874213-42874213 on Build GRCh38
rs376808261 Chr.3: 42874347-42874347 on Build GRCh38
rs534735240 Chr.3: 42873931-42873931 on Build GRCh38
rs993234194 Chr.3: 42874061-42874061 on Build GRCh38
rs1026409858 Chr.3: 42874063-42874063 on Build GRCh38
rs368285524 Chr.3: 42874329-42874329 on Build GRCh38
rs1052491236 Chr.3: 42874188-42874189 on Build GRCh38
rs760124342 Chr.3: 42874357-42874364 on Build GRCh38
rs79930470 Chr.3: 42873963-42873963 on Build GRCh38
rs112111925 Chr.3: 42874093-42874093 on Build GRCh38
rs370752214 Chr.3: 42874356-42874356 on Build GRCh38
rs753535558 Chr.3: 42874272-42874272 on Build GRCh38
rs776931947 Chr.3: 42874387-42874387 on Build GRCh38
rs931786658 Chr.3: 42874194-42874194 on Build GRCh38
rs201871190 Chr.3: 42874090-42874090 on Build GRCh38
rs745387448 Chr.3: 42874100-42874100 on Build GRCh38
rs746971123 Chr.3: 42874363-42874363 on Build GRCh38
rs913972115 Chr.3: 42874201-42874201 on Build GRCh38
rs889062370 Chr.3: 42874248-42874248 on Build GRCh38
rs547979461 Chr.3: 42873937-42873937 on Build GRCh38
rs138532585 Chr.3: 42874358-42874358 on Build GRCh38
rs949289121 Chr.3: 42874092-42874092 on Build GRCh38
rs1007428881 Chr.3: 42873999-42873999 on Build GRCh38
rs754959169 Chr.3: 42874364-42874364 on Build GRCh38
rs963477167 Chr.3: 42874057-42874057 on Build GRCh38
rs114153667 Chr.3: 42874149-42874149 on Build GRCh38
rs992903188 Chr.3: 42874078-42874078 on Build GRCh38
rs959696265 Chr.3: 42874064-42874064 on Build GRCh38
rs145159025 Chr.3: 42874336-42874336 on Build GRCh38
rs541149108 Chr.3: 42874283-42874283 on Build GRCh38
rs780818431 Chr.3: 42874001-42874001 on Build GRCh38
rs735320 Chr.3: 42874386-42874386 on Build GRCh38
rs6782601 Chr.3: 42873893-42873893 on Build GRCh38
rs372447955 Chr.3: 42874331-42874331 on Build GRCh38
rs79670156 Chr.3: 42873983-42873983 on Build GRCh38
rs201949466 Chr.3: 42874350-42874350 on Build GRCh38
rs749318881 Chr.3: 42874378-42874378 on Build GRCh38
rs1007459738 Chr.3: 42874288-42874288 on Build GRCh38
rs528553610 Chr.3: 42874302-42874302 on Build GRCh38
rs925424321 Chr.3: 42874146-42874146 on Build GRCh38
rs778750576 Chr.3: 42874277-42874277 on Build GRCh38
rs771278880 Chr.3: 42874382-42874382 on Build GRCh38
rs1051754627 Chr.3: 42874226-42874226 on Build GRCh38
rs144198819 Chr.3: 42874353-42874353 on Build GRCh38

View Full Product Details