Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2520385-2520936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG7
Entrez Gene ID
2788
Gene Name
G protein subunit gamma 7
Gene Aliases
-
UniGene
Hs.515544
Gene Chromosome Location
Chr.19: 2511219-2702748 on Build GRCh38
Transcripts
NM_052847.2 XM_017026606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1392267 c.53G>A p.R18H Chr.19: 2520636-2520636 on Build GRCh38
6229416 c.74G>A p.R25H Chr.19: 2520615-2520615 on Build GRCh38
1392268 c.1A>G p.M1V Chr.19: 2520688-2520688 on Build GRCh38
255869 c.28G>A p.A10T Chr.19: 2520661-2520661 on Build GRCh38
4645980 c.17A>G p.N6S Chr.19: 2520672-2520672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914909964 Chr.19: 2520634-2520634 on Build GRCh38
rs369990544 Chr.19: 2520681-2520681 on Build GRCh38
rs576755199 Chr.19: 2520790-2520790 on Build GRCh38
rs936756315 Chr.19: 2520579-2520579 on Build GRCh38
rs997741596 Chr.19: 2520897-2520897 on Build GRCh38
rs532716854 Chr.19: 2520902-2520902 on Build GRCh38
rs778000559 Chr.19: 2520713-2520713 on Build GRCh38
rs922387990 Chr.19: 2520413-2520413 on Build GRCh38
rs772815638 Chr.19: 2520590-2520590 on Build GRCh38
rs751743205 Chr.19: 2520628-2520628 on Build GRCh38
rs369190563 Chr.19: 2520580-2520580 on Build GRCh38
rs573215355 Chr.19: 2520733-2520733 on Build GRCh38
rs748944977 Chr.19: 2520609-2520609 on Build GRCh38
rs891956987 Chr.19: 2520903-2520903 on Build GRCh38
rs1032454062 Chr.19: 2520809-2520809 on Build GRCh38
rs551027417 Chr.19: 2520633-2520633 on Build GRCh38
rs746935907 Chr.19: 2520557-2520557 on Build GRCh38
rs572018660 Chr.19: 2520565-2520565 on Build GRCh38
rs118126180 Chr.19: 2520584-2520584 on Build GRCh38
rs753275850 Chr.19: 2520658-2520658 on Build GRCh38
rs766472325 Chr.19: 2520597-2520597 on Build GRCh38
rs865952383 Chr.19: 2520636-2520636 on Build GRCh38
rs956433318 Chr.19: 2520808-2520808 on Build GRCh38
rs968116201 Chr.19: 2520817-2520817 on Build GRCh38
rs916569690 Chr.19: 2520591-2520591 on Build GRCh38
rs1004656682 Chr.19: 2520453-2520453 on Build GRCh38
rs200094859 Chr.19: 2520737-2520737 on Build GRCh38
rs1020251523 Chr.19: 2520744-2520744 on Build GRCh38
rs987429863 Chr.19: 2520505-2520508 on Build GRCh38
rs565465766 Chr.19: 2520865-2520865 on Build GRCh38
rs980487515 Chr.19: 2520825-2520825 on Build GRCh38
rs185432826 Chr.19: 2520556-2520556 on Build GRCh38
rs369491069 Chr.19: 2520626-2520626 on Build GRCh38
rs3764583 Chr.19: 2520474-2520474 on Build GRCh38
rs376510778 Chr.19: 2520592-2520592 on Build GRCh38
rs562489533 Chr.19: 2520484-2520484 on Build GRCh38
rs574377485 Chr.19: 2520814-2520814 on Build GRCh38
rs981088498 Chr.19: 2520710-2520710 on Build GRCh38
rs779717170 Chr.19: 2520636-2520637 on Build GRCh38
rs907705707 Chr.19: 2520656-2520656 on Build GRCh38
rs982062048 Chr.19: 2520850-2520850 on Build GRCh38
rs943345348 Chr.19: 2520879-2520879 on Build GRCh38
rs3764585 Chr.19: 2520793-2520793 on Build GRCh38
rs987788748 Chr.19: 2520461-2520461 on Build GRCh38
rs779999497 Chr.19: 2520409-2520409 on Build GRCh38
rs764011188 Chr.19: 2520881-2520881 on Build GRCh38
rs886245364 Chr.19: 2520434-2520434 on Build GRCh38
rs536221370 Chr.19: 2520719-2520719 on Build GRCh38
rs1009274205 Chr.19: 2520811-2520811 on Build GRCh38
rs747489287 Chr.19: 2520458-2520458 on Build GRCh38
rs181346972 Chr.19: 2520527-2520527 on Build GRCh38
rs1056342963 Chr.19: 2520730-2520730 on Build GRCh38
rs868531287 Chr.19: 2520640-2520640 on Build GRCh38
rs1022079214 Chr.19: 2520813-2520813 on Build GRCh38
rs956483357 Chr.19: 2520619-2520619 on Build GRCh38
rs181918274 Chr.19: 2520827-2520827 on Build GRCh38
rs902074093 Chr.19: 2520882-2520882 on Build GRCh38
rs1027672954 Chr.19: 2520561-2520561 on Build GRCh38
rs1044881961 Chr.19: 2520497-2520497 on Build GRCh38
rs1015983823 Chr.19: 2520457-2520457 on Build GRCh38
rs544117198 Chr.19: 2520791-2520791 on Build GRCh38
rs1031812673 Chr.19: 2520807-2520807 on Build GRCh38
rs188061507 Chr.19: 2520504-2520504 on Build GRCh38
rs915148848 Chr.19: 2520466-2520466 on Build GRCh38
rs758704103 Chr.19: 2520913-2520913 on Build GRCh38
rs189944029 Chr.19: 2520732-2520732 on Build GRCh38
rs1133692 Chr.19: 2520718-2520718 on Build GRCh38
rs559946090 Chr.19: 2520534-2520534 on Build GRCh38
rs759515987 Chr.19: 2520775-2520775 on Build GRCh38
rs558326245 Chr.19: 2520770-2520770 on Build GRCh38
rs929396230 Chr.19: 2520734-2520734 on Build GRCh38
rs186619477 Chr.19: 2520906-2520906 on Build GRCh38
rs760497239 Chr.19: 2520828-2520828 on Build GRCh38
rs779517125 Chr.19: 2520721-2520721 on Build GRCh38
rs887780996 Chr.19: 2520760-2520760 on Build GRCh38
rs541957987 Chr.19: 2520826-2520826 on Build GRCh38
rs898841493 Chr.19: 2520464-2520464 on Build GRCh38
rs967957523 Chr.19: 2520702-2520702 on Build GRCh38
rs139249433 Chr.19: 2520637-2520637 on Build GRCh38
rs61603373 Chr.19: 2520912-2520912 on Build GRCh38
rs763152930 Chr.19: 2520596-2520596 on Build GRCh38
rs529625736 Chr.19: 2520526-2520526 on Build GRCh38
rs997625935 Chr.19: 2520801-2520801 on Build GRCh38
rs547244865 Chr.19: 2520861-2520861 on Build GRCh38
rs909336548 Chr.19: 2520878-2520878 on Build GRCh38
rs1050130841 Chr.19: 2520910-2520910 on Build GRCh38
rs919097105 Chr.19: 2520860-2520860 on Build GRCh38
rs768100642 Chr.19: 2520657-2520657 on Build GRCh38
rs142838913 Chr.19: 2520855-2520855 on Build GRCh38
rs201004439 Chr.19: 2520558-2520558 on Build GRCh38
rs558894525 Chr.19: 2520512-2520512 on Build GRCh38

View Full Product Details