Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 981947-982438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PERM1
Entrez Gene ID
84808
Gene Name
PPARGC1 and ESRR induced regulator, muscle 1
Gene Aliases
C1orf170
UniGene
Hs.271462
Gene Chromosome Location
Chr.1: 975198-982117 on Build GRCh38
Transcripts
NM_001291366.1 NM_001291367.1 XM_017002583.1 XM_017002584.1

SNP Detail

SNP ID Chromosome Location
rs995944178 Chr.1: 982198-982198 on Build GRCh38
rs550252786 Chr.1: 982183-982183 on Build GRCh38
rs762948675 Chr.1: 982415-982415 on Build GRCh38
rs978421586 Chr.1: 982281-982281 on Build GRCh38
rs947045661 Chr.1: 982107-982107 on Build GRCh38
rs775507284 Chr.1: 982116-982116 on Build GRCh38
rs1006717108 Chr.1: 982215-982215 on Build GRCh38
rs529381225 Chr.1: 982169-982169 on Build GRCh38
rs375345061 Chr.1: 982275-982275 on Build GRCh38
rs925525613 Chr.1: 982289-982289 on Build GRCh38
rs866121447 Chr.1: 982364-982364 on Build GRCh38
rs552688509 Chr.1: 982347-982347 on Build GRCh38
rs1008658116 Chr.1: 982101-982101 on Build GRCh38
rs995641434 Chr.1: 982077-982077 on Build GRCh38
rs968901331 Chr.1: 982392-982392 on Build GRCh38
rs543249182 Chr.1: 982207-982207 on Build GRCh38
rs967858758 Chr.1: 981967-981967 on Build GRCh38
rs1048456902 Chr.1: 982361-982361 on Build GRCh38
rs541007519 Chr.1: 982053-982053 on Build GRCh38
rs978493845 Chr.1: 982393-982393 on Build GRCh38
rs761050770 Chr.1: 982073-982073 on Build GRCh38
rs765373771 Chr.1: 982102-982102 on Build GRCh38
rs372846343 Chr.1: 982303-982303 on Build GRCh38
rs746183744 Chr.1: 982010-982010 on Build GRCh38
rs985332962 Chr.1: 982061-982061 on Build GRCh38
rs777163408 Chr.1: 982089-982089 on Build GRCh38
rs939972763 Chr.1: 982192-982192 on Build GRCh38
rs1019546602 Chr.1: 982365-982365 on Build GRCh38
rs1013646089 Chr.1: 982150-982150 on Build GRCh38
rs1026909476 Chr.1: 982042-982042 on Build GRCh38
rs6658898 Chr.1: 982382-982382 on Build GRCh38
rs1036835594 Chr.1: 982066-982066 on Build GRCh38
rs891798530 Chr.1: 982306-982306 on Build GRCh38
rs140127909 Chr.1: 982416-982416 on Build GRCh38
rs751529225 Chr.1: 982074-982074 on Build GRCh38
rs867433972 Chr.1: 982114-982114 on Build GRCh38
rs151100411 Chr.1: 982021-982021 on Build GRCh38
rs1033722242 Chr.1: 981973-981973 on Build GRCh38
rs555114511 Chr.1: 981966-981966 on Build GRCh38
rs571886408 Chr.1: 982276-982276 on Build GRCh38
rs961692539 Chr.1: 982012-982012 on Build GRCh38
rs897268499 Chr.1: 982346-982346 on Build GRCh38
rs543786728 Chr.1: 982105-982105 on Build GRCh38
rs965188492 Chr.1: 982085-982085 on Build GRCh38
rs41285816 Chr.1: 982260-982260 on Build GRCh38
rs1026903006 Chr.1: 982208-982208 on Build GRCh38
rs771779744 Chr.1: 982022-982022 on Build GRCh38
rs991864696 Chr.1: 982394-982394 on Build GRCh38
rs1024634083 Chr.1: 982395-982395 on Build GRCh38
rs1037524059 Chr.1: 982191-982191 on Build GRCh38
rs28434575 Chr.1: 982112-982112 on Build GRCh38
rs770954674 Chr.1: 982372-982372 on Build GRCh38
rs926380206 Chr.1: 982162-982162 on Build GRCh38
rs1019432003 Chr.1: 982254-982254 on Build GRCh38
rs969274399 Chr.1: 982132-982132 on Build GRCh38
rs569434313 Chr.1: 982362-982362 on Build GRCh38
rs755911057 Chr.1: 982155-982155 on Build GRCh38
rs747516275 Chr.1: 982316-982316 on Build GRCh38
rs896598604 Chr.1: 981974-981974 on Build GRCh38
rs991791165 Chr.1: 982183-982184 on Build GRCh38
rs776711902 Chr.1: 982390-982390 on Build GRCh38
rs1013990337 Chr.1: 981977-981977 on Build GRCh38
rs191948107 Chr.1: 982090-982090 on Build GRCh38
rs111479166 Chr.1: 982308-982308 on Build GRCh38
rs747124102 Chr.1: 982130-982130 on Build GRCh38
rs1010519313 Chr.1: 982380-982380 on Build GRCh38
rs866247170 Chr.1: 982263-982263 on Build GRCh38
rs768604439 Chr.1: 982148-982148 on Build GRCh38
rs938163116 Chr.1: 982120-982120 on Build GRCh38
rs748497122 Chr.1: 982109-982110 on Build GRCh38
rs995490240 Chr.1: 982354-982354 on Build GRCh38
rs779799027 Chr.1: 982126-982126 on Build GRCh38
rs897117849 Chr.1: 982194-982194 on Build GRCh38
rs559944388 Chr.1: 982256-982256 on Build GRCh38
rs949589034 Chr.1: 982335-982335 on Build GRCh38
rs558030838 Chr.1: 982081-982081 on Build GRCh38
rs370496376 Chr.1: 982076-982076 on Build GRCh38
rs1055249002 Chr.1: 982139-982139 on Build GRCh38
rs953969608 Chr.1: 982211-982211 on Build GRCh38
rs750271971 Chr.1: 982147-982147 on Build GRCh38
rs114920174 Chr.1: 982336-982336 on Build GRCh38
rs111743280 Chr.1: 982113-982113 on Build GRCh38

View Full Product Details