Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93921954-93922413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM181A
Entrez Gene ID
90050
Gene Name
family with sequence similarity 181 member A
Gene Aliases
C14orf152
UniGene
Hs.525550
Gene Chromosome Location
Chr.14: 93918894-93929608 on Build GRCh38
Transcripts
NM_001207071.1 NM_138344.4 NR_027004.1

Gene Symbol
FAM181A-AS1
Entrez Gene ID
283592
Gene Name
FAM181A antisense RNA 1
Gene Aliases
C14orf86
UniGene
Hs.585648
Gene Chromosome Location
Chr.14: 93904730-93926372 on Build GRCh38
Transcripts
NM_001207071.1 NM_138344.4 NR_027004.1

SNP Detail

SNP ID Chromosome Location
rs898554168 Chr.14: 93922226-93922226 on Build GRCh38
rs7154542 Chr.14: 93922382-93922382 on Build GRCh38
rs1032588150 Chr.14: 93922122-93922122 on Build GRCh38
rs923754010 Chr.14: 93922330-93922330 on Build GRCh38
rs573278706 Chr.14: 93922244-93922244 on Build GRCh38
rs968173550 Chr.14: 93922297-93922297 on Build GRCh38
rs976150048 Chr.14: 93922203-93922203 on Build GRCh38
rs935068434 Chr.14: 93922338-93922338 on Build GRCh38
rs926251879 Chr.14: 93922380-93922380 on Build GRCh38
rs543826934 Chr.14: 93922112-93922112 on Build GRCh38
rs942255292 Chr.14: 93922023-93922023 on Build GRCh38
rs534290340 Chr.14: 93922262-93922262 on Build GRCh38
rs557113152 Chr.14: 93922295-93922295 on Build GRCh38
rs963627815 Chr.14: 93922261-93922261 on Build GRCh38
rs78193516 Chr.14: 93921999-93921999 on Build GRCh38
rs571743077 Chr.14: 93922045-93922045 on Build GRCh38
rs1046152577 Chr.14: 93922099-93922099 on Build GRCh38
rs937896186 Chr.14: 93922132-93922132 on Build GRCh38
rs7155685 Chr.14: 93922296-93922296 on Build GRCh38
rs1006728887 Chr.14: 93922148-93922148 on Build GRCh38
rs775735975 Chr.14: 93922070-93922070 on Build GRCh38
rs147036435 Chr.14: 93922004-93922004 on Build GRCh38
rs912698055 Chr.14: 93922009-93922009 on Build GRCh38
rs983675618 Chr.14: 93922255-93922255 on Build GRCh38
rs551114004 Chr.14: 93922123-93922123 on Build GRCh38
rs890047286 Chr.14: 93922258-93922258 on Build GRCh38
rs987943793 Chr.14: 93922050-93922050 on Build GRCh38
rs552663529 Chr.14: 93922278-93922278 on Build GRCh38
rs949136076 Chr.14: 93922098-93922098 on Build GRCh38
rs574339978 Chr.14: 93922285-93922285 on Build GRCh38
rs893757674 Chr.14: 93922128-93922128 on Build GRCh38
rs896362018 Chr.14: 93922186-93922186 on Build GRCh38
rs150665534 Chr.14: 93922228-93922228 on Build GRCh38
rs561632476 Chr.14: 93922309-93922309 on Build GRCh38
rs1028595379 Chr.14: 93921989-93921989 on Build GRCh38
rs956191749 Chr.14: 93922010-93922010 on Build GRCh38
rs144135565 Chr.14: 93922202-93922202 on Build GRCh38
rs902248815 Chr.14: 93922111-93922112 on Build GRCh38
rs1003992767 Chr.14: 93922200-93922200 on Build GRCh38
rs866025406 Chr.14: 93922355-93922355 on Build GRCh38
rs138514423 Chr.14: 93922156-93922156 on Build GRCh38
rs779944921 Chr.14: 93922075-93922075 on Build GRCh38
rs1019871872 Chr.14: 93922286-93922286 on Build GRCh38
rs913628832 Chr.14: 93922056-93922056 on Build GRCh38
rs538875671 Chr.14: 93922101-93922101 on Build GRCh38
rs187904138 Chr.14: 93922288-93922288 on Build GRCh38
rs1043763349 Chr.14: 93922110-93922110 on Build GRCh38
rs764473106 Chr.14: 93922310-93922310 on Build GRCh38
rs1017561426 Chr.14: 93922171-93922171 on Build GRCh38

View Full Product Details