Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63600387-63600889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL22
Entrez Gene ID
283807
Gene Name
F-box and leucine rich repeat protein 22
Gene Aliases
Fbl22
UniGene
Hs.631163
Gene Chromosome Location
Chr.15: 63596225-63608898 on Build GRCh38
Transcripts
NM_203373.2 XM_005254320.3 XM_006720478.1 XM_011521468.2 XM_011521469.1 XM_011521472.2 XM_017022086.1 NR_034080.1

Gene Symbol
USP3-AS1
Entrez Gene ID
100130855
Gene Name
USP3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.631163
Gene Chromosome Location
Chr.15: 63587710-63600827 on Build GRCh38
Transcripts
NM_203373.2 XM_005254320.3 XM_006720478.1 XM_011521468.2 XM_011521469.1 XM_011521472.2 XM_017022086.1 NR_034080.1

SNP Detail

SNP ID Chromosome Location
rs911724850 Chr.15: 63600487-63600487 on Build GRCh38
rs764704360 Chr.15: 63600530-63600530 on Build GRCh38
rs941362907 Chr.15: 63600461-63600461 on Build GRCh38
rs902188660 Chr.15: 63600746-63600746 on Build GRCh38
rs577122984 Chr.15: 63600546-63600546 on Build GRCh38
rs1055064356 Chr.15: 63600735-63600735 on Build GRCh38
rs879373550 Chr.15: 63600628-63600628 on Build GRCh38
rs953018568 Chr.15: 63600639-63600639 on Build GRCh38
rs944575441 Chr.15: 63600674-63600674 on Build GRCh38
rs893530084 Chr.15: 63600737-63600737 on Build GRCh38
rs1019142608 Chr.15: 63600772-63600772 on Build GRCh38
rs955357113 Chr.15: 63600843-63600843 on Build GRCh38
rs889969750 Chr.15: 63600509-63600509 on Build GRCh38
rs575000392 Chr.15: 63600676-63600676 on Build GRCh38
rs1014423965 Chr.15: 63600570-63600570 on Build GRCh38
rs960509386 Chr.15: 63600564-63600564 on Build GRCh38
rs529024679 Chr.15: 63600844-63600844 on Build GRCh38
rs1017197896 Chr.15: 63600765-63600765 on Build GRCh38
rs1020593994 Chr.15: 63600588-63600588 on Build GRCh38
rs147096577 Chr.15: 63600548-63600548 on Build GRCh38
rs986311445 Chr.15: 63600643-63600643 on Build GRCh38
rs937307763 Chr.15: 63600715-63600715 on Build GRCh38
rs1052804980 Chr.15: 63600515-63600515 on Build GRCh38
rs995060760 Chr.15: 63600762-63600762 on Build GRCh38
rs1043087049 Chr.15: 63600536-63600536 on Build GRCh38
rs189797805 Chr.15: 63600792-63600792 on Build GRCh38
rs571923129 Chr.15: 63600706-63600706 on Build GRCh38
rs892379847 Chr.15: 63600524-63600524 on Build GRCh38
rs937375537 Chr.15: 63600818-63600818 on Build GRCh38
rs959232002 Chr.15: 63600648-63600648 on Build GRCh38
rs1010621934 Chr.15: 63600743-63600743 on Build GRCh38
rs543090485 Chr.15: 63600766-63600766 on Build GRCh38
rs117539833 Chr.15: 63600470-63600470 on Build GRCh38
rs904552772 Chr.15: 63600714-63600714 on Build GRCh38
rs934250045 Chr.15: 63600478-63600478 on Build GRCh38
rs538591757 Chr.15: 63600798-63600798 on Build GRCh38
rs973036036 Chr.15: 63600467-63600467 on Build GRCh38
rs985811067 Chr.15: 63600770-63600770 on Build GRCh38
rs966569563 Chr.15: 63600593-63600593 on Build GRCh38
rs926525381 Chr.15: 63600817-63600817 on Build GRCh38
rs911504184 Chr.15: 63600709-63600709 on Build GRCh38
rs1014722671 Chr.15: 63600757-63600757 on Build GRCh38
rs988256226 Chr.15: 63600790-63600790 on Build GRCh38
rs776207721 Chr.15: 63600457-63600457 on Build GRCh38
rs576214671 Chr.15: 63600749-63600749 on Build GRCh38
rs541604748 Chr.15: 63600671-63600671 on Build GRCh38
rs760066196 Chr.15: 63600435-63600435 on Build GRCh38
rs931733551 Chr.15: 63600477-63600477 on Build GRCh38
rs374593605 Chr.15: 63600846-63600846 on Build GRCh38
rs1050157451 Chr.15: 63600482-63600482 on Build GRCh38
rs904501807 Chr.15: 63600547-63600547 on Build GRCh38
rs188022622 Chr.15: 63600454-63600454 on Build GRCh38
rs12899671 Chr.15: 63600712-63600712 on Build GRCh38
rs962280162 Chr.15: 63600594-63600594 on Build GRCh38
rs561818357 Chr.15: 63600812-63600812 on Build GRCh38
rs1015097984 Chr.15: 63600758-63600758 on Build GRCh38
rs531293000 Chr.15: 63600760-63600760 on Build GRCh38
rs185344665 Chr.15: 63600670-63600670 on Build GRCh38
rs973588577 Chr.15: 63600608-63600608 on Build GRCh38
rs1051351593 Chr.15: 63600707-63600707 on Build GRCh38
rs918552854 Chr.15: 63600827-63600827 on Build GRCh38
rs897972684 Chr.15: 63600759-63600759 on Build GRCh38
rs1007683363 Chr.15: 63600538-63600538 on Build GRCh38
rs1056323569 Chr.15: 63600725-63600725 on Build GRCh38
rs556267212 Chr.15: 63600745-63600745 on Build GRCh38
rs965830647 Chr.15: 63600788-63600788 on Build GRCh38
rs559010071 Chr.15: 63600492-63600492 on Build GRCh38
rs1027293828 Chr.15: 63600610-63600610 on Build GRCh38
rs953071071 Chr.15: 63600768-63600768 on Build GRCh38
rs1034775417 Chr.15: 63600645-63600645 on Build GRCh38
rs1015628242 Chr.15: 63600587-63600587 on Build GRCh38
rs192063792 Chr.15: 63600561-63600561 on Build GRCh38
rs913703093 Chr.15: 63600681-63600681 on Build GRCh38

View Full Product Details