Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 83508284-83508816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH13
Entrez Gene ID
1012
Gene Name
cadherin 13
Gene Aliases
CDHH, P105
UniGene
Hs.654386
Gene Chromosome Location
Chr.16: 82626794-83796610 on Build GRCh38
Transcripts
NM_001220488.1 NM_001220489.1 NM_001220490.1 NM_001257.4 XM_011522804.2 XM_017022848.1 NR_036147.1

Gene Symbol
MIR3182
Entrez Gene ID
100422853
Gene Name
microRNA 3182
Gene Aliases
mir-3182
UniGene
Gene Chromosome Location
Chr.16: 83508346-83508408 on Build GRCh38
Transcripts
NM_001220488.1 NM_001220489.1 NM_001220490.1 NM_001257.4 XM_011522804.2 XM_017022848.1 NR_036147.1

SNP Detail

SNP ID Chromosome Location
rs931820156 Chr.16: 83508382-83508382 on Build GRCh38
rs774286983 Chr.16: 83508404-83508404 on Build GRCh38
rs1012361159 Chr.16: 83508642-83508642 on Build GRCh38
rs370649035 Chr.16: 83508430-83508430 on Build GRCh38
rs566522155 Chr.16: 83508319-83508319 on Build GRCh38
rs188269015 Chr.16: 83508386-83508386 on Build GRCh38
rs534033923 Chr.16: 83508789-83508789 on Build GRCh38
rs1055439914 Chr.16: 83508527-83508527 on Build GRCh38
rs971036156 Chr.16: 83508334-83508334 on Build GRCh38
rs377183499 Chr.16: 83508779-83508779 on Build GRCh38
rs919087668 Chr.16: 83508722-83508722 on Build GRCh38
rs145343431 Chr.16: 83508719-83508719 on Build GRCh38
rs73605824 Chr.16: 83508307-83508307 on Build GRCh38
rs146394071 Chr.16: 83508375-83508375 on Build GRCh38
rs1003184069 Chr.16: 83508792-83508792 on Build GRCh38
rs953167222 Chr.16: 83508542-83508542 on Build GRCh38
rs1031642764 Chr.16: 83508513-83508513 on Build GRCh38
rs759494487 Chr.16: 83508547-83508547 on Build GRCh38
rs758603687 Chr.16: 83508655-83508655 on Build GRCh38
rs982767517 Chr.16: 83508341-83508341 on Build GRCh38
rs112877512 Chr.16: 83508470-83508470 on Build GRCh38
rs1004096425 Chr.16: 83508676-83508676 on Build GRCh38
rs578021969 Chr.16: 83508450-83508450 on Build GRCh38
rs940505639 Chr.16: 83508497-83508497 on Build GRCh38
rs1045173382 Chr.16: 83508780-83508780 on Build GRCh38
rs991631941 Chr.16: 83508680-83508680 on Build GRCh38
rs767301810 Chr.16: 83508487-83508487 on Build GRCh38
rs940450302 Chr.16: 83508320-83508323 on Build GRCh38
rs538176217 Chr.16: 83508713-83508713 on Build GRCh38
rs985842520 Chr.16: 83508548-83508548 on Build GRCh38
rs1048989229 Chr.16: 83508728-83508728 on Build GRCh38
rs539550585 Chr.16: 83508342-83508342 on Build GRCh38
rs1048458508 Chr.16: 83508630-83508630 on Build GRCh38
rs1040571652 Chr.16: 83508721-83508721 on Build GRCh38
rs890279064 Chr.16: 83508405-83508405 on Build GRCh38
rs367772162 Chr.16: 83508665-83508665 on Build GRCh38
rs923654044 Chr.16: 83508365-83508365 on Build GRCh38
rs989203066 Chr.16: 83508372-83508372 on Build GRCh38
rs981192411 Chr.16: 83508500-83508500 on Build GRCh38
rs180679894 Chr.16: 83508514-83508514 on Build GRCh38
rs1023898981 Chr.16: 83508325-83508325 on Build GRCh38
rs114548535 Chr.16: 83508553-83508553 on Build GRCh38
rs887782430 Chr.16: 83508747-83508747 on Build GRCh38
rs565510974 Chr.16: 83508574-83508574 on Build GRCh38
rs564683390 Chr.16: 83508360-83508360 on Build GRCh38
rs944527770 Chr.16: 83508573-83508573 on Build GRCh38
rs139736497 Chr.16: 83508385-83508385 on Build GRCh38
rs186120552 Chr.16: 83508601-83508601 on Build GRCh38
rs757650856 Chr.16: 83508755-83508755 on Build GRCh38
rs376190368 Chr.16: 83508374-83508374 on Build GRCh38
rs532879899 Chr.16: 83508586-83508586 on Build GRCh38
rs189432655 Chr.16: 83508669-83508669 on Build GRCh38
rs944681318 Chr.16: 83508425-83508425 on Build GRCh38
rs377076264 Chr.16: 83508366-83508366 on Build GRCh38
rs369064261 Chr.16: 83508720-83508720 on Build GRCh38
rs115777277 Chr.16: 83508477-83508477 on Build GRCh38
rs575622679 Chr.16: 83508409-83508409 on Build GRCh38
rs915001261 Chr.16: 83508534-83508534 on Build GRCh38
rs752823513 Chr.16: 83508631-83508631 on Build GRCh38
rs150045618 Chr.16: 83508502-83508502 on Build GRCh38
rs1019025040 Chr.16: 83508436-83508436 on Build GRCh38
rs929967139 Chr.16: 83508622-83508622 on Build GRCh38
rs114424746 Chr.16: 83508794-83508794 on Build GRCh38
rs200484212 Chr.16: 83508667-83508668 on Build GRCh38
rs549019358 Chr.16: 83508672-83508672 on Build GRCh38
rs570425242 Chr.16: 83508339-83508339 on Build GRCh38
rs372828591 Chr.16: 83508546-83508546 on Build GRCh38
rs557988407 Chr.16: 83508657-83508657 on Build GRCh38
rs907693359 Chr.16: 83508320-83508321 on Build GRCh38
rs149172273 Chr.16: 83508735-83508735 on Build GRCh38
rs546264806 Chr.16: 83508367-83508367 on Build GRCh38
rs866348421 Chr.16: 83508383-83508383 on Build GRCh38
rs4782541 Chr.16: 83508621-83508621 on Build GRCh38
rs1040873601 Chr.16: 83508590-83508590 on Build GRCh38
rs937015195 Chr.16: 83508520-83508520 on Build GRCh38
rs538008506 Chr.16: 83508433-83508433 on Build GRCh38
rs535361829 Chr.16: 83508768-83508768 on Build GRCh38
rs114268765 Chr.16: 83508447-83508447 on Build GRCh38
rs746971300 Chr.16: 83508626-83508626 on Build GRCh38
rs952020093 Chr.16: 83508628-83508628 on Build GRCh38
rs996647416 Chr.16: 83508616-83508616 on Build GRCh38
rs551575929 Chr.16: 83508335-83508335 on Build GRCh38
rs181216326 Chr.16: 83508726-83508726 on Build GRCh38
rs1006193393 Chr.16: 83508753-83508753 on Build GRCh38
rs533750761 Chr.16: 83508743-83508743 on Build GRCh38
rs1031255810 Chr.16: 83508677-83508677 on Build GRCh38
rs764365970 Chr.16: 83508695-83508695 on Build GRCh38
rs560282788 Chr.16: 83508466-83508466 on Build GRCh38
rs117609775 Chr.16: 83508510-83508510 on Build GRCh38
rs1011107557 Chr.16: 83508691-83508691 on Build GRCh38
rs4782540 Chr.16: 83508456-83508456 on Build GRCh38
rs79510224 Chr.16: 83508690-83508690 on Build GRCh38
rs999608409 Chr.16: 83508560-83508560 on Build GRCh38
rs906571893 Chr.16: 83508787-83508787 on Build GRCh38
rs532235818 Chr.16: 83508432-83508432 on Build GRCh38
rs907745808 Chr.16: 83508488-83508488 on Build GRCh38
rs989067243 Chr.16: 83508519-83508519 on Build GRCh38
rs911605364 Chr.16: 83508715-83508715 on Build GRCh38
rs1021958019 Chr.16: 83508699-83508699 on Build GRCh38
rs953033325 Chr.16: 83508703-83508703 on Build GRCh38
rs1043957141 Chr.16: 83508712-83508712 on Build GRCh38
rs1012328625 Chr.16: 83508464-83508464 on Build GRCh38

View Full Product Details