Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13427277-13427775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD4
Entrez Gene ID
753
Gene Name
low density lipoprotein receptor class A domain containing 4
Gene Aliases
C18orf1
UniGene
Hs.731853
Gene Chromosome Location
Chr.18: 13217691-13652754 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1 NR_040031.1

Gene Symbol
LDLRAD4-AS1
Entrez Gene ID
100288122
Gene Name
LDLRAD4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.646264
Gene Chromosome Location
Chr.18: 13419499-13427534 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1 NR_040031.1

SNP Detail

SNP ID Chromosome Location
rs926564492 Chr.18: 13427442-13427442 on Build GRCh38
rs980727212 Chr.18: 13427419-13427419 on Build GRCh38
rs956447349 Chr.18: 13427541-13427541 on Build GRCh38
rs1003032889 Chr.18: 13427522-13427522 on Build GRCh38
rs368172945 Chr.18: 13427723-13427723 on Build GRCh38
rs1049494939 Chr.18: 13427468-13427468 on Build GRCh38
rs1016989928 Chr.18: 13427415-13427415 on Build GRCh38
rs1048987940 Chr.18: 13427709-13427709 on Build GRCh38
rs759067309 Chr.18: 13427619-13427619 on Build GRCh38
rs1004349092 Chr.18: 13427610-13427610 on Build GRCh38
rs560996739 Chr.18: 13427560-13427560 on Build GRCh38
rs919100683 Chr.18: 13427662-13427662 on Build GRCh38
rs887805139 Chr.18: 13427474-13427474 on Build GRCh38
rs910301065 Chr.18: 13427588-13427588 on Build GRCh38
rs55800405 Chr.18: 13427604-13427604 on Build GRCh38
rs897728873 Chr.18: 13427416-13427416 on Build GRCh38
rs573714020 Chr.18: 13427515-13427515 on Build GRCh38
rs137969018 Chr.18: 13427521-13427521 on Build GRCh38
rs571406841 Chr.18: 13427661-13427661 on Build GRCh38
rs79061269 Chr.18: 13427548-13427548 on Build GRCh38
rs760289461 Chr.18: 13427513-13427513 on Build GRCh38
rs939239834 Chr.18: 13427454-13427454 on Build GRCh38
rs772905540 Chr.18: 13427420-13427420 on Build GRCh38
rs992030525 Chr.18: 13427471-13427471 on Build GRCh38
rs544849406 Chr.18: 13427323-13427323 on Build GRCh38
rs985855851 Chr.18: 13427402-13427402 on Build GRCh38
rs976286717 Chr.18: 13427627-13427627 on Build GRCh38
rs765667842 Chr.18: 13427725-13427725 on Build GRCh38
rs988176058 Chr.18: 13427349-13427349 on Build GRCh38
rs1038668541 Chr.18: 13427544-13427544 on Build GRCh38
rs149462229 Chr.18: 13427585-13427585 on Build GRCh38
rs140235022 Chr.18: 13427403-13427403 on Build GRCh38
rs1002237307 Chr.18: 13427716-13427716 on Build GRCh38
rs914913633 Chr.18: 13427359-13427359 on Build GRCh38
rs1040247783 Chr.18: 13427411-13427411 on Build GRCh38
rs1023043799 Chr.18: 13427517-13427517 on Build GRCh38
rs929247982 Chr.18: 13427441-13427441 on Build GRCh38
rs1022349606 Chr.18: 13427573-13427573 on Build GRCh38
rs150325467 Chr.18: 13427405-13427405 on Build GRCh38
rs956348502 Chr.18: 13427312-13427312 on Build GRCh38
rs771804995 Chr.18: 13427329-13427332 on Build GRCh38
rs913786341 Chr.18: 13427746-13427746 on Build GRCh38
rs373025378 Chr.18: 13427732-13427732 on Build GRCh38
rs929193150 Chr.18: 13427665-13427665 on Build GRCh38
rs775961130 Chr.18: 13427528-13427528 on Build GRCh38
rs571117982 Chr.18: 13427575-13427575 on Build GRCh38
rs962783424 Chr.18: 13427672-13427672 on Build GRCh38
rs1017031659 Chr.18: 13427668-13427668 on Build GRCh38
rs946445535 Chr.18: 13427380-13427380 on Build GRCh38

View Full Product Details