Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10192502-10192993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf48
Entrez Gene ID
348738
Gene Name
chromosome 2 open reading frame 48
Gene Aliases
-
UniGene
Hs.454981
Gene Chromosome Location
Chr.2: 10141382-10211730 on Build GRCh38
Transcripts
NM_182626.3 NR_036222.1

Gene Symbol
MIR4261
Entrez Gene ID
100422929
Gene Name
microRNA 4261
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 10192614-10192671 on Build GRCh38
Transcripts
NM_182626.3 NR_036222.1

SNP Detail

SNP ID Chromosome Location
rs754989195 Chr.2: 10192892-10192892 on Build GRCh38
rs184441452 Chr.2: 10192928-10192928 on Build GRCh38
rs920867012 Chr.2: 10192794-10192794 on Build GRCh38
rs1010530812 Chr.2: 10192681-10192681 on Build GRCh38
rs1040770278 Chr.2: 10192660-10192660 on Build GRCh38
rs964386845 Chr.2: 10192829-10192829 on Build GRCh38
rs575451039 Chr.2: 10192757-10192757 on Build GRCh38
rs922749515 Chr.2: 10192618-10192618 on Build GRCh38
rs200644374 Chr.2: 10192759-10192759 on Build GRCh38
rs1000096638 Chr.2: 10192944-10192944 on Build GRCh38
rs892507653 Chr.2: 10192678-10192678 on Build GRCh38
rs904336591 Chr.2: 10192702-10192702 on Build GRCh38
rs947705067 Chr.2: 10192929-10192929 on Build GRCh38
rs1010812959 Chr.2: 10192806-10192806 on Build GRCh38
rs916219131 Chr.2: 10192704-10192704 on Build GRCh38
rs910097662 Chr.2: 10192688-10192688 on Build GRCh38
rs930804645 Chr.2: 10192591-10192591 on Build GRCh38
rs561278314 Chr.2: 10192813-10192813 on Build GRCh38
rs1052273047 Chr.2: 10192772-10192772 on Build GRCh38
rs941636088 Chr.2: 10192694-10192694 on Build GRCh38
rs577344769 Chr.2: 10192612-10192612 on Build GRCh38
rs867685316 Chr.2: 10192548-10192548 on Build GRCh38
rs532668655 Chr.2: 10192894-10192894 on Build GRCh38
rs544773188 Chr.2: 10192690-10192690 on Build GRCh38
rs759921963 Chr.2: 10192594-10192594 on Build GRCh38
rs566045458 Chr.2: 10192750-10192750 on Build GRCh38
rs951467984 Chr.2: 10192869-10192869 on Build GRCh38
rs372326630 Chr.2: 10192864-10192864 on Build GRCh38
rs56228505 Chr.2: 10192751-10192751 on Build GRCh38
rs796486033 Chr.2: 10192758-10192758 on Build GRCh38
rs565350878 Chr.2: 10192890-10192890 on Build GRCh38
rs1050479757 Chr.2: 10192607-10192607 on Build GRCh38
rs779961413 Chr.2: 10192858-10192858 on Build GRCh38
rs958685603 Chr.2: 10192753-10192753 on Build GRCh38
rs927661596 Chr.2: 10192972-10192972 on Build GRCh38
rs542556987 Chr.2: 10192778-10192778 on Build GRCh38
rs189325293 Chr.2: 10192583-10192583 on Build GRCh38
rs981811706 Chr.2: 10192930-10192930 on Build GRCh38
rs192288636 Chr.2: 10192629-10192629 on Build GRCh38
rs992885054 Chr.2: 10192521-10192521 on Build GRCh38
rs530363843 Chr.2: 10192912-10192912 on Build GRCh38
rs1017071332 Chr.2: 10192885-10192885 on Build GRCh38
rs528481253 Chr.2: 10192854-10192854 on Build GRCh38
rs916949400 Chr.2: 10192542-10192542 on Build GRCh38
rs879796753 Chr.2: 10192755-10192755 on Build GRCh38
rs551112149 Chr.2: 10192895-10192895 on Build GRCh38
rs962881166 Chr.2: 10192893-10192893 on Build GRCh38
rs1034168752 Chr.2: 10192717-10192717 on Build GRCh38
rs972380475 Chr.2: 10192582-10192582 on Build GRCh38
rs910758951 Chr.2: 10192828-10192828 on Build GRCh38
rs114679793 Chr.2: 10192899-10192899 on Build GRCh38
rs951541030 Chr.2: 10192638-10192638 on Build GRCh38
rs572914154 Chr.2: 10192771-10192771 on Build GRCh38
rs750227468 Chr.2: 10192653-10192653 on Build GRCh38
rs944786237 Chr.2: 10192842-10192842 on Build GRCh38
rs563350901 Chr.2: 10192705-10192705 on Build GRCh38
rs906316027 Chr.2: 10192756-10192756 on Build GRCh38
rs185770523 Chr.2: 10192564-10192564 on Build GRCh38
rs1037342421 Chr.2: 10192699-10192699 on Build GRCh38
rs10187710 Chr.2: 10192884-10192884 on Build GRCh38

View Full Product Details