Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25035111-25035631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27-AS1
Entrez Gene ID
729723
Gene Name
DNAJC27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.436366
Gene Chromosome Location
Chr.2: 24972112-25039694 on Build GRCh38
Transcripts
NR_034113.1

SNP Detail

SNP ID Chromosome Location
rs753503587 Chr.2: 25035402-25035402 on Build GRCh38
rs1042676086 Chr.2: 25035250-25035250 on Build GRCh38
rs6760296 Chr.2: 25035324-25035324 on Build GRCh38
rs1023641897 Chr.2: 25035424-25035424 on Build GRCh38
rs1031280454 Chr.2: 25035329-25035329 on Build GRCh38
rs1000906490 Chr.2: 25035268-25035268 on Build GRCh38
rs150547049 Chr.2: 25035408-25035408 on Build GRCh38
rs927766158 Chr.2: 25035500-25035500 on Build GRCh38
rs969425987 Chr.2: 25035434-25035434 on Build GRCh38
rs947863769 Chr.2: 25035142-25035142 on Build GRCh38
rs190527693 Chr.2: 25035537-25035537 on Build GRCh38
rs376963353 Chr.2: 25035182-25035182 on Build GRCh38
rs760225059 Chr.2: 25035176-25035176 on Build GRCh38
rs899522371 Chr.2: 25035586-25035586 on Build GRCh38
rs13395059 Chr.2: 25035446-25035446 on Build GRCh38
rs1005254326 Chr.2: 25035248-25035248 on Build GRCh38
rs945184860 Chr.2: 25035600-25035600 on Build GRCh38
rs899553460 Chr.2: 25035291-25035291 on Build GRCh38
rs1046222710 Chr.2: 25035167-25035167 on Build GRCh38
rs541407386 Chr.2: 25035432-25035432 on Build GRCh38
rs10445939 Chr.2: 25035528-25035528 on Build GRCh38
rs1000166710 Chr.2: 25035326-25035326 on Build GRCh38
rs1046998938 Chr.2: 25035196-25035196 on Build GRCh38
rs971805938 Chr.2: 25035137-25035137 on Build GRCh38
rs913338371 Chr.2: 25035551-25035551 on Build GRCh38
rs185144695 Chr.2: 25035371-25035371 on Build GRCh38
rs796396695 Chr.2: 25035411-25035411 on Build GRCh38
rs765860632 Chr.2: 25035316-25035316 on Build GRCh38
rs982204227 Chr.2: 25035495-25035495 on Build GRCh38
rs1057179327 Chr.2: 25035164-25035164 on Build GRCh38
rs554402287 Chr.2: 25035272-25035272 on Build GRCh38
rs575710178 Chr.2: 25035292-25035292 on Build GRCh38
rs192947233 Chr.2: 25035301-25035301 on Build GRCh38
rs145147045 Chr.2: 25035300-25035300 on Build GRCh38
rs980470109 Chr.2: 25035520-25035520 on Build GRCh38
rs929857058 Chr.2: 25035516-25035516 on Build GRCh38
rs544405022 Chr.2: 25035399-25035399 on Build GRCh38
rs559985087 Chr.2: 25035453-25035453 on Build GRCh38
rs1016899251 Chr.2: 25035391-25035391 on Build GRCh38
rs74447731 Chr.2: 25035536-25035536 on Build GRCh38
rs983638339 Chr.2: 25035372-25035372 on Build GRCh38
rs1039311444 Chr.2: 25035542-25035542 on Build GRCh38
rs989533035 Chr.2: 25035379-25035379 on Build GRCh38
rs1039425745 Chr.2: 25035262-25035262 on Build GRCh38
rs35027159 Chr.2: 25035209-25035209 on Build GRCh38
rs888099998 Chr.2: 25035232-25035232 on Build GRCh38

View Full Product Details