Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199848689-199849201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTCDNL1
Entrez Gene ID
348751
Gene Name
formiminotransferase cyclodeaminase N-terminal like
Gene Aliases
FONG
UniGene
Hs.734578
Gene Chromosome Location
Chr.2: 199760536-199851173 on Build GRCh38
Transcripts
NR_034096.1

SNP Detail

SNP ID Chromosome Location
rs963363416 Chr.2: 199849059-199849059 on Build GRCh38
rs548171302 Chr.2: 199848809-199848809 on Build GRCh38
rs529738721 Chr.2: 199848714-199848714 on Build GRCh38
rs983416980 Chr.2: 199848749-199848749 on Build GRCh38
rs901299393 Chr.2: 199848787-199848787 on Build GRCh38
rs112327156 Chr.2: 199849175-199849175 on Build GRCh38
rs73988960 Chr.2: 199848955-199848955 on Build GRCh38
rs187422297 Chr.2: 199849058-199849058 on Build GRCh38
rs756148000 Chr.2: 199848938-199848938 on Build GRCh38
rs189318648 Chr.2: 199848882-199848882 on Build GRCh38
rs1041489375 Chr.2: 199848785-199848785 on Build GRCh38
rs980535599 Chr.2: 199849105-199849105 on Build GRCh38
rs980197750 Chr.2: 199848871-199848871 on Build GRCh38
rs111738686 Chr.2: 199848814-199848814 on Build GRCh38
rs772049933 Chr.2: 199848744-199848747 on Build GRCh38
rs556409509 Chr.2: 199849042-199849042 on Build GRCh38
rs978238076 Chr.2: 199848987-199848987 on Build GRCh38
rs4672569 Chr.2: 199849028-199849028 on Build GRCh38
rs1017335705 Chr.2: 199848988-199848988 on Build GRCh38
rs369211175 Chr.2: 199849002-199849002 on Build GRCh38
rs1031151373 Chr.2: 199848992-199848992 on Build GRCh38
rs1000516183 Chr.2: 199848966-199848966 on Build GRCh38
rs769511203 Chr.2: 199849112-199849112 on Build GRCh38
rs751000971 Chr.2: 199848899-199848899 on Build GRCh38
rs761580889 Chr.2: 199848893-199848893 on Build GRCh38
rs751212645 Chr.2: 199848818-199848818 on Build GRCh38
rs886289878 Chr.2: 199848830-199848830 on Build GRCh38
rs1011953483 Chr.2: 199848935-199848935 on Build GRCh38
rs552443435 Chr.2: 199848937-199848937 on Build GRCh38
rs927390521 Chr.2: 199849148-199849148 on Build GRCh38
rs970531441 Chr.2: 199848972-199848972 on Build GRCh38
rs942694689 Chr.2: 199849033-199849034 on Build GRCh38
rs1030588457 Chr.2: 199849033-199849033 on Build GRCh38
rs746885682 Chr.2: 199848939-199848939 on Build GRCh38
rs909215945 Chr.2: 199848768-199848768 on Build GRCh38
rs946303604 Chr.2: 199848921-199848921 on Build GRCh38
rs1006065010 Chr.2: 199848850-199848850 on Build GRCh38
rs950647706 Chr.2: 199848728-199848728 on Build GRCh38
rs1057491633 Chr.2: 199848897-199848897 on Build GRCh38
rs1016245982 Chr.2: 199849049-199849049 on Build GRCh38
rs900289235 Chr.2: 199849022-199849022 on Build GRCh38
rs942726975 Chr.2: 199848756-199848756 on Build GRCh38
rs779269055 Chr.2: 199848889-199848889 on Build GRCh38
rs983387348 Chr.2: 199849043-199849043 on Build GRCh38
rs746603798 Chr.2: 199849083-199849083 on Build GRCh38
rs745350369 Chr.2: 199849084-199849084 on Build GRCh38

View Full Product Details