Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25763563-25764066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM182B
Entrez Gene ID
728882
Gene Name
family with sequence similarity 182 member B
Gene Aliases
-
UniGene
Hs.682103
Gene Chromosome Location
Chr.20: 25763466-25801291 on Build GRCh38
Transcripts
NR_027061.2

SNP Detail

SNP ID Chromosome Location
rs201135551 Chr.20: 25763772-25763772 on Build GRCh38
rs542841432 Chr.20: 25763895-25763895 on Build GRCh38
rs6050876 Chr.20: 25763766-25763766 on Build GRCh38
rs796616458 Chr.20: 25764040-25764040 on Build GRCh38
rs375999069 Chr.20: 25763615-25763615 on Build GRCh38
rs141108487 Chr.20: 25763903-25763903 on Build GRCh38
rs77861871 Chr.20: 25763605-25763605 on Build GRCh38
rs530502246 Chr.20: 25763969-25763969 on Build GRCh38
rs199883094 Chr.20: 25763885-25763885 on Build GRCh38
rs543480339 Chr.20: 25763681-25763681 on Build GRCh38
rs888203153 Chr.20: 25763779-25763779 on Build GRCh38
rs978583926 Chr.20: 25764008-25764008 on Build GRCh38
rs113648357 Chr.20: 25763731-25763731 on Build GRCh38
rs865909440 Chr.20: 25763832-25763832 on Build GRCh38
rs368994213 Chr.20: 25763739-25763739 on Build GRCh38
rs75956650 Chr.20: 25763583-25763583 on Build GRCh38
rs200941897 Chr.20: 25763888-25763888 on Build GRCh38
rs1005749205 Chr.20: 25763826-25763826 on Build GRCh38
rs201723030 Chr.20: 25764010-25764010 on Build GRCh38
rs934538509 Chr.20: 25763628-25763628 on Build GRCh38
rs75845810 Chr.20: 25763662-25763662 on Build GRCh38
rs751861732 Chr.20: 25763872-25763872 on Build GRCh38
rs868434944 Chr.20: 25763673-25763673 on Build GRCh38
rs186895380 Chr.20: 25763682-25763682 on Build GRCh38
rs80331156 Chr.20: 25763945-25763945 on Build GRCh38
rs200428226 Chr.20: 25763948-25763948 on Build GRCh38
rs6115283 Chr.20: 25763967-25763967 on Build GRCh38
rs924443707 Chr.20: 25764019-25764019 on Build GRCh38
rs201631879 Chr.20: 25763666-25763666 on Build GRCh38
rs982330287 Chr.20: 25763960-25763960 on Build GRCh38
rs1035255655 Chr.20: 25763965-25763965 on Build GRCh38
rs946788225 Chr.20: 25763986-25763986 on Build GRCh38
rs78235498 Chr.20: 25764033-25764033 on Build GRCh38
rs6115282 Chr.20: 25763925-25763925 on Build GRCh38
rs186448127 Chr.20: 25763785-25763785 on Build GRCh38
rs529409743 Chr.20: 25763876-25763876 on Build GRCh38
rs367606621 Chr.20: 25763595-25763595 on Build GRCh38
rs990814376 Chr.20: 25763973-25763973 on Build GRCh38
rs376251071 Chr.20: 25763732-25763732 on Build GRCh38
rs76953668 Chr.20: 25763651-25763651 on Build GRCh38
rs78029083 Chr.20: 25763838-25763838 on Build GRCh38
rs1040960511 Chr.20: 25763631-25763631 on Build GRCh38
rs796624336 Chr.20: 25763995-25763995 on Build GRCh38
rs551666946 Chr.20: 25763933-25763933 on Build GRCh38
rs867576758 Chr.20: 25763988-25763988 on Build GRCh38
rs563787817 Chr.20: 25763847-25763847 on Build GRCh38
rs563366263 Chr.20: 25764000-25764000 on Build GRCh38
rs915262220 Chr.20: 25763979-25763979 on Build GRCh38
rs369723238 Chr.20: 25763623-25763623 on Build GRCh38
rs996456074 Chr.20: 25763680-25763680 on Build GRCh38
rs577944503 Chr.20: 25763584-25763584 on Build GRCh38
rs796343046 Chr.20: 25763989-25763989 on Build GRCh38
rs559541863 Chr.20: 25763910-25763910 on Build GRCh38
rs796258774 Chr.20: 25763996-25763996 on Build GRCh38
rs1027969845 Chr.20: 25763721-25763721 on Build GRCh38

View Full Product Details