Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19018775-19019273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR5
Entrez Gene ID
26220
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
Gene Aliases
LINC00037, NCRNA00037
UniGene
Hs.646438
Gene Chromosome Location
Chr.22: 18970498-19031242 on Build GRCh38
Transcripts
NR_110533.1 NR_024159.1

Gene Symbol
DGCR9
Entrez Gene ID
25787
Gene Name
DiGeorge syndrome critical region gene 9 (non-protein coding)
Gene Aliases
DGS-A
UniGene
Hs.106311
Gene Chromosome Location
Chr.22: 19017834-19020248 on Build GRCh38
Transcripts
NR_110533.1 NR_024159.1

SNP Detail

SNP ID Chromosome Location
rs1018297178 Chr.22: 19018959-19018959 on Build GRCh38
rs114655471 Chr.22: 19019237-19019237 on Build GRCh38
rs1026884234 Chr.22: 19019095-19019095 on Build GRCh38
rs1006001163 Chr.22: 19018949-19018949 on Build GRCh38
rs949776414 Chr.22: 19019018-19019018 on Build GRCh38
rs751606492 Chr.22: 19018879-19018879 on Build GRCh38
rs527694752 Chr.22: 19018923-19018923 on Build GRCh38
rs374738861 Chr.22: 19019114-19019114 on Build GRCh38
rs991384477 Chr.22: 19019232-19019232 on Build GRCh38
rs1046525564 Chr.22: 19018859-19018859 on Build GRCh38
rs906707680 Chr.22: 19018872-19018872 on Build GRCh38
rs1032854586 Chr.22: 19019205-19019205 on Build GRCh38
rs554253621 Chr.22: 19019056-19019056 on Build GRCh38
rs747781212 Chr.22: 19018994-19018994 on Build GRCh38
rs925621388 Chr.22: 19018811-19018811 on Build GRCh38
rs1042052702 Chr.22: 19018801-19018801 on Build GRCh38
rs938891984 Chr.22: 19018998-19018998 on Build GRCh38
rs867994893 Chr.22: 19019100-19019100 on Build GRCh38
rs955218690 Chr.22: 19018885-19018885 on Build GRCh38
rs899685727 Chr.22: 19019240-19019240 on Build GRCh38
rs890886783 Chr.22: 19019233-19019233 on Build GRCh38
rs890875140 Chr.22: 19019042-19019042 on Build GRCh38
rs114664207 Chr.22: 19018995-19018995 on Build GRCh38
rs967236698 Chr.22: 19019191-19019191 on Build GRCh38
rs964086736 Chr.22: 19018962-19018962 on Build GRCh38
rs147023150 Chr.22: 19019087-19019087 on Build GRCh38
rs775968497 Chr.22: 19019073-19019073 on Build GRCh38
rs770237022 Chr.22: 19018796-19018796 on Build GRCh38
rs377656497 Chr.22: 19018851-19018851 on Build GRCh38
rs999177446 Chr.22: 19019035-19019035 on Build GRCh38
rs747516603 Chr.22: 19019162-19019162 on Build GRCh38
rs994777112 Chr.22: 19019036-19019036 on Build GRCh38
rs548597182 Chr.22: 19018804-19018804 on Build GRCh38
rs894737729 Chr.22: 19019016-19019016 on Build GRCh38
rs878991219 Chr.22: 19018873-19018873 on Build GRCh38
rs112853840 Chr.22: 19019131-19019131 on Build GRCh38
rs768630716 Chr.22: 19019158-19019158 on Build GRCh38
rs573815478 Chr.22: 19018924-19018924 on Build GRCh38
rs1043729313 Chr.22: 19019218-19019218 on Build GRCh38
rs947563296 Chr.22: 19019185-19019185 on Build GRCh38
rs754818904 Chr.22: 19018977-19018977 on Build GRCh38
rs1026341169 Chr.22: 19018881-19018881 on Build GRCh38
rs550251070 Chr.22: 19019113-19019113 on Build GRCh38
rs949749694 Chr.22: 19019239-19019239 on Build GRCh38
rs766397423 Chr.22: 19018825-19018825 on Build GRCh38
rs1029788718 Chr.22: 19019151-19019151 on Build GRCh38
rs755747785 Chr.22: 19019022-19019022 on Build GRCh38
rs906687737 Chr.22: 19019020-19019020 on Build GRCh38
rs113697007 Chr.22: 19019051-19019051 on Build GRCh38
rs976854355 Chr.22: 19018982-19018982 on Build GRCh38
rs542952945 Chr.22: 19019000-19019000 on Build GRCh38
rs899848842 Chr.22: 19019121-19019121 on Build GRCh38
rs577101449 Chr.22: 19019146-19019146 on Build GRCh38
rs770482402 Chr.22: 19019057-19019057 on Build GRCh38
rs974372996 Chr.22: 19019080-19019080 on Build GRCh38
rs562749453 Chr.22: 19019225-19019225 on Build GRCh38
rs957487612 Chr.22: 19019216-19019216 on Build GRCh38
rs1018228755 Chr.22: 19018941-19018941 on Build GRCh38
rs967638321 Chr.22: 19018990-19018990 on Build GRCh38
rs556934262 Chr.22: 19018839-19018839 on Build GRCh38
rs539631661 Chr.22: 19018997-19018997 on Build GRCh38
rs998260207 Chr.22: 19019135-19019135 on Build GRCh38
rs974109063 Chr.22: 19018987-19018987 on Build GRCh38
rs369649180 Chr.22: 19019019-19019019 on Build GRCh38
rs1055460966 Chr.22: 19018829-19018829 on Build GRCh38
rs562013025 Chr.22: 19018855-19018855 on Build GRCh38
rs562177249 Chr.22: 19019238-19019238 on Build GRCh38
rs553760759 Chr.22: 19019136-19019136 on Build GRCh38
rs781141726 Chr.22: 19018989-19018989 on Build GRCh38
rs777302374 Chr.22: 19019037-19019037 on Build GRCh38
rs545669949 Chr.22: 19019198-19019198 on Build GRCh38
rs576158909 Chr.22: 19019231-19019231 on Build GRCh38
rs952741169 Chr.22: 19018899-19018899 on Build GRCh38
rs536617046 Chr.22: 19018832-19018832 on Build GRCh38
rs903542040 Chr.22: 19019029-19019029 on Build GRCh38
rs951419834 Chr.22: 19019103-19019103 on Build GRCh38
rs116468634 Chr.22: 19018996-19018996 on Build GRCh38
rs576138943 Chr.22: 19018825-19018826 on Build GRCh38
rs916042790 Chr.22: 19019168-19019168 on Build GRCh38
rs568859389 Chr.22: 19019046-19019046 on Build GRCh38
rs986622107 Chr.22: 19018922-19018922 on Build GRCh38
rs533565620 Chr.22: 19019089-19019089 on Build GRCh38
rs895888130 Chr.22: 19019248-19019248 on Build GRCh38

View Full Product Details