Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42732475-42732982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
UniGene
Hs.604182
Gene Chromosome Location
Chr.3: 42704118-42773253 on Build GRCh38
Transcripts
NM_144719.3 XM_005264898.3 XM_011533418.2 XM_011533419.2 XM_017005781.1 NR_046732.1

Gene Symbol
CCDC13-AS1
Entrez Gene ID
100874114
Gene Name
CCDC13 antisense RNA 1
Gene Aliases
-
UniGene
Hs.669654
Gene Chromosome Location
Chr.3: 42732575-42745158 on Build GRCh38
Transcripts
NM_144719.3 XM_005264898.3 XM_011533418.2 XM_011533419.2 XM_017005781.1 NR_046732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7270718 c.1512-4A>G p.? Chr.3: 42732974-42732974 on Build GRCh38
5570082 c.1556C>A p.T519K Chr.3: 42732926-42732926 on Build GRCh38
5479629 c.1579_1580CA>AC p.H527T Chr.3: 42732902-42732903 on Build GRCh38
6511033 c.1572C>T p.P524P Chr.3: 42732910-42732910 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751437870 Chr.3: 42732840-42732840 on Build GRCh38
rs1004394229 Chr.3: 42732945-42732945 on Build GRCh38
rs556573992 Chr.3: 42732845-42732845 on Build GRCh38
rs961757605 Chr.3: 42732761-42732761 on Build GRCh38
rs779679755 Chr.3: 42732849-42732850 on Build GRCh38
rs199935877 Chr.3: 42732953-42732953 on Build GRCh38
rs534259848 Chr.3: 42732651-42732651 on Build GRCh38
rs540162764 Chr.3: 42732806-42732806 on Build GRCh38
rs1049285536 Chr.3: 42732876-42732876 on Build GRCh38
rs768778002 Chr.3: 42732501-42732501 on Build GRCh38
rs886573465 Chr.3: 42732502-42732502 on Build GRCh38
rs759351124 Chr.3: 42732947-42732947 on Build GRCh38
rs1020629408 Chr.3: 42732775-42732775 on Build GRCh38
rs747412610 Chr.3: 42732893-42732893 on Build GRCh38
rs1045826534 Chr.3: 42732582-42732582 on Build GRCh38
rs775956906 Chr.3: 42732900-42732900 on Build GRCh38
rs758828255 Chr.3: 42732878-42732878 on Build GRCh38
rs1041496756 Chr.3: 42732872-42732872 on Build GRCh38
rs908140010 Chr.3: 42732883-42732883 on Build GRCh38
rs762793296 Chr.3: 42732933-42732933 on Build GRCh38
rs986465452 Chr.3: 42732833-42732833 on Build GRCh38
rs866561558 Chr.3: 42732644-42732644 on Build GRCh38
rs748764673 Chr.3: 42732922-42732922 on Build GRCh38
rs920255912 Chr.3: 42732776-42732776 on Build GRCh38
rs779250951 Chr.3: 42732875-42732875 on Build GRCh38
rs367680312 Chr.3: 42732925-42732925 on Build GRCh38
rs772848305 Chr.3: 42732930-42732930 on Build GRCh38
rs567513299 Chr.3: 42732616-42732616 on Build GRCh38
rs368326533 Chr.3: 42732867-42732867 on Build GRCh38
rs570841055 Chr.3: 42732781-42732781 on Build GRCh38
rs746272549 Chr.3: 42732877-42732877 on Build GRCh38
rs117005887 Chr.3: 42732944-42732944 on Build GRCh38
rs772093104 Chr.3: 42732676-42732676 on Build GRCh38
rs973100459 Chr.3: 42732774-42732774 on Build GRCh38
rs867525995 Chr.3: 42732550-42732550 on Build GRCh38
rs1045921727 Chr.3: 42732923-42732923 on Build GRCh38
rs1014685626 Chr.3: 42732658-42732658 on Build GRCh38
rs768248165 Chr.3: 42732898-42732898 on Build GRCh38
rs967212373 Chr.3: 42732780-42732780 on Build GRCh38
rs180980060 Chr.3: 42732932-42732932 on Build GRCh38
rs372263809 Chr.3: 42732928-42732928 on Build GRCh38
rs1032964689 Chr.3: 42732503-42732503 on Build GRCh38
rs1005477277 Chr.3: 42732509-42732509 on Build GRCh38
rs536964687 Chr.3: 42732631-42732631 on Build GRCh38
rs776348972 Chr.3: 42732948-42732948 on Build GRCh38
rs34594350 Chr.3: 42732868-42732868 on Build GRCh38
rs371554766 Chr.3: 42732926-42732926 on Build GRCh38
rs1054411157 Chr.3: 42732887-42732887 on Build GRCh38
rs774570348 Chr.3: 42732516-42732516 on Build GRCh38
rs189301090 Chr.3: 42732500-42732500 on Build GRCh38
rs1012892895 Chr.3: 42732913-42732913 on Build GRCh38
rs766028854 Chr.3: 42732937-42732937 on Build GRCh38
rs7617368 Chr.3: 42732656-42732656 on Build GRCh38
rs757502175 Chr.3: 42732874-42732874 on Build GRCh38
rs773255739 Chr.3: 42732802-42732802 on Build GRCh38
rs147142085 Chr.3: 42732901-42732901 on Build GRCh38
rs958984124 Chr.3: 42732611-42732611 on Build GRCh38
rs756230729 Chr.3: 42732850-42732850 on Build GRCh38
rs542065348 Chr.3: 42732871-42732871 on Build GRCh38
rs780627607 Chr.3: 42732879-42732879 on Build GRCh38
rs766336445 Chr.3: 42732910-42732910 on Build GRCh38

View Full Product Details