Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49550810-49551319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSN-AS2
Entrez Gene ID
100132677
Gene Name
BSN antisense RNA 2 (head to head)
Gene Aliases
-
UniGene
Hs.435651
Gene Chromosome Location
Chr.3: 49549306-49554366 on Build GRCh38
Transcripts
NR_038866.1

SNP Detail

SNP ID Chromosome Location
rs534728621 Chr.3: 49550846-49550846 on Build GRCh38
rs756069801 Chr.3: 49550967-49550968 on Build GRCh38
rs888636002 Chr.3: 49550951-49550951 on Build GRCh38
rs956665989 Chr.3: 49551037-49551037 on Build GRCh38
rs541714060 Chr.3: 49551007-49551007 on Build GRCh38
rs1019533294 Chr.3: 49551176-49551176 on Build GRCh38
rs574106523 Chr.3: 49550860-49550860 on Build GRCh38
rs903683428 Chr.3: 49551008-49551008 on Build GRCh38
rs930268169 Chr.3: 49551253-49551253 on Build GRCh38
rs952890910 Chr.3: 49551237-49551237 on Build GRCh38
rs541517428 Chr.3: 49551279-49551279 on Build GRCh38
rs896424539 Chr.3: 49550871-49550871 on Build GRCh38
rs911846525 Chr.3: 49551149-49551149 on Build GRCh38
rs745855159 Chr.3: 49550964-49550964 on Build GRCh38
rs540408422 Chr.3: 49551136-49551136 on Build GRCh38
rs930382763 Chr.3: 49550915-49550915 on Build GRCh38
rs564322176 Chr.3: 49551131-49551131 on Build GRCh38
rs1048821536 Chr.3: 49550950-49550952 on Build GRCh38
rs1012818664 Chr.3: 49550968-49550968 on Build GRCh38
rs138255404 Chr.3: 49551236-49551236 on Build GRCh38
rs947915430 Chr.3: 49550916-49550916 on Build GRCh38
rs779500887 Chr.3: 49550982-49550982 on Build GRCh38
rs1027148272 Chr.3: 49551235-49551235 on Build GRCh38
rs918946790 Chr.3: 49551249-49551249 on Build GRCh38
rs1030967408 Chr.3: 49551034-49551034 on Build GRCh38
rs1023914884 Chr.3: 49550972-49550972 on Build GRCh38
rs762052059 Chr.3: 49551031-49551031 on Build GRCh38
rs989541895 Chr.3: 49551047-49551047 on Build GRCh38
rs944629429 Chr.3: 49551188-49551188 on Build GRCh38
rs940354738 Chr.3: 49550852-49550852 on Build GRCh38
rs975628276 Chr.3: 49551205-49551205 on Build GRCh38
rs575075177 Chr.3: 49551099-49551099 on Build GRCh38
rs768950815 Chr.3: 49550872-49550872 on Build GRCh38
rs192375128 Chr.3: 49551106-49551106 on Build GRCh38
rs964269860 Chr.3: 49551197-49551197 on Build GRCh38
rs556886511 Chr.3: 49551081-49551081 on Build GRCh38
rs552651365 Chr.3: 49550847-49550847 on Build GRCh38
rs1048873246 Chr.3: 49551285-49551285 on Build GRCh38

View Full Product Details