Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 7323639-7324142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66B
Entrez Gene ID
100128890
Gene Name
family with sequence similarity 66 member B
Gene Aliases
FAM66E
UniGene
Hs.694680
Gene Chromosome Location
Chr.8: 7301611-7355354 on Build GRCh38
Transcripts
NR_027423.1

SNP Detail

SNP ID Chromosome Location
rs1046277109 Chr.8: 7323990-7323990 on Build GRCh38
rs182932329 Chr.8: 7324112-7324112 on Build GRCh38
rs953586467 Chr.8: 7323834-7323834 on Build GRCh38
rs35859946 Chr.8: 7323879-7323879 on Build GRCh38
rs1014559126 Chr.8: 7324014-7324014 on Build GRCh38
rs1005042159 Chr.8: 7324014-7324016 on Build GRCh38
rs947897468 Chr.8: 7323965-7323965 on Build GRCh38
rs894960712 Chr.8: 7323963-7323963 on Build GRCh38
rs898861494 Chr.8: 7324036-7324036 on Build GRCh38
rs895005151 Chr.8: 7323676-7323676 on Build GRCh38
rs530525636 Chr.8: 7324081-7324081 on Build GRCh38
rs1054149349 Chr.8: 7323958-7323958 on Build GRCh38
rs974072410 Chr.8: 7323833-7323833 on Build GRCh38
rs1029683309 Chr.8: 7324069-7324069 on Build GRCh38
rs925952065 Chr.8: 7323942-7323942 on Build GRCh38
rs752980394 Chr.8: 7323762-7323762 on Build GRCh38
rs936592152 Chr.8: 7323954-7323954 on Build GRCh38
rs112095013 Chr.8: 7323814-7323814 on Build GRCh38
rs946149669 Chr.8: 7323887-7323887 on Build GRCh38
rs977427076 Chr.8: 7323893-7323893 on Build GRCh38
rs1024753517 Chr.8: 7323757-7323757 on Build GRCh38
rs34791945 Chr.8: 7323995-7323997 on Build GRCh38
rs963625542 Chr.8: 7323822-7323822 on Build GRCh38
rs1004521633 Chr.8: 7323795-7323795 on Build GRCh38
rs1011998617 Chr.8: 7323691-7323691 on Build GRCh38
rs1015021474 Chr.8: 7323800-7323800 on Build GRCh38
rs912008667 Chr.8: 7323875-7323875 on Build GRCh38
rs906257472 Chr.8: 7324007-7324007 on Build GRCh38
rs987500035 Chr.8: 7323855-7323855 on Build GRCh38
rs564903907 Chr.8: 7324063-7324063 on Build GRCh38
rs953535856 Chr.8: 7324105-7324105 on Build GRCh38

View Full Product Details