Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 101075071-101075574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ42969
Entrez Gene ID
441374
Gene Name
uncharacterized LOC441374
Gene Aliases
-
UniGene
Hs.652045
Gene Chromosome Location
Chr.8: 101052054-101076251 on Build GRCh38
Transcripts
NR_033962.1

SNP Detail

SNP ID Chromosome Location
rs932110604 Chr.8: 101075192-101075192 on Build GRCh38
rs552046674 Chr.8: 101075419-101075419 on Build GRCh38
rs959867617 Chr.8: 101075135-101075135 on Build GRCh38
rs923340011 Chr.8: 101075169-101075169 on Build GRCh38
rs139400364 Chr.8: 101075230-101075230 on Build GRCh38
rs12679969 Chr.8: 101075321-101075321 on Build GRCh38
rs1057329244 Chr.8: 101075289-101075289 on Build GRCh38
rs1015941699 Chr.8: 101075380-101075380 on Build GRCh38
rs978935959 Chr.8: 101075451-101075451 on Build GRCh38
rs1050412622 Chr.8: 101075214-101075214 on Build GRCh38
rs967207493 Chr.8: 101075449-101075449 on Build GRCh38
rs1041272972 Chr.8: 101075344-101075344 on Build GRCh38
rs72669519 Chr.8: 101075501-101075501 on Build GRCh38
rs763050511 Chr.8: 101075121-101075121 on Build GRCh38
rs751059995 Chr.8: 101075200-101075200 on Build GRCh38
rs992588105 Chr.8: 101075144-101075144 on Build GRCh38
rs1012581141 Chr.8: 101075518-101075518 on Build GRCh38
rs1012654757 Chr.8: 101075343-101075343 on Build GRCh38
rs938969290 Chr.8: 101075492-101075500 on Build GRCh38
rs779398699 Chr.8: 101075327-101075327 on Build GRCh38
rs549806851 Chr.8: 101075452-101075452 on Build GRCh38
rs367765960 Chr.8: 101075418-101075419 on Build GRCh38
rs148652955 Chr.8: 101075476-101075476 on Build GRCh38
rs1026573873 Chr.8: 101075354-101075354 on Build GRCh38
rs144040336 Chr.8: 101075423-101075423 on Build GRCh38
rs978520752 Chr.8: 101075235-101075235 on Build GRCh38
rs888086255 Chr.8: 101075542-101075542 on Build GRCh38
rs985825456 Chr.8: 101075489-101075489 on Build GRCh38
rs897311609 Chr.8: 101075348-101075348 on Build GRCh38
rs754544954 Chr.8: 101075201-101075201 on Build GRCh38
rs1034237686 Chr.8: 101075414-101075414 on Build GRCh38
rs145138635 Chr.8: 101075418-101075418 on Build GRCh38
rs921148751 Chr.8: 101075415-101075415 on Build GRCh38
rs918765166 Chr.8: 101075537-101075537 on Build GRCh38
rs183843014 Chr.8: 101075164-101075164 on Build GRCh38
rs539684702 Chr.8: 101075238-101075238 on Build GRCh38
rs535583721 Chr.8: 101075502-101075502 on Build GRCh38
rs113488334 Chr.8: 101075145-101075145 on Build GRCh38
rs192401007 Chr.8: 101075147-101075147 on Build GRCh38
rs765130493 Chr.8: 101075118-101075118 on Build GRCh38
rs111574828 Chr.8: 101075418-101075418 on Build GRCh38
rs370673026 Chr.8: 101075508-101075508 on Build GRCh38
rs59158548 Chr.8: 101075388-101075388 on Build GRCh38
rs911441262 Chr.8: 101075322-101075322 on Build GRCh38
rs925685416 Chr.8: 101075455-101075455 on Build GRCh38
rs372930822 Chr.8: 101075258-101075258 on Build GRCh38
rs913060213 Chr.8: 101075165-101075165 on Build GRCh38
rs750298366 Chr.8: 101075120-101075120 on Build GRCh38
rs994691150 Chr.8: 101075358-101075358 on Build GRCh38
rs562461845 Chr.8: 101075262-101075262 on Build GRCh38
rs149942957 Chr.8: 101075377-101075377 on Build GRCh38
rs117385462 Chr.8: 101075397-101075397 on Build GRCh38

View Full Product Details