Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39475421-39475878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM3A
Entrez Gene ID
1587
Gene Name
ADAM metallopeptidase domain 3A (pseudogene)
Gene Aliases
ADAM3, CYRN1, tMDCI
UniGene
Hs.98848
Gene Chromosome Location
Chr.8: 39451045-39522989 on Build GRCh38
Transcripts
NR_001569.3 NR_024106.2 NR_024107.2 NR_073423.1

SNP Detail

SNP ID Chromosome Location
rs1036158068 Chr.8: 39475501-39475501 on Build GRCh38
rs762716622 Chr.8: 39475466-39475466 on Build GRCh38
rs150158281 Chr.8: 39475720-39475720 on Build GRCh38
rs746408469 Chr.8: 39475789-39475789 on Build GRCh38
rs147348375 Chr.8: 39475844-39475844 on Build GRCh38
rs552225920 Chr.8: 39475812-39475812 on Build GRCh38
rs749652023 Chr.8: 39475616-39475616 on Build GRCh38
rs890588885 Chr.8: 39475765-39475765 on Build GRCh38
rs527673646 Chr.8: 39475774-39475774 on Build GRCh38
rs771443995 Chr.8: 39475689-39475689 on Build GRCh38
rs138681189 Chr.8: 39475761-39475761 on Build GRCh38
rs182774876 Chr.8: 39475717-39475717 on Build GRCh38
rs1051928604 Chr.8: 39475625-39475625 on Build GRCh38
rs192911808 Chr.8: 39475821-39475821 on Build GRCh38
rs142713841 Chr.8: 39475834-39475834 on Build GRCh38
rs187743556 Chr.8: 39475773-39475773 on Build GRCh38
rs987256731 Chr.8: 39475845-39475845 on Build GRCh38
rs373190056 Chr.8: 39475568-39475568 on Build GRCh38
rs932172870 Chr.8: 39475593-39475593 on Build GRCh38
rs112427411 Chr.8: 39475509-39475509 on Build GRCh38
rs779242427 Chr.8: 39475710-39475710 on Build GRCh38

View Full Product Details