Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81916099-81916652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D5P
Entrez Gene ID
347127
Gene Name
SPATA31 subfamily D member 5, pseudogene
Gene Aliases
FAM75D5, FAM75D5P
UniGene
Hs.215235
Gene Chromosome Location
Chr.9: 81913437-81919927 on Build GRCh38
Transcripts
NR_026851.1

SNP Detail

SNP ID Chromosome Location
rs1015531913 Chr.9: 81916503-81916503 on Build GRCh38
rs901785950 Chr.9: 81916630-81916630 on Build GRCh38
rs754699351 Chr.9: 81916317-81916317 on Build GRCh38
rs764645302 Chr.9: 81916230-81916230 on Build GRCh38
rs760118993 Chr.9: 81916202-81916202 on Build GRCh38
rs779060070 Chr.9: 81916303-81916303 on Build GRCh38
rs768000296 Chr.9: 81916281-81916281 on Build GRCh38
rs762541211 Chr.9: 81916179-81916179 on Build GRCh38
rs777746561 Chr.9: 81916229-81916229 on Build GRCh38
rs115919238 Chr.9: 81916464-81916464 on Build GRCh38
rs577288260 Chr.9: 81916460-81916460 on Build GRCh38
rs776113440 Chr.9: 81916156-81916156 on Build GRCh38
rs746870719 Chr.9: 81916247-81916247 on Build GRCh38
rs777109402 Chr.9: 81916538-81916538 on Build GRCh38
rs570222889 Chr.9: 81916235-81916235 on Build GRCh38
rs527288487 Chr.9: 81916147-81916147 on Build GRCh38
rs146387432 Chr.9: 81916176-81916176 on Build GRCh38
rs774766687 Chr.9: 81916279-81916279 on Build GRCh38
rs780991981 Chr.9: 81916319-81916319 on Build GRCh38
rs754043426 Chr.9: 81916234-81916234 on Build GRCh38
rs945908382 Chr.9: 81916616-81916616 on Build GRCh38
rs544658322 Chr.9: 81916478-81916478 on Build GRCh38
rs186823698 Chr.9: 81916375-81916375 on Build GRCh38
rs992854201 Chr.9: 81916511-81916511 on Build GRCh38
rs777995423 Chr.9: 81916129-81916129 on Build GRCh38
rs905730732 Chr.9: 81916144-81916144 on Build GRCh38
rs1019385023 Chr.9: 81916187-81916187 on Build GRCh38
rs748025461 Chr.9: 81916325-81916325 on Build GRCh38
rs748080431 Chr.9: 81916256-81916256 on Build GRCh38
rs756197463 Chr.9: 81916227-81916227 on Build GRCh38
rs769304467 Chr.9: 81916175-81916175 on Build GRCh38
rs935758227 Chr.9: 81916591-81916591 on Build GRCh38
rs781394955 Chr.9: 81916124-81916124 on Build GRCh38
rs750653570 Chr.9: 81916316-81916316 on Build GRCh38
rs768566512 Chr.9: 81916251-81916251 on Build GRCh38
rs1044136148 Chr.9: 81916333-81916333 on Build GRCh38
rs771860906 Chr.9: 81916141-81916141 on Build GRCh38
rs112844174 Chr.9: 81916596-81916596 on Build GRCh38
rs200456940 Chr.9: 81916264-81916264 on Build GRCh38
rs761953485 Chr.9: 81916285-81916285 on Build GRCh38
rs779922355 Chr.9: 81916276-81916277 on Build GRCh38
rs554103133 Chr.9: 81916332-81916332 on Build GRCh38
rs527940439 Chr.9: 81916569-81916569 on Build GRCh38
rs564083601 Chr.9: 81916607-81916607 on Build GRCh38
rs568215921 Chr.9: 81916280-81916280 on Build GRCh38
rs753694634 Chr.9: 81916299-81916299 on Build GRCh38
rs753271677 Chr.9: 81916207-81916207 on Build GRCh38
rs1999662 Chr.9: 81916532-81916532 on Build GRCh38
rs979784599 Chr.9: 81916552-81916552 on Build GRCh38
rs766747310 Chr.9: 81916184-81916184 on Build GRCh38
rs556514551 Chr.9: 81916499-81916499 on Build GRCh38
rs149370879 Chr.9: 81916598-81916598 on Build GRCh38
rs558943887 Chr.9: 81916438-81916438 on Build GRCh38
rs138774243 Chr.9: 81916551-81916551 on Build GRCh38
rs768198836 Chr.9: 81916204-81916204 on Build GRCh38
rs755702165 Chr.9: 81916238-81916238 on Build GRCh38
rs761367233 Chr.9: 81916158-81916158 on Build GRCh38
rs765263344 Chr.9: 81916356-81916356 on Build GRCh38
rs569017696 Chr.9: 81916342-81916342 on Build GRCh38
rs996135240 Chr.9: 81916430-81916430 on Build GRCh38
rs753904030 Chr.9: 81916296-81916296 on Build GRCh38
rs749258768 Chr.9: 81916277-81916278 on Build GRCh38
rs763139427 Chr.9: 81916199-81916199 on Build GRCh38
rs549276430 Chr.9: 81916336-81916336 on Build GRCh38
rs757495130 Chr.9: 81916297-81916297 on Build GRCh38
rs749478205 Chr.9: 81916137-81916137 on Build GRCh38
rs748308261 Chr.9: 81916125-81916125 on Build GRCh38
rs540070614 Chr.9: 81916154-81916154 on Build GRCh38
rs559745668 Chr.9: 81916128-81916128 on Build GRCh38
rs945991433 Chr.9: 81916393-81916393 on Build GRCh38
rs753218408 Chr.9: 81916282-81916282 on Build GRCh38
rs531324075 Chr.9: 81916620-81916620 on Build GRCh38
rs927014905 Chr.9: 81916589-81916589 on Build GRCh38
rs908091028 Chr.9: 81916245-81916245 on Build GRCh38
rs749871555 Chr.9: 81916174-81916176 on Build GRCh38
rs755594474 Chr.9: 81916123-81916123 on Build GRCh38
rs901817359 Chr.9: 81916416-81916416 on Build GRCh38
rs377687087 Chr.9: 81916286-81916286 on Build GRCh38
rs1040365509 Chr.9: 81916399-81916399 on Build GRCh38
rs549733735 Chr.9: 81916278-81916278 on Build GRCh38
rs534192575 Chr.9: 81916379-81916379 on Build GRCh38
rs779868152 Chr.9: 81916244-81916244 on Build GRCh38
rs772615903 Chr.9: 81916270-81916270 on Build GRCh38
rs762640221 Chr.9: 81916274-81916274 on Build GRCh38
rs891691866 Chr.9: 81916384-81916384 on Build GRCh38
rs542163492 Chr.9: 81916547-81916547 on Build GRCh38
rs1010223706 Chr.9: 81916186-81916186 on Build GRCh38
rs535420364 Chr.9: 81916283-81916283 on Build GRCh38
rs766170151 Chr.9: 81916298-81916298 on Build GRCh38

View Full Product Details