Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171339332-171339884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO4
Entrez Gene ID
2329
Gene Name
flavin containing monooxygenase 4
Gene Aliases
FMO2
UniGene
Hs.386502
Gene Chromosome Location
Chr.1: 171314131-171342084 on Build GRCh38
Transcripts
NM_002022.2 XM_005245045.3 XM_005245046.3 XM_005245048.3 XM_006711243.3 XM_011509347.2 XM_011509348.2 NR_002719.2

Gene Symbol
TOP1P1
Entrez Gene ID
7151
Gene Name
topoisomerase (DNA) I pseudogene 1
Gene Aliases
-
UniGene
Hs.661092
Gene Chromosome Location
Chr.1: 171338896-171341326 on Build GRCh38
Transcripts
NM_002022.2 XM_005245045.3 XM_005245046.3 XM_005245048.3 XM_006711243.3 XM_011509347.2 XM_011509348.2 NR_002719.2

SNP Detail

SNP ID Chromosome Location
rs771717378 Chr.1: 171339442-171339442 on Build GRCh38
rs1003089110 Chr.1: 171339813-171339813 on Build GRCh38
rs551650246 Chr.1: 171339645-171339645 on Build GRCh38
rs971242979 Chr.1: 171339443-171339443 on Build GRCh38
rs940338160 Chr.1: 171339430-171339430 on Build GRCh38
rs878986060 Chr.1: 171339412-171339412 on Build GRCh38
rs565773404 Chr.1: 171339728-171339728 on Build GRCh38
rs774089669 Chr.1: 171339409-171339409 on Build GRCh38
rs71635698 Chr.1: 171339711-171339711 on Build GRCh38
rs958895099 Chr.1: 171339647-171339647 on Build GRCh38
rs61731205 Chr.1: 171339527-171339527 on Build GRCh38
rs975033037 Chr.1: 171339526-171339526 on Build GRCh38
rs956159597 Chr.1: 171339489-171339489 on Build GRCh38
rs907136063 Chr.1: 171339811-171339811 on Build GRCh38
rs1032136613 Chr.1: 171339478-171339478 on Build GRCh38
rs889063410 Chr.1: 171339820-171339820 on Build GRCh38
rs939810214 Chr.1: 171339675-171339675 on Build GRCh38
rs560542968 Chr.1: 171339488-171339488 on Build GRCh38
rs1036055585 Chr.1: 171339432-171339432 on Build GRCh38
rs964953145 Chr.1: 171339522-171339522 on Build GRCh38
rs1044253745 Chr.1: 171339807-171339807 on Build GRCh38
rs111814817 Chr.1: 171339391-171339391 on Build GRCh38
rs2223477 Chr.1: 171339693-171339693 on Build GRCh38
rs981720470 Chr.1: 171339550-171339550 on Build GRCh38
rs184305603 Chr.1: 171339627-171339627 on Build GRCh38
rs530551371 Chr.1: 171339788-171339788 on Build GRCh38
rs879149447 Chr.1: 171339480-171339480 on Build GRCh38
rs908265869 Chr.1: 171339649-171339649 on Build GRCh38
rs922978277 Chr.1: 171339766-171339766 on Build GRCh38
rs990531668 Chr.1: 171339492-171339492 on Build GRCh38
rs879224953 Chr.1: 171339472-171339472 on Build GRCh38
rs879191529 Chr.1: 171339704-171339704 on Build GRCh38
rs373762925 Chr.1: 171339849-171339849 on Build GRCh38
rs560535572 Chr.1: 171339400-171339400 on Build GRCh38
rs571792410 Chr.1: 171339451-171339451 on Build GRCh38
rs982003712 Chr.1: 171339373-171339373 on Build GRCh38
rs937048150 Chr.1: 171339405-171339405 on Build GRCh38
rs1048850605 Chr.1: 171339407-171339407 on Build GRCh38
rs879026898 Chr.1: 171339655-171339655 on Build GRCh38
rs45589839 Chr.1: 171339758-171339758 on Build GRCh38
rs898401157 Chr.1: 171339402-171339402 on Build GRCh38
rs565141783 Chr.1: 171339650-171339650 on Build GRCh38
rs933443907 Chr.1: 171339595-171339595 on Build GRCh38
rs914598487 Chr.1: 171339493-171339493 on Build GRCh38
rs745849939 Chr.1: 171339427-171339427 on Build GRCh38
rs972027318 Chr.1: 171339535-171339535 on Build GRCh38
rs888956476 Chr.1: 171339453-171339453 on Build GRCh38
rs547535989 Chr.1: 171339695-171339695 on Build GRCh38
rs567767389 Chr.1: 171339840-171339840 on Build GRCh38
rs1053573505 Chr.1: 171339842-171339842 on Build GRCh38
rs45556934 Chr.1: 171339742-171339742 on Build GRCh38
rs542342406 Chr.1: 171339456-171339456 on Build GRCh38
rs878899645 Chr.1: 171339434-171339434 on Build GRCh38
rs1004257765 Chr.1: 171339369-171339369 on Build GRCh38
rs776484014 Chr.1: 171339844-171339844 on Build GRCh38
rs933192484 Chr.1: 171339775-171339775 on Build GRCh38
rs781444364 Chr.1: 171339455-171339455 on Build GRCh38

View Full Product Details