Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19329271-19329833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P3
Entrez Gene ID
100101938
Gene Name
ankyrin repeat domain 26 pseudogene 3
Gene Aliases
LINC00414
UniGene
Hs.129283
Gene Chromosome Location
Chr.13: 19262800-19344973 on Build GRCh38
Transcripts
NR_027248.3

SNP Detail

SNP ID Chromosome Location
rs528575334 Chr.13: 19329687-19329687 on Build GRCh38
rs184963286 Chr.13: 19329734-19329734 on Build GRCh38
rs936509362 Chr.13: 19329534-19329534 on Build GRCh38
rs886488639 Chr.13: 19329704-19329704 on Build GRCh38
rs778926941 Chr.13: 19329665-19329665 on Build GRCh38
rs748169643 Chr.13: 19329781-19329781 on Build GRCh38
rs991464432 Chr.13: 19329764-19329764 on Build GRCh38
rs1046941971 Chr.13: 19329322-19329322 on Build GRCh38
rs982681188 Chr.13: 19329601-19329601 on Build GRCh38
rs926936118 Chr.13: 19329621-19329621 on Build GRCh38
rs889731098 Chr.13: 19329569-19329569 on Build GRCh38
rs115597427 Chr.13: 19329384-19329384 on Build GRCh38
rs901834668 Chr.13: 19329683-19329683 on Build GRCh38
rs990517616 Chr.13: 19329471-19329471 on Build GRCh38
rs997997144 Chr.13: 19329374-19329374 on Build GRCh38
rs550440755 Chr.13: 19329480-19329480 on Build GRCh38
rs1007574675 Chr.13: 19329296-19329296 on Build GRCh38
rs893833745 Chr.13: 19329810-19329810 on Build GRCh38
rs938202428 Chr.13: 19329674-19329674 on Build GRCh38
rs761694112 Chr.13: 19329731-19329754 on Build GRCh38
rs1026403976 Chr.13: 19329703-19329703 on Build GRCh38
rs78797075 Chr.13: 19329360-19329360 on Build GRCh38
rs562124863 Chr.13: 19329589-19329589 on Build GRCh38
rs970794638 Chr.13: 19329775-19329775 on Build GRCh38
rs71985553 Chr.13: 19329729-19329754 on Build GRCh38
rs76974310 Chr.13: 19329727-19329754 on Build GRCh38
rs2065881 Chr.13: 19329807-19329807 on Build GRCh38
rs564645014 Chr.13: 19329533-19329533 on Build GRCh38
rs373221892 Chr.13: 19329754-19329755 on Build GRCh38
rs2065882 Chr.13: 19329808-19329808 on Build GRCh38
rs899769792 Chr.13: 19329785-19329785 on Build GRCh38
rs879875677 Chr.13: 19329807-19329808 on Build GRCh38
rs886922344 Chr.13: 19329323-19329323 on Build GRCh38
rs1006407404 Chr.13: 19329720-19329720 on Build GRCh38
rs867655633 Chr.13: 19329754-19329754 on Build GRCh38
rs9319349 Chr.13: 19329440-19329440 on Build GRCh38
rs146923826 Chr.13: 19329529-19329529 on Build GRCh38
rs575426545 Chr.13: 19329448-19329448 on Build GRCh38
rs984918271 Chr.13: 19329456-19329456 on Build GRCh38
rs909471354 Chr.13: 19329464-19329464 on Build GRCh38
rs1042111293 Chr.13: 19329504-19329504 on Build GRCh38
rs949191125 Chr.13: 19329495-19329495 on Build GRCh38
rs776373974 Chr.13: 19329725-19329725 on Build GRCh38
rs997639923 Chr.13: 19329689-19329689 on Build GRCh38
rs768453938 Chr.13: 19329590-19329590 on Build GRCh38
rs151152630 Chr.13: 19329343-19329343 on Build GRCh38
rs768093811 Chr.13: 19329728-19329729 on Build GRCh38
rs931066833 Chr.13: 19329697-19329697 on Build GRCh38
rs140109260 Chr.13: 19329355-19329355 on Build GRCh38
rs561710736 Chr.13: 19329766-19329766 on Build GRCh38
rs1053524226 Chr.13: 19329568-19329568 on Build GRCh38
rs959861339 Chr.13: 19329470-19329470 on Build GRCh38
rs34848933 Chr.13: 19329807-19329808 on Build GRCh38
rs749513042 Chr.13: 19329441-19329441 on Build GRCh38
rs970950990 Chr.13: 19329597-19329597 on Build GRCh38

View Full Product Details