Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 14338761-14339314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F35P
Entrez Gene ID
284233
Gene Name
cytochrome P450 family 4 subfamily F member 35, pseudogene
Gene Aliases
-
UniGene
Hs.607579
Gene Chromosome Location
Chr.18: 14337423-14342524 on Build GRCh38
Transcripts
NR_026756.1

SNP Detail

SNP ID Chromosome Location
rs796543004 Chr.18: 14339028-14339028 on Build GRCh38
rs923552399 Chr.18: 14339012-14339012 on Build GRCh38
rs758780001 Chr.18: 14339205-14339205 on Build GRCh38
rs34742814 Chr.18: 14338836-14338837 on Build GRCh38
rs567748205 Chr.18: 14339197-14339197 on Build GRCh38
rs901803269 Chr.18: 14338809-14338809 on Build GRCh38
rs889359833 Chr.18: 14338786-14338786 on Build GRCh38
rs1016751031 Chr.18: 14338870-14338870 on Build GRCh38
rs190789220 Chr.18: 14339284-14339284 on Build GRCh38
rs374286298 Chr.18: 14338844-14338844 on Build GRCh38
rs1969142 Chr.18: 14339095-14339095 on Build GRCh38
rs960930996 Chr.18: 14339154-14339154 on Build GRCh38
rs1002038610 Chr.18: 14338875-14338875 on Build GRCh38
rs1984679 Chr.18: 14339109-14339109 on Build GRCh38
rs796683659 Chr.18: 14338975-14338975 on Build GRCh38
rs368458041 Chr.18: 14338874-14338874 on Build GRCh38
rs185634099 Chr.18: 14339104-14339104 on Build GRCh38
rs563572467 Chr.18: 14339246-14339246 on Build GRCh38
rs979135173 Chr.18: 14339192-14339192 on Build GRCh38
rs796874969 Chr.18: 14339243-14339243 on Build GRCh38
rs532915546 Chr.18: 14338970-14338970 on Build GRCh38
rs910699712 Chr.18: 14338964-14338964 on Build GRCh38
rs534157945 Chr.18: 14339189-14339189 on Build GRCh38
rs374654227 Chr.18: 14339221-14339222 on Build GRCh38
rs1043941828 Chr.18: 14338785-14338785 on Build GRCh38
rs559630530 Chr.18: 14339124-14339124 on Build GRCh38
rs556389886 Chr.18: 14338799-14338799 on Build GRCh38
rs1006232057 Chr.18: 14339100-14339100 on Build GRCh38
rs993913472 Chr.18: 14338827-14338827 on Build GRCh38
rs576584340 Chr.18: 14338986-14338986 on Build GRCh38
rs1047987999 Chr.18: 14339230-14339230 on Build GRCh38
rs909179587 Chr.18: 14339231-14339231 on Build GRCh38
rs1040595999 Chr.18: 14339011-14339011 on Build GRCh38
rs569098407 Chr.18: 14339108-14339108 on Build GRCh38
rs771385645 Chr.18: 14338850-14338850 on Build GRCh38
rs1044453589 Chr.18: 14339239-14339239 on Build GRCh38
rs551664859 Chr.18: 14339112-14339112 on Build GRCh38
rs796887219 Chr.18: 14339010-14339010 on Build GRCh38
rs988252691 Chr.18: 14338920-14338920 on Build GRCh38
rs3927192 Chr.18: 14339022-14339022 on Build GRCh38
rs58630609 Chr.18: 14339191-14339191 on Build GRCh38
rs979190502 Chr.18: 14338953-14338953 on Build GRCh38
rs1034943243 Chr.18: 14339150-14339150 on Build GRCh38
rs576197175 Chr.18: 14339250-14339250 on Build GRCh38
rs868826835 Chr.18: 14338940-14338940 on Build GRCh38
rs576306779 Chr.18: 14338908-14338908 on Build GRCh38
rs866173248 Chr.18: 14338941-14338941 on Build GRCh38
rs138156812 Chr.18: 14339249-14339249 on Build GRCh38
rs896333825 Chr.18: 14339251-14339251 on Build GRCh38
rs779252409 Chr.18: 14339196-14339196 on Build GRCh38
rs2175665 Chr.18: 14339212-14339212 on Build GRCh38
rs565253894 Chr.18: 14338932-14338932 on Build GRCh38
rs141007590 Chr.18: 14339182-14339182 on Build GRCh38
rs747231231 Chr.18: 14339228-14339228 on Build GRCh38
rs1035358746 Chr.18: 14338881-14338881 on Build GRCh38
rs181357914 Chr.18: 14338836-14338836 on Build GRCh38
rs905999097 Chr.18: 14339244-14339244 on Build GRCh38
rs561927679 Chr.18: 14339099-14339099 on Build GRCh38
rs1020604416 Chr.18: 14339186-14339186 on Build GRCh38
rs942045754 Chr.18: 14339233-14339233 on Build GRCh38
rs796071975 Chr.18: 14338971-14338971 on Build GRCh38
rs753742209 Chr.18: 14339253-14339253 on Build GRCh38
rs796738792 Chr.18: 14339240-14339240 on Build GRCh38
rs887899681 Chr.18: 14338845-14338845 on Build GRCh38
rs761528779 Chr.18: 14339079-14339079 on Build GRCh38
rs558254353 Chr.18: 14338820-14338820 on Build GRCh38
rs753120582 Chr.18: 14339204-14339204 on Build GRCh38
rs560151939 Chr.18: 14339065-14339065 on Build GRCh38
rs543199147 Chr.18: 14338789-14338789 on Build GRCh38
rs964866511 Chr.18: 14339200-14339200 on Build GRCh38
rs776982363 Chr.18: 14338990-14338990 on Build GRCh38
rs146617792 Chr.18: 14339001-14339001 on Build GRCh38
rs913632629 Chr.18: 14338928-14338928 on Build GRCh38
rs545328936 Chr.18: 14338917-14338917 on Build GRCh38
rs929575485 Chr.18: 14339218-14339218 on Build GRCh38
rs34800058 Chr.18: 14338965-14338965 on Build GRCh38

View Full Product Details