Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10046265-10046820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3P1
Entrez Gene ID
388503
Gene Name
complement component 3 precursor pseudogene
Gene Aliases
CPLP
UniGene
Hs.655363
Gene Chromosome Location
Chr.19: 10041356-10074138 on Build GRCh38
Transcripts
NR_027300.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3772481 c.25G>A p.E9K Chr.19: 10046795-10046795 on Build GRCh38
1246844 c.29G>C p.S10T Chr.19: 10046799-10046799 on Build GRCh38
3528062 c.46T>C p.F16L Chr.19: 10046816-10046816 on Build GRCh38
6149801 c.3G>A p.M1I Chr.19: 10046773-10046773 on Build GRCh38
3528063 c.48C>T p.F16F Chr.19: 10046818-10046818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139683096 Chr.19: 10046778-10046778 on Build GRCh38
rs550423640 Chr.19: 10046597-10046597 on Build GRCh38
rs573555153 Chr.19: 10046475-10046475 on Build GRCh38
rs116673803 Chr.19: 10046751-10046751 on Build GRCh38
rs890046980 Chr.19: 10046767-10046767 on Build GRCh38
rs142012481 Chr.19: 10046451-10046451 on Build GRCh38
rs574172929 Chr.19: 10046407-10046407 on Build GRCh38
rs538154905 Chr.19: 10046510-10046510 on Build GRCh38
rs368068080 Chr.19: 10046731-10046731 on Build GRCh38
rs933898101 Chr.19: 10046315-10046315 on Build GRCh38
rs776612416 Chr.19: 10046776-10046776 on Build GRCh38
rs749179698 Chr.19: 10046733-10046733 on Build GRCh38
rs1045405608 Chr.19: 10046324-10046324 on Build GRCh38
rs201490083 Chr.19: 10046750-10046750 on Build GRCh38
rs766758502 Chr.19: 10046786-10046786 on Build GRCh38
rs368636103 Chr.19: 10046781-10046781 on Build GRCh38
rs767198878 Chr.19: 10046789-10046789 on Build GRCh38
rs754392901 Chr.19: 10046769-10046769 on Build GRCh38
rs986759917 Chr.19: 10046558-10046558 on Build GRCh38
rs199654654 Chr.19: 10046797-10046797 on Build GRCh38
rs146373238 Chr.19: 10046711-10046711 on Build GRCh38
rs939578864 Chr.19: 10046645-10046645 on Build GRCh38
rs1045215729 Chr.19: 10046426-10046426 on Build GRCh38
rs778811703 Chr.19: 10046749-10046749 on Build GRCh38
rs1045199269 Chr.19: 10046616-10046616 on Build GRCh38
rs750865743 Chr.19: 10046461-10046461 on Build GRCh38
rs926974265 Chr.19: 10046361-10046361 on Build GRCh38
rs555241329 Chr.19: 10046471-10046471 on Build GRCh38
rs1022358325 Chr.19: 10046512-10046512 on Build GRCh38
rs955262472 Chr.19: 10046556-10046556 on Build GRCh38
rs1042969186 Chr.19: 10046423-10046423 on Build GRCh38
rs902755907 Chr.19: 10046450-10046450 on Build GRCh38
rs939675814 Chr.19: 10046582-10046582 on Build GRCh38
rs766987156 Chr.19: 10046727-10046727 on Build GRCh38
rs150693493 Chr.19: 10046795-10046795 on Build GRCh38
rs11883054 Chr.19: 10046543-10046543 on Build GRCh38
rs1011046098 Chr.19: 10046410-10046410 on Build GRCh38
rs763379616 Chr.19: 10046784-10046784 on Build GRCh38
rs868367999 Chr.19: 10046710-10046710 on Build GRCh38
rs1054250952 Chr.19: 10046373-10046373 on Build GRCh38
rs1054543004 Chr.19: 10046682-10046682 on Build GRCh38
rs757122801 Chr.19: 10046744-10046744 on Build GRCh38
rs986864268 Chr.19: 10046318-10046318 on Build GRCh38
rs756040326 Chr.19: 10046577-10046577 on Build GRCh38
rs766756945 Chr.19: 10046706-10046706 on Build GRCh38
rs369064208 Chr.19: 10046403-10046403 on Build GRCh38
rs550462099 Chr.19: 10046780-10046780 on Build GRCh38
rs187467635 Chr.19: 10046585-10046585 on Build GRCh38
rs774678235 Chr.19: 10046788-10046788 on Build GRCh38
rs375395421 Chr.19: 10046633-10046633 on Build GRCh38
rs560604560 Chr.19: 10046694-10046694 on Build GRCh38
rs539124735 Chr.19: 10046364-10046364 on Build GRCh38
rs936951145 Chr.19: 10046601-10046601 on Build GRCh38
rs556051721 Chr.19: 10046523-10046523 on Build GRCh38
rs879451104 Chr.19: 10046436-10046436 on Build GRCh38
rs566106929 Chr.19: 10046421-10046421 on Build GRCh38
rs373620015 Chr.19: 10046730-10046730 on Build GRCh38
rs918438176 Chr.19: 10046570-10046570 on Build GRCh38
rs926943582 Chr.19: 10046599-10046599 on Build GRCh38
rs1056693041 Chr.19: 10046677-10046677 on Build GRCh38
rs542398440 Chr.19: 10046327-10046327 on Build GRCh38
rs898265425 Chr.19: 10046404-10046404 on Build GRCh38
rs142999843 Chr.19: 10046794-10046794 on Build GRCh38
rs919639891 Chr.19: 10046591-10046591 on Build GRCh38
rs972566255 Chr.19: 10046544-10046544 on Build GRCh38
rs374079030 Chr.19: 10046402-10046402 on Build GRCh38
rs968132111 Chr.19: 10046514-10046514 on Build GRCh38

View Full Product Details