Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23359481-23360036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES5AP1
Entrez Gene ID
649264
Gene Name
carboxylesterase 5A pseudogene 1
Gene Aliases
-
UniGene
Hs.600549
Gene Chromosome Location
Chr.22: 23359606-23382126 on Build GRCh38
Transcripts
NR_037839.1

SNP Detail

SNP ID Chromosome Location
rs6003651 Chr.22: 23359576-23359576 on Build GRCh38
rs1032501784 Chr.22: 23359859-23359859 on Build GRCh38
rs747096774 Chr.22: 23359719-23359719 on Build GRCh38
rs867362601 Chr.22: 23359878-23359878 on Build GRCh38
rs953381288 Chr.22: 23359767-23359767 on Build GRCh38
rs553194381 Chr.22: 23359723-23359723 on Build GRCh38
rs76411639 Chr.22: 23359512-23359512 on Build GRCh38
rs972313157 Chr.22: 23359562-23359562 on Build GRCh38
rs949015339 Chr.22: 23359571-23359572 on Build GRCh38
rs201199473 Chr.22: 23359718-23359718 on Build GRCh38
rs113651797 Chr.22: 23359565-23359565 on Build GRCh38
rs781708030 Chr.22: 23359787-23359787 on Build GRCh38
rs181011090 Chr.22: 23359770-23359770 on Build GRCh38
rs775752487 Chr.22: 23359980-23359980 on Build GRCh38
rs942211955 Chr.22: 23359522-23359522 on Build GRCh38
rs937694829 Chr.22: 23359986-23359986 on Build GRCh38
rs771641485 Chr.22: 23360003-23360003 on Build GRCh38
rs778548780 Chr.22: 23359566-23359566 on Build GRCh38
rs532540007 Chr.22: 23359894-23359894 on Build GRCh38
rs571553916 Chr.22: 23359577-23359577 on Build GRCh38
rs10427608 Chr.22: 23359943-23359943 on Build GRCh38
rs970944712 Chr.22: 23359849-23359849 on Build GRCh38
rs1030329648 Chr.22: 23359762-23359762 on Build GRCh38
rs770995166 Chr.22: 23359724-23359724 on Build GRCh38
rs575542754 Chr.22: 23359806-23359806 on Build GRCh38
rs558765100 Chr.22: 23359777-23359777 on Build GRCh38
rs6003652 Chr.22: 23359617-23359617 on Build GRCh38
rs560839626 Chr.22: 23359865-23359865 on Build GRCh38
rs982356668 Chr.22: 23359942-23359942 on Build GRCh38
rs559619583 Chr.22: 23359517-23359517 on Build GRCh38
rs528620047 Chr.22: 23359570-23359570 on Build GRCh38
rs567626299 Chr.22: 23359681-23359681 on Build GRCh38
rs6003653 Chr.22: 23359998-23359998 on Build GRCh38
rs202236671 Chr.22: 23359587-23359587 on Build GRCh38
rs1009829800 Chr.22: 23359664-23359664 on Build GRCh38
rs544259298 Chr.22: 23359852-23359852 on Build GRCh38
rs538984139 Chr.22: 23359775-23359775 on Build GRCh38
rs1053970308 Chr.22: 23359588-23359588 on Build GRCh38
rs998143083 Chr.22: 23359707-23359707 on Build GRCh38
rs144148701 Chr.22: 23359702-23359702 on Build GRCh38
rs563590538 Chr.22: 23359592-23359592 on Build GRCh38
rs746353512 Chr.22: 23359843-23359843 on Build GRCh38

View Full Product Details