Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 75421678-75422225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86DP
Entrez Gene ID
692099
Gene Name
family with sequence similarity 86, member D, pseudogene
Gene Aliases
FAM86D
UniGene
Hs.591652
Gene Chromosome Location
Chr.3: 75421552-75435115 on Build GRCh38
Transcripts
NR_024241.1

SNP Detail

SNP ID Chromosome Location
rs11717001 Chr.3: 75422144-75422144 on Build GRCh38
rs750906192 Chr.3: 75421905-75421906 on Build GRCh38
rs1036225756 Chr.3: 75422094-75422094 on Build GRCh38
rs149189373 Chr.3: 75421852-75421852 on Build GRCh38
rs578022619 Chr.3: 75421983-75421983 on Build GRCh38
rs553195471 Chr.3: 75421728-75421728 on Build GRCh38
rs867473142 Chr.3: 75422061-75422061 on Build GRCh38
rs940582160 Chr.3: 75421849-75421849 on Build GRCh38
rs578192091 Chr.3: 75421753-75421753 on Build GRCh38
rs547741720 Chr.3: 75422133-75422133 on Build GRCh38
rs756712546 Chr.3: 75421971-75421971 on Build GRCh38
rs143367038 Chr.3: 75421856-75421856 on Build GRCh38
rs199534045 Chr.3: 75421890-75421890 on Build GRCh38
rs199754293 Chr.3: 75421845-75421845 on Build GRCh38
rs180987196 Chr.3: 75422138-75422138 on Build GRCh38
rs71627425 Chr.3: 75421976-75421977 on Build GRCh38
rs556705178 Chr.3: 75422189-75422189 on Build GRCh38
rs71627426 Chr.3: 75421981-75421981 on Build GRCh38
rs933153875 Chr.3: 75422102-75422111 on Build GRCh38
rs1053958780 Chr.3: 75422163-75422163 on Build GRCh38
rs997334762 Chr.3: 75421870-75421870 on Build GRCh38
rs748610946 Chr.3: 75421951-75421951 on Build GRCh38
rs563534815 Chr.3: 75422063-75422063 on Build GRCh38
rs532432247 Chr.3: 75422092-75422092 on Build GRCh38
rs888905503 Chr.3: 75421920-75421920 on Build GRCh38
rs76883378 Chr.3: 75421977-75421977 on Build GRCh38
rs956591467 Chr.3: 75421930-75421930 on Build GRCh38
rs11716994 Chr.3: 75422079-75422079 on Build GRCh38
rs184464852 Chr.3: 75422176-75422176 on Build GRCh38
rs1025259369 Chr.3: 75421961-75421961 on Build GRCh38
rs926716454 Chr.3: 75421752-75421752 on Build GRCh38
rs980974571 Chr.3: 75421991-75421991 on Build GRCh38
rs531359396 Chr.3: 75421954-75421954 on Build GRCh38
rs201238417 Chr.3: 75421844-75421844 on Build GRCh38
rs371991085 Chr.3: 75422060-75422060 on Build GRCh38
rs1050256450 Chr.3: 75422107-75422107 on Build GRCh38
rs980838802 Chr.3: 75421742-75421742 on Build GRCh38
rs892116712 Chr.3: 75422173-75422173 on Build GRCh38
rs908518339 Chr.3: 75422077-75422078 on Build GRCh38
rs375350160 Chr.3: 75421706-75421706 on Build GRCh38
rs940031266 Chr.3: 75422080-75422080 on Build GRCh38
rs778161125 Chr.3: 75422002-75422002 on Build GRCh38
rs931301406 Chr.3: 75421763-75421763 on Build GRCh38
rs879123146 Chr.3: 75422170-75422170 on Build GRCh38
rs565278656 Chr.3: 75421948-75421948 on Build GRCh38
rs573248594 Chr.3: 75421814-75421814 on Build GRCh38
rs923216428 Chr.3: 75422101-75422101 on Build GRCh38
rs563908279 Chr.3: 75421857-75421857 on Build GRCh38
rs190244632 Chr.3: 75421994-75421994 on Build GRCh38
rs538920825 Chr.3: 75422202-75422202 on Build GRCh38
rs887113006 Chr.3: 75421813-75421813 on Build GRCh38
rs1000933282 Chr.3: 75421921-75421921 on Build GRCh38
rs952790278 Chr.3: 75422020-75422020 on Build GRCh38
rs1028683144 Chr.3: 75421892-75421892 on Build GRCh38
rs184945375 Chr.3: 75421851-75421851 on Build GRCh38
rs1031973512 Chr.3: 75421927-75421927 on Build GRCh38
rs1048423106 Chr.3: 75421770-75421770 on Build GRCh38
rs199815626 Chr.3: 75422073-75422073 on Build GRCh38
rs569732890 Chr.3: 75422158-75422158 on Build GRCh38
rs11709789 Chr.3: 75422190-75422190 on Build GRCh38
rs536268198 Chr.3: 75422141-75422141 on Build GRCh38
rs1025227302 Chr.3: 75422182-75422182 on Build GRCh38
rs368132602 Chr.3: 75421714-75421714 on Build GRCh38
rs201431043 Chr.3: 75421887-75421887 on Build GRCh38

View Full Product Details