Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30370983-30371538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP586I1420
Entrez Gene ID
222161
Gene Name
uncharacterized protein DKFZp586I1420
Gene Aliases
-
UniGene
Hs.112423
Gene Chromosome Location
Chr.7: 30370050-30372809 on Build GRCh38
Transcripts
NR_002186.1

SNP Detail

SNP ID Chromosome Location
rs139599524 Chr.7: 30371488-30371488 on Build GRCh38
rs140038319 Chr.7: 30371029-30371029 on Build GRCh38
rs539992528 Chr.7: 30371167-30371167 on Build GRCh38
rs958105993 Chr.7: 30371055-30371055 on Build GRCh38
rs926032199 Chr.7: 30371477-30371477 on Build GRCh38
rs953599030 Chr.7: 30371066-30371066 on Build GRCh38
rs977571490 Chr.7: 30371454-30371454 on Build GRCh38
rs953648780 Chr.7: 30371370-30371370 on Build GRCh38
rs976999576 Chr.7: 30371110-30371110 on Build GRCh38
rs1046113671 Chr.7: 30371254-30371254 on Build GRCh38
rs1052351619 Chr.7: 30371280-30371280 on Build GRCh38
rs987446064 Chr.7: 30371106-30371106 on Build GRCh38
rs547061554 Chr.7: 30371057-30371057 on Build GRCh38
rs777210549 Chr.7: 30371317-30371317 on Build GRCh38
rs186512588 Chr.7: 30371291-30371291 on Build GRCh38
rs150598899 Chr.7: 30371432-30371432 on Build GRCh38
rs1004670543 Chr.7: 30371307-30371307 on Build GRCh38
rs184608846 Chr.7: 30371382-30371382 on Build GRCh38
rs1045973571 Chr.7: 30371455-30371455 on Build GRCh38
rs982440823 Chr.7: 30371144-30371144 on Build GRCh38
rs967460523 Chr.7: 30371139-30371139 on Build GRCh38
rs898878061 Chr.7: 30371335-30371335 on Build GRCh38
rs957442844 Chr.7: 30371478-30371478 on Build GRCh38
rs1029951406 Chr.7: 30371371-30371371 on Build GRCh38
rs570698105 Chr.7: 30371039-30371039 on Build GRCh38
rs779710555 Chr.7: 30371421-30371421 on Build GRCh38
rs572681142 Chr.7: 30371378-30371378 on Build GRCh38
rs967342621 Chr.7: 30371419-30371419 on Build GRCh38
rs142270870 Chr.7: 30371191-30371191 on Build GRCh38
rs1010921409 Chr.7: 30371304-30371304 on Build GRCh38
rs909510727 Chr.7: 30371412-30371412 on Build GRCh38
rs1018404682 Chr.7: 30371080-30371080 on Build GRCh38
rs906506058 Chr.7: 30371021-30371021 on Build GRCh38
rs951603398 Chr.7: 30371377-30371377 on Build GRCh38
rs999515068 Chr.7: 30371209-30371209 on Build GRCh38
rs542202717 Chr.7: 30371427-30371427 on Build GRCh38
rs42602 Chr.7: 30371349-30371349 on Build GRCh38
rs10267283 Chr.7: 30371169-30371169 on Build GRCh38
rs35842421 Chr.7: 30371178-30371178 on Build GRCh38
rs964663046 Chr.7: 30371108-30371108 on Build GRCh38
rs916490870 Chr.7: 30371198-30371198 on Build GRCh38
rs906178850 Chr.7: 30371258-30371258 on Build GRCh38
rs184904600 Chr.7: 30371112-30371112 on Build GRCh38
rs950022765 Chr.7: 30371180-30371180 on Build GRCh38
rs977462104 Chr.7: 30371162-30371162 on Build GRCh38
rs754601791 Chr.7: 30371513-30371513 on Build GRCh38
rs571930596 Chr.7: 30371317-30371320 on Build GRCh38
rs117545137 Chr.7: 30371510-30371510 on Build GRCh38
rs191353259 Chr.7: 30371362-30371362 on Build GRCh38
rs5883238 Chr.7: 30371410-30371410 on Build GRCh38
rs962473683 Chr.7: 30371413-30371413 on Build GRCh38
rs995140475 Chr.7: 30371036-30371036 on Build GRCh38
rs551807908 Chr.7: 30371217-30371217 on Build GRCh38
rs1036520951 Chr.7: 30371316-30371316 on Build GRCh38
rs923830818 Chr.7: 30371164-30371164 on Build GRCh38
rs947921641 Chr.7: 30371201-30371201 on Build GRCh38
rs778580003 Chr.7: 30371514-30371514 on Build GRCh38
rs1018931187 Chr.7: 30371418-30371418 on Build GRCh38

View Full Product Details