Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 35123545-35124136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P1
Entrez Gene ID
554236
Gene Name
DPY19L2 pseudogene 1
Gene Aliases
-
UniGene
Hs.594815
Gene Chromosome Location
Chr.7: 35081287-35186162 on Build GRCh38
Transcripts
NR_002833.2

SNP Detail

SNP ID Chromosome Location
rs1027034625 Chr.7: 35123764-35123764 on Build GRCh38
rs912829318 Chr.7: 35123799-35123799 on Build GRCh38
rs753780716 Chr.7: 35124102-35124102 on Build GRCh38
rs577209410 Chr.7: 35123758-35123758 on Build GRCh38
rs201663573 Chr.7: 35123617-35123617 on Build GRCh38
rs1023628966 Chr.7: 35123750-35123752 on Build GRCh38
rs766283920 Chr.7: 35124100-35124100 on Build GRCh38
rs1031371169 Chr.7: 35123694-35123694 on Build GRCh38
rs771539984 Chr.7: 35123973-35123973 on Build GRCh38
rs769865128 Chr.7: 35124108-35124108 on Build GRCh38
rs562437263 Chr.7: 35123764-35123765 on Build GRCh38
rs147124727 Chr.7: 35123906-35123906 on Build GRCh38
rs754794959 Chr.7: 35124111-35124111 on Build GRCh38
rs141869341 Chr.7: 35123897-35123897 on Build GRCh38
rs747818960 Chr.7: 35123956-35123956 on Build GRCh38
rs745591131 Chr.7: 35124087-35124088 on Build GRCh38
rs746327735 Chr.7: 35123994-35123994 on Build GRCh38
rs575579672 Chr.7: 35123690-35123690 on Build GRCh38
rs559888610 Chr.7: 35124072-35124072 on Build GRCh38
rs774568185 Chr.7: 35124041-35124041 on Build GRCh38
rs549394890 Chr.7: 35123840-35123841 on Build GRCh38
rs542409808 Chr.7: 35123919-35123919 on Build GRCh38
rs969350862 Chr.7: 35123750-35123750 on Build GRCh38
rs1043461112 Chr.7: 35123626-35123626 on Build GRCh38
rs574429323 Chr.7: 35123936-35123936 on Build GRCh38
rs760788474 Chr.7: 35124099-35124099 on Build GRCh38
rs540361227 Chr.7: 35123955-35123955 on Build GRCh38
rs539741765 Chr.7: 35123829-35123829 on Build GRCh38
rs562292070 Chr.7: 35123920-35123920 on Build GRCh38
rs1039997883 Chr.7: 35123576-35123576 on Build GRCh38
rs1013617846 Chr.7: 35123707-35123710 on Build GRCh38
rs975606998 Chr.7: 35123842-35123842 on Build GRCh38
rs770455133 Chr.7: 35123997-35123997 on Build GRCh38
rs778716214 Chr.7: 35123871-35123871 on Build GRCh38
rs569565736 Chr.7: 35123757-35123758 on Build GRCh38
rs2908117 Chr.7: 35123592-35123592 on Build GRCh38
rs761954783 Chr.7: 35124048-35124048 on Build GRCh38
rs758990783 Chr.7: 35123885-35123886 on Build GRCh38
rs750537282 Chr.7: 35124085-35124085 on Build GRCh38
rs780880437 Chr.7: 35124054-35124054 on Build GRCh38
rs763353250 Chr.7: 35124011-35124011 on Build GRCh38
rs767537398 Chr.7: 35124055-35124055 on Build GRCh38
rs777431496 Chr.7: 35123979-35123979 on Build GRCh38
rs921623001 Chr.7: 35123872-35123872 on Build GRCh38
rs982967799 Chr.7: 35123783-35123783 on Build GRCh38
rs903687779 Chr.7: 35123681-35123681 on Build GRCh38
rs2536282 Chr.7: 35123648-35123648 on Build GRCh38
rs191719410 Chr.7: 35123589-35123589 on Build GRCh38
rs776178545 Chr.7: 35124010-35124010 on Build GRCh38
rs900288103 Chr.7: 35123598-35123598 on Build GRCh38
rs768970204 Chr.7: 35124039-35124039 on Build GRCh38
rs754781621 Chr.7: 35123953-35123953 on Build GRCh38
rs931745015 Chr.7: 35123600-35123600 on Build GRCh38
rs2908118 Chr.7: 35123647-35123647 on Build GRCh38

View Full Product Details