Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22002973-22003439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA3A
Entrez Gene ID
10136
Gene Name
chymotrypsin like elastase family member 3A
Gene Aliases
ELA3, ELA3A
UniGene
Hs.181289
Gene Chromosome Location
Chr.1: 22001656-22012542 on Build GRCh38
Transcripts
NM_005747.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1960248 c.85G>A p.V29I Chr.1: 22003044-22003044 on Build GRCh38
5898708 c.122C>T p.P41L Chr.1: 22003081-22003081 on Build GRCh38
904384 c.44C>A p.A15D Chr.1: 22003003-22003003 on Build GRCh38
5468676 c.121T>C p.S41P Chr.1: 22003032-22003032 on Build GRCh38
5453068 c.139C>A p.H47N Chr.1: 22003050-22003050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959836422 Chr.1: 22003163-22003163 on Build GRCh38
rs368374089 Chr.1: 22003010-22003010 on Build GRCh38
rs41307874 Chr.1: 22003110-22003110 on Build GRCh38
rs766462894 Chr.1: 22002996-22002996 on Build GRCh38
rs928069851 Chr.1: 22003126-22003126 on Build GRCh38
rs767378718 Chr.1: 22003091-22003091 on Build GRCh38
rs942368403 Chr.1: 22003349-22003349 on Build GRCh38
rs778895690 Chr.1: 22003137-22003137 on Build GRCh38
rs536134870 Chr.1: 22003295-22003295 on Build GRCh38
rs749409675 Chr.1: 22003038-22003038 on Build GRCh38
rs1048823416 Chr.1: 22003307-22003307 on Build GRCh38
rs550611930 Chr.1: 22003063-22003063 on Build GRCh38
rs4381156 Chr.1: 22003194-22003194 on Build GRCh38
rs780794710 Chr.1: 22003015-22003015 on Build GRCh38
rs57573643 Chr.1: 22003385-22003385 on Build GRCh38
rs549681282 Chr.1: 22003084-22003084 on Build GRCh38
rs7519660 Chr.1: 22003050-22003050 on Build GRCh38
rs141652575 Chr.1: 22003068-22003068 on Build GRCh38
rs551108357 Chr.1: 22003332-22003332 on Build GRCh38
rs972600399 Chr.1: 22003176-22003176 on Build GRCh38
rs746312349 Chr.1: 22003043-22003043 on Build GRCh38
rs761421252 Chr.1: 22003116-22003116 on Build GRCh38
rs4543768 Chr.1: 22003152-22003152 on Build GRCh38
rs769383326 Chr.1: 22003054-22003054 on Build GRCh38
rs752481465 Chr.1: 22003109-22003109 on Build GRCh38
rs4543769 Chr.1: 22003333-22003333 on Build GRCh38
rs777947108 Chr.1: 22003037-22003037 on Build GRCh38
rs545148620 Chr.1: 22003041-22003041 on Build GRCh38
rs762622931 Chr.1: 22003064-22003064 on Build GRCh38
rs918458005 Chr.1: 22003243-22003243 on Build GRCh38
rs573687841 Chr.1: 22003383-22003383 on Build GRCh38
rs1023634936 Chr.1: 22002993-22002993 on Build GRCh38
rs41302021 Chr.1: 22003239-22003239 on Build GRCh38
rs528135718 Chr.1: 22003122-22003122 on Build GRCh38
rs564169007 Chr.1: 22003387-22003387 on Build GRCh38
rs201536611 Chr.1: 22003044-22003044 on Build GRCh38
rs1052620086 Chr.1: 22003418-22003418 on Build GRCh38
rs751556750 Chr.1: 22003081-22003081 on Build GRCh38
rs7533776 Chr.1: 22003032-22003032 on Build GRCh38
rs189505124 Chr.1: 22003371-22003371 on Build GRCh38
rs969396319 Chr.1: 22003067-22003067 on Build GRCh38
rs386629419 Chr.1: 22003030-22003032 on Build GRCh38
rs144477023 Chr.1: 22003014-22003014 on Build GRCh38
rs569056040 Chr.1: 22003260-22003260 on Build GRCh38
rs41300124 Chr.1: 22003188-22003188 on Build GRCh38
rs372304926 Chr.1: 22003121-22003121 on Build GRCh38
rs762533821 Chr.1: 22002995-22002995 on Build GRCh38
rs931241801 Chr.1: 22003258-22003258 on Build GRCh38
rs7531336 Chr.1: 22003030-22003030 on Build GRCh38

View Full Product Details