Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161820831-161821170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF6
Entrez Gene ID
22926
Gene Name
activating transcription factor 6
Gene Aliases
ACHM7, ATF6A
UniGene
Hs.492740
Gene Chromosome Location
Chr.1: 161766244-161964070 on Build GRCh38
Transcripts
NM_007348.3 XM_006711224.1 XM_011509308.1 XM_011509309.1 XM_011509310.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898617 c.1127G>A p.R376Q Chr.1: 161821101-161821101 on Build GRCh38
898616 c.1126C>T p.R376* Chr.1: 161821100-161821100 on Build GRCh38
4576500 c.1175A>G p.Y392C Chr.1: 161821149-161821149 on Build GRCh38
3705189 c.1168C>G p.L390V Chr.1: 161821142-161821142 on Build GRCh38
4576499 c.1118G>C p.S373T Chr.1: 161821092-161821092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765936076 Chr.1: 161821049-161821049 on Build GRCh38
rs750797724 Chr.1: 161821139-161821139 on Build GRCh38
rs925169089 Chr.1: 161820926-161820926 on Build GRCh38
rs780340461 Chr.1: 161821098-161821098 on Build GRCh38
rs867820530 Chr.1: 161821122-161821122 on Build GRCh38
rs888035149 Chr.1: 161820862-161820862 on Build GRCh38
rs752848119 Chr.1: 161820966-161820967 on Build GRCh38
rs747921362 Chr.1: 161821123-161821123 on Build GRCh38
rs762415950 Chr.1: 161821041-161821041 on Build GRCh38
rs548052568 Chr.1: 161820868-161820868 on Build GRCh38
rs878871603 Chr.1: 161820925-161820925 on Build GRCh38
rs201720200 Chr.1: 161820925-161820926 on Build GRCh38
rs775960355 Chr.1: 161821052-161821052 on Build GRCh38
rs757452190 Chr.1: 161821078-161821078 on Build GRCh38
rs754184728 Chr.1: 161821077-161821077 on Build GRCh38
rs774855413 Chr.1: 161820924-161820925 on Build GRCh38
rs764451128 Chr.1: 161821058-161821058 on Build GRCh38
rs552323571 Chr.1: 161821010-161821010 on Build GRCh38
rs758549801 Chr.1: 161821097-161821097 on Build GRCh38
rs969004560 Chr.1: 161820877-161820877 on Build GRCh38
rs913710719 Chr.1: 161820963-161820963 on Build GRCh38
rs1009190912 Chr.1: 161821007-161821007 on Build GRCh38
rs957608484 Chr.1: 161820917-161820917 on Build GRCh38
rs931933672 Chr.1: 161820980-161820980 on Build GRCh38
rs765354804 Chr.1: 161821081-161821081 on Build GRCh38
rs181456626 Chr.1: 161821028-161821028 on Build GRCh38
rs79406872 Chr.1: 161820939-161820940 on Build GRCh38
rs533520427 Chr.1: 161820982-161820982 on Build GRCh38
rs765524240 Chr.1: 161821092-161821092 on Build GRCh38
rs769666743 Chr.1: 161821136-161821136 on Build GRCh38
rs781003969 Chr.1: 161821119-161821119 on Build GRCh38
rs537882677 Chr.1: 161821040-161821040 on Build GRCh38
rs367613392 Chr.1: 161821082-161821082 on Build GRCh38
rs980408551 Chr.1: 161820891-161820891 on Build GRCh38
rs942229101 Chr.1: 161820872-161820872 on Build GRCh38
rs186397514 Chr.1: 161821053-161821053 on Build GRCh38
rs564681246 Chr.1: 161820976-161820976 on Build GRCh38
rs771128039 Chr.1: 161821072-161821072 on Build GRCh38
rs754973294 Chr.1: 161821101-161821101 on Build GRCh38
rs746887984 Chr.1: 161821100-161821100 on Build GRCh38
rs797045174 Chr.1: 161821084-161821085 on Build GRCh38
rs890836540 Chr.1: 161820983-161820983 on Build GRCh38
rs967951251 Chr.1: 161821011-161821011 on Build GRCh38
rs866111607 Chr.1: 161820940-161820940 on Build GRCh38
rs1033501312 Chr.1: 161821065-161821065 on Build GRCh38
rs988114987 Chr.1: 161820947-161820947 on Build GRCh38
rs887815344 Chr.1: 161820994-161820994 on Build GRCh38
rs1039239325 Chr.1: 161820914-161820914 on Build GRCh38

View Full Product Details