Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50948467-50948788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPM1
Entrez Gene ID
8813
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
Gene Aliases
CDGIE, MPDS
UniGene
Hs.654951
Gene Chromosome Location
Chr.20: 50934855-50958564 on Build GRCh38
Transcripts
NM_001317034.1 NM_001317035.1 NM_001317036.1 NM_003859.2 NR_133648.1 XM_011529093.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446710 c.266T>C p.L89P Chr.20: 50948658-50948658 on Build GRCh38
4099361 c.273A>G p.P91P Chr.20: 50948651-50948651 on Build GRCh38
6446711 c.264T>C p.L88L Chr.20: 50948658-50948658 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753337775 Chr.20: 50948706-50948706 on Build GRCh38
rs766462970 Chr.20: 50948651-50948651 on Build GRCh38
rs774244338 Chr.20: 50948668-50948668 on Build GRCh38
rs1041527850 Chr.20: 50948499-50948499 on Build GRCh38
rs757819188 Chr.20: 50948598-50948598 on Build GRCh38
rs763645722 Chr.20: 50948703-50948703 on Build GRCh38
rs749795581 Chr.20: 50948596-50948596 on Build GRCh38
rs746263556 Chr.20: 50948608-50948608 on Build GRCh38
rs1050370119 Chr.20: 50948521-50948521 on Build GRCh38
rs554958858 Chr.20: 50948523-50948523 on Build GRCh38
rs772233855 Chr.20: 50948609-50948609 on Build GRCh38
rs573286911 Chr.20: 50948571-50948571 on Build GRCh38
rs767218987 Chr.20: 50948675-50948675 on Build GRCh38
rs904366156 Chr.20: 50948537-50948537 on Build GRCh38
rs779225695 Chr.20: 50948602-50948602 on Build GRCh38
rs755788332 Chr.20: 50948701-50948701 on Build GRCh38
rs771823278 Chr.20: 50948709-50948711 on Build GRCh38
rs775797102 Chr.20: 50948622-50948622 on Build GRCh38
rs121908583 Chr.20: 50948650-50948650 on Build GRCh38
rs1008695624 Chr.20: 50948548-50948548 on Build GRCh38
rs1031495562 Chr.20: 50948600-50948600 on Build GRCh38
rs569940029 Chr.20: 50948500-50948500 on Build GRCh38
rs200920587 Chr.20: 50948583-50948583 on Build GRCh38
rs994461170 Chr.20: 50948729-50948729 on Build GRCh38
rs752397824 Chr.20: 50948692-50948692 on Build GRCh38
rs1025779156 Chr.20: 50948749-50948749 on Build GRCh38
rs570292112 Chr.20: 50948741-50948742 on Build GRCh38
rs768779546 Chr.20: 50948639-50948639 on Build GRCh38
rs758553918 Chr.20: 50948621-50948621 on Build GRCh38
rs536920409 Chr.20: 50948508-50948508 on Build GRCh38
rs780244460 Chr.20: 50948638-50948638 on Build GRCh38
rs759423847 Chr.20: 50948671-50948671 on Build GRCh38
rs202112706 Chr.20: 50948579-50948579 on Build GRCh38
rs906775483 Chr.20: 50948702-50948702 on Build GRCh38
rs935728066 Chr.20: 50948544-50948544 on Build GRCh38
rs751754266 Chr.20: 50948565-50948565 on Build GRCh38

View Full Product Details