Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63624028-63624334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2
Entrez Gene ID
283417
Gene Name
dpy-19 like 2
Gene Aliases
SPATA34, SPGF9
UniGene
Hs.533644
Gene Chromosome Location
Chr.12: 63558913-63669201 on Build GRCh38
Transcripts
NM_173812.4 XM_006719352.2 XM_011538215.2 XM_017019188.1 XM_017019189.1 XM_017019190.1 XM_017019191.1 XM_017019192.1 XM_017019193.1 XM_017019194.1 XM_017019195.1 XM_017019196.1 XM_017019197.1 XM_017019198.1 XM_017019199.1 XM_017019200.1 XM_017019201.1 XM_017019202.1 XM_017019203.1 XM_017019204.1 XM_017019205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244138 c.912T>C p.P304P Chr.12: 63624081-63624081 on Build GRCh38
942410 c.949C>A p.L317I Chr.12: 63624044-63624044 on Build GRCh38
3782757 c.869G>A p.R290H Chr.12: 63624124-63624124 on Build GRCh38
4147382 c.862G>T p.A288S Chr.12: 63624131-63624131 on Build GRCh38
1606487 c.871G>A p.V291M Chr.12: 63624122-63624122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918223214 Chr.12: 63624279-63624279 on Build GRCh38
rs747628376 Chr.12: 63624165-63624165 on Build GRCh38
rs587777206 Chr.12: 63624101-63624101 on Build GRCh38
rs756355358 Chr.12: 63624149-63624149 on Build GRCh38
rs7300764 Chr.12: 63624176-63624176 on Build GRCh38
rs777793491 Chr.12: 63624164-63624164 on Build GRCh38
rs774183925 Chr.12: 63624082-63624082 on Build GRCh38
rs542329575 Chr.12: 63624190-63624190 on Build GRCh38
rs146876165 Chr.12: 63624183-63624183 on Build GRCh38
rs917679038 Chr.12: 63624197-63624197 on Build GRCh38
rs371875973 Chr.12: 63624172-63624172 on Build GRCh38
rs773028091 Chr.12: 63624075-63624075 on Build GRCh38
rs750857060 Chr.12: 63624142-63624142 on Build GRCh38
rs770656278 Chr.12: 63624079-63624079 on Build GRCh38
rs546639994 Chr.12: 63624191-63624191 on Build GRCh38
rs760880725 Chr.12: 63624251-63624251 on Build GRCh38
rs139405673 Chr.12: 63624073-63624073 on Build GRCh38
rs972371972 Chr.12: 63624255-63624255 on Build GRCh38
rs754673809 Chr.12: 63624150-63624150 on Build GRCh38
rs764591745 Chr.12: 63624113-63624113 on Build GRCh38
rs560155512 Chr.12: 63624235-63624235 on Build GRCh38
rs573019515 Chr.12: 63624086-63624086 on Build GRCh38
rs769715208 Chr.12: 63624072-63624072 on Build GRCh38
rs780818533 Chr.12: 63624061-63624061 on Build GRCh38
rs781047908 Chr.12: 63624163-63624163 on Build GRCh38
rs529033071 Chr.12: 63624289-63624289 on Build GRCh38
rs145052038 Chr.12: 63624078-63624078 on Build GRCh38
rs970426032 Chr.12: 63624238-63624238 on Build GRCh38
rs199562120 Chr.12: 63624117-63624117 on Build GRCh38
rs752764341 Chr.12: 63624100-63624100 on Build GRCh38
rs147579680 Chr.12: 63624124-63624124 on Build GRCh38
rs760679938 Chr.12: 63624104-63624104 on Build GRCh38
rs754330251 Chr.12: 63624115-63624115 on Build GRCh38
rs759725841 Chr.12: 63624085-63624085 on Build GRCh38
rs931255858 Chr.12: 63624309-63624309 on Build GRCh38

View Full Product Details