Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50970415-50970545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2C
Entrez Gene ID
1031
Gene Name
cyclin dependent kinase inhibitor 2C
Gene Aliases
INK4C, p18, p18-INK4C
UniGene
Hs.525324
Gene Chromosome Location
Chr.1: 50968695-50974637 on Build GRCh38
Transcripts
NM_001262.2 NM_078626.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2192270 c.62_63delTT p.L21fs*2 Chr.1: 50970430-50970431 on Build GRCh38
2192272 c.109T>A p.F37I Chr.1: 50970477-50970477 on Build GRCh38
4619045 c.103A>G p.N35D Chr.1: 50970471-50970471 on Build GRCh38
5967629 c.76C>T p.Q26* Chr.1: 50970444-50970444 on Build GRCh38
4407874 c.64A>C p.T22P Chr.1: 50970432-50970432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371529586 Chr.1: 50970448-50970448 on Build GRCh38
rs555689863 Chr.1: 50970438-50970438 on Build GRCh38
rs61751018 Chr.1: 50970488-50970488 on Build GRCh38
rs869243400 Chr.1: 50970516-50970516 on Build GRCh38
rs781499255 Chr.1: 50970463-50970463 on Build GRCh38
rs753009206 Chr.1: 50970514-50970514 on Build GRCh38
rs200956660 Chr.1: 50970459-50970459 on Build GRCh38
rs954236531 Chr.1: 50970512-50970512 on Build GRCh38
rs150509670 Chr.1: 50970458-50970458 on Build GRCh38
rs149643969 Chr.1: 50970464-50970464 on Build GRCh38
rs869283778 Chr.1: 50970505-50970505 on Build GRCh38
rs372401911 Chr.1: 50970507-50970507 on Build GRCh38
rs758585248 Chr.1: 50970515-50970515 on Build GRCh38
rs375535785 Chr.1: 50970516-50970516 on Build GRCh38
rs753867120 Chr.1: 50970472-50970472 on Build GRCh38
rs766360495 Chr.1: 50970469-50970469 on Build GRCh38

View Full Product Details