Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102325214-102325364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC3
Entrez Gene ID
330
Gene Name
baculoviral IAP repeat containing 3
Gene Aliases
AIP1, API2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49, c-IAP2
UniGene
Hs.127799
Gene Chromosome Location
Chr.11: 102317450-102339403 on Build GRCh38
Transcripts
NM_001165.4 NM_182962.2 XM_017017643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328890 c.771C>A p.A257A Chr.11: 102325280-102325280 on Build GRCh38
1684943 c.710A>T p.N237I Chr.11: 102325219-102325219 on Build GRCh38
6936869 c.711T>G p.N237K Chr.11: 102325220-102325220 on Build GRCh38
73837 c.743C>G p.S248C Chr.11: 102325252-102325252 on Build GRCh38
6005782 c.754A>C p.M252L Chr.11: 102325263-102325263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2276113 Chr.11: 102325288-102325288 on Build GRCh38
rs370712639 Chr.11: 102325323-102325323 on Build GRCh38
rs760734130 Chr.11: 102325302-102325302 on Build GRCh38
rs755783862 Chr.11: 102325257-102325257 on Build GRCh38
rs747649672 Chr.11: 102325255-102325255 on Build GRCh38
rs373514370 Chr.11: 102325252-102325252 on Build GRCh38
rs750897944 Chr.11: 102325246-102325246 on Build GRCh38
rs765661781 Chr.11: 102325325-102325325 on Build GRCh38
rs147757750 Chr.11: 102325253-102325253 on Build GRCh38
rs924808678 Chr.11: 102325259-102325259 on Build GRCh38
rs755873313 Chr.11: 102325263-102325263 on Build GRCh38
rs201739570 Chr.11: 102325326-102325326 on Build GRCh38
rs771950433 Chr.11: 102325296-102325296 on Build GRCh38
rs777456061 Chr.11: 102325260-102325260 on Build GRCh38
rs571701272 Chr.11: 102325272-102325272 on Build GRCh38
rs967841484 Chr.11: 102325274-102325274 on Build GRCh38
rs201026275 Chr.11: 102325306-102325306 on Build GRCh38
rs747731925 Chr.11: 102325242-102325243 on Build GRCh38
rs377560709 Chr.11: 102325305-102325305 on Build GRCh38
rs753471138 Chr.11: 102325314-102325314 on Build GRCh38
rs181325614 Chr.11: 102325307-102325307 on Build GRCh38
rs774161644 Chr.11: 102325278-102325278 on Build GRCh38
rs770645813 Chr.11: 102325276-102325276 on Build GRCh38
rs775611041 Chr.11: 102325300-102325300 on Build GRCh38

View Full Product Details